Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 22

Accusense™ Pressure Transducer   to   Acetal Copolymer Hose Clamp

Image
  Product Detail
SKU

AccuSense™ Pressure Transducer, 0-50 PSIG

Category: Pressure Transmitters and Pressure Transducers
Description: AccuSense High-Accuracy Pressure Transducers
Brand: Setra
Model Number: AccuSense


AccuSense™ Pressure Transducer, 0-500 PSIG

Category: Pressure Transmitters and Pressure Transducers
Description: AccuSense High-Accuracy Pressure Transducers
Brand: Setra
Model Number: AccuSense


AccuSense™ Pressure Transducer, 0-750 PSIG

Category: Pressure Transmitters and Pressure Transducers
Description: AccuSense High-Accuracy Pressure Transducers
Brand: Setra
Model Number: AccuSense


AccuSense™ Pressure Transducer, 30-0" HG

Category: Pressure Transmitters and Pressure Transducers
Description: AccuSense High-Accuracy Pressure Transducers
Brand: Setra
Model Number: AccuSense


Acd-10:Pro Digital Clamp-On Multimeter (ACD)

Category: Amprobe Clamp Meters and Multimeters
Description: Amprobe 10 Series Digital Clamp-On Multimeters
Brand: Amprobe
Model Number: ACD


ACD-16 PRO: 1000A DMM Data Logging Clamp-On Multimeter (ACD)

Category: Clamp Meters
Description: Amprobe 1000A Data Logging Clamp-On Multimeters
Brand: Amprobe
Model Number: ACD


ACD-16 TRMS-PRO: 1000A DMM Data Logging Clamp-On Multimeter (ACD)

Category: Amprobe Clamp Meters and Multimeters
Description: Amprobe 1000A Data Logging Clamp-On Multimeters
Brand: Amprobe
Model Number: ACD


Acd-56-Hpq:Power Quality Clamp-On Meter (ACD)

Category: Clamp Meters
Description: Amprobe Power Quality Clamp Meter
Brand: AMPROBE
Model Number: ACD


ACD-6 PRO: 1000A Digital Clamp-On Multimeter (ACD)

Category: Amprobe Clamp Meters and Multimeters
Description: Amprobe 1000A Digital Clamp-On Meters
Brand: Amprobe
Model Number: ACD


ACD-6 TRMS PRO: 1000A Digital Clamp-On Meter (ACD)

Category: Amprobe Clamp Meters and Multimeters
Description: Amprobe 1000A Digital Clamp-On Meters
Brand: Amprobe
Model Number: ACD


ACDC400:Dig Ac/Dc Clamp-On Multimeter (ACDC)

Category: Amprobe Clamp Meters and Multimeters
Description: Amprobe AC-DC 400 Digital Clamp-On Multimeter
Brand: Amprobe
Model Number: ACDC


ACE GLASS Separatory Funnels, 1000 mL (YO-34731-08)

Category: Separatory Funnels
Description: ACE GLASS Separatory Funnels
Brand: Ace Glass


ACE GLASS Separatory Funnels, 125 mL (YO-34731-02)

Category: Separatory Funnels
Description: ACE GLASS Separatory Funnels
Brand: Ace Glass


ACE GLASS Separatory Funnels, 2000 mL (YO-34731-10)

Category: Separatory Funnels
Description: ACE GLASS Separatory Funnels
Brand: Ace Glass


ACE GLASS Separatory Funnels, 250 mL (YO-34731-04)

Category: Separatory Funnels
Description: ACE GLASS Separatory Funnels
Brand: Ace Glass


ACE GLASS Separatory Funnels, 500 mL (YO-34731-06)

Category: Separatory Funnels
Description: ACE GLASS Separatory Funnels
Brand: Ace Glass


ACE GLASS Separatory Funnels, 60 mL (YO-34731-00)

Category: Separatory Funnels
Description: ACE GLASS Separatory Funnels
Brand: Ace Glass


Ace Glass Tube Adapter with Hose Barb, 24/40 Joint (5261-16)

Category: Adapters
Description: Ace Glass Tube Adapter
Brand: Cole-Parmer
Model Number: 5261-16


Ace Glass Tube Adapter, 24/40 Joint (5028-30)

Category: Adapters
Description: Ace Glass Tube Adapter
Brand: Cole-Parmer
Model Number: 5028-30


Acessory Starter Kit for Evidence Drying Cabinets 28616-00 and -02. In

Category: Labconco Protector Evidence Drying Cabinet
Description: Accessories for Evidence Drying Cabinet
Brand: Labconco


ACET ACS SAF-CT 6X2.5L CS (YO-88051-49)

Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand: Cole-Parmer


Acetal Check Valve, 1/4" NPT(F) inlet/1/4" NPT(F) outlet (0354300)

Category: Check Valves
Description: Acetal Check Valves
Brand: Cole-Parmer
Model Number: 0354300


Acetal Check Valve, 1/4" NPT(F) inlet/1/4" NPT(M) outlet (0354303)

Category: Check Valves
Description: Acetal Check Valves
Brand: Cole-Parmer
Model Number: 0354303


Acetal Copolymer Hose Clamps Kit, White

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Acetal Copolymer Hose Clamps and Kits
Brand: Cole-Parmer


Acetal Copolymer Hose Clamps, Black, 0.228" x 0.256", 100/pack

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Acetal Copolymer Hose Clamps and Kits
Brand: Cole-Parmer


Acetal Copolymer Hose Clamps, White, 0.228" x 0.256", 100/pack

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Acetal Copolymer Hose Clamps and Kits
Brand: Cole-Parmer


Acetal Copolymer Hose Clamps, White, 0.269" x 0.291", 100/pack

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Acetal Copolymer Hose Clamps and Kits
Brand: Cole-Parmer


Acetal Copolymer Hose Clamps, White, 0.307" x 0.342", 100/pack

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Acetal Copolymer Hose Clamps and Kits
Brand: Cole-Parmer


Acetal Copolymer Hose Clamps, White, 0.351" x 0.394", 100/pack

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Acetal Copolymer Hose Clamps and Kits
Brand: Cole-Parmer


Acetal Copolymer Hose Clamps, White, 0.387" x 0.440", 100/pack

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Acetal Copolymer Hose Clamps and Kits
Brand: Cole-Parmer




Accusense™ Pressure Transducer to Acetal Copolymer Hose Clamp

>