Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 400

Cole-Parmer Penny Head Hollow Stopper #19   to   Cole-Parmer Ph Electrode

Image
  Product Detail
SKU

Cole-Parmer Penny Head Hollow Stopper #19

Category: Stoppers
Description: Cole-Parmer Penny Head Hollow Stoppers, PTFE
Brand: Cole-Parmer


Cole-Parmer Penny Head Hollow Stopper #9

Category: Stoppers
Description: Cole-Parmer Penny Head Hollow Stoppers, PTFE
Brand: Cole-Parmer


Cole-Parmer Personal Microcentrifuge; 115V; 60 Hz (YO-17307-00)

Category: Microcentrifuges
Description: Cole-Parmer Personal Microcentrifuge
Brand: Cole-Parmer


Cole-Parmer Personal Microcentrifuge; 230V; 50Hz (YO-17307-05)

Category: Microcentrifuges
Description: Cole-Parmer Personal Microcentrifuge
Brand: Cole-Parmer


Cole-Parmer pH 10 buffer solution, 1 L bottle, NIST-traceable referenc

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 10 buffer solution, 4 L bottle, NIST-traceable referenc

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 10 buffer solution, 500 mL bottle, NIST-traceable refer

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 10 buffer, blue, 1L, NIST-traceable

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 10 buffer, blue, 4L, NIST-traceable

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 10 buffer, blue, 500mL, NIST-traceable

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 4 buffer solution, 1 L bottle, NIST-traceable reference

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 4 buffer solution, 4 L bottle, NIST-traceable reference

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 4 buffer solution, 500 mL bottle, NIST-traceable refere

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 4 buffer, red, 1L, NIST-traceable

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 4 buffer, red, 4L, NIST-traceable

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 4 buffer, red, 500mL, NIST-traceable

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 7 buffer solution, 1 L bottle, NIST-traceable reference

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 7 buffer solution, 4 L bottle, NIST-traceable reference

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 7 buffer solution, 500 mL bottle, NIST-traceable refere

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 7 buffer, yellow, 1L, NIST-traceable

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 7 buffer, yellow, 4L, NIST-traceable

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH 7 buffer, yellow, 500mL, NIST-traceable

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer pH Electrode for Ultrapure Water

Category: Cole-Parmer Ultrapure Water pH Measurement System
Description: Cole-Parmer Ultrapure Water pH electrode
Brand: Cole-Parmer


Cole-Parmer pH electrode, epoxy-body, spear tip, double-junction, BNC

Category: Cole-Parmer Epoxy-Body Combination pH and ORP Electrodes
Description: Cole-Parmer Epoxy-Body Special-purpose Electrodes
Brand: Cole-Parmer


Cole-Parmer pH electrode, in-line, double-junction, 100 Ohm RTD w/mini (YO-05993-29)

Category: Cole-Parmer Economical In-Line pH and ORP Electrodes
Description: Cole-Parmer Economical In-Line pH Electrodes
Brand: Cole-Parmer


Cole-Parmer pH electrode, in-line, double-junction, 100 Ohm RTD w/mini (YO-27001-70)

Category: Cole-Parmer Economical In-Line pH and ORP Electrodes
Description: Cole-Parmer Economical In-Line pH Electrodes
Brand: Cole-Parmer


Cole-Parmer pH electrode, in-line, double-junction, 100 Ohm RTD w/tinn (YO-05993-38)

Category: Cole-Parmer Economical In-Line pH and ORP Electrodes
Description: Cole-Parmer Economical In-Line pH Electrodes
Brand: Cole-Parmer


Cole-Parmer pH electrode, in-line, double-junction, 100 Ohm RTD w/tinn (YO-27001-72)

Category: Cole-Parmer Economical In-Line pH and ORP Electrodes
Description: Cole-Parmer Economical In-Line pH Electrodes
Brand: Cole-Parmer


Cole-Parmer pH electrode, in-line, double-junction, 1000 Ohm RTD, 10-f (YO-05993-48)

Category: Cole-Parmer Economical In-Line pH and ORP Electrodes
Description: Cole-Parmer Economical In-Line pH Electrodes
Brand: Cole-Parmer


Cole-Parmer pH electrode, in-line, double-junction, 1000 Ohm RTD, 10-f (YO-27001-91)

Category: Cole-Parmer Economical In-Line pH and ORP Electrodes
Description: Cole-Parmer Economical In-Line pH Electrodes
Brand: Cole-Parmer




Cole-Parmer Penny Head Hollow Stopper #19 to Cole-Parmer Ph Electrode

>