Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 5
A&D Hlwp Portion Control Washdown Scale
to
a&D Moisture Determination Balance
Image
Product Detail
SKU
A&D HLWP Portion Control Washdown Scale, 300 G X 0.1 G
Category: A&D Weighing Food Portion Scales
Description: A&D Weighing HLWP Portable Washdown Scales
Brand:
A&D Weighing
Model Number:
HL-300WP
YO-11114-81
More Details
A&D HLWP Portion Control Washdown Scale, 3000 G X 1 G
Category: A&D Weighing Food Portion Scales
Description: A&D Weighing HLWP Portable Washdown Scales
Brand:
A&D Weighing
Model Number:
HL-3000WP
YO-11114-83
More Details
A&D HLWP Portion Control Washdown Scale, 3000 G X 1 G (large Weigh Pan
Category: A&D Weighing Food Portion Scales
Description: A&D Weighing HLWP Portable Washdown Scales
Brand:
A&D Weighing
Model Number:
HL-3000LWP
YO-11114-85
More Details
A&D HLWP Portion Control Washdown Scale, 3000 G X 1 G (ntep, With Larg
Category: A&D Weighing Food Portion Scales
Description: A&D Weighing HLWP Portable Washdown Scales
Brand:
A&D Weighing
Model Number:
HL-3000LWPN
YO-11114-86
More Details
A&D HLWP Portion Control Washdown Scale, 3000 G X 1 G (ntep)
Category: A&D Weighing Food Portion Scales
Description: A&D Weighing HLWP Portable Washdown Scales
Brand:
A&D Weighing
Model Number:
HL-3000WPN
YO-11114-84
More Details
A&D HT-Series Compact Scales 3100g x 1g incl carry case
Category: A&D Weighing Portable Balances
Description: A&D HT Portable Scales
Brand:
A&D Weighing
YO-11111-31
More Details
A&D HT-Series Compact Scales 310g x 0.1g incl carry case
Category: A&D Weighing Portable Balances
Description: A&D HT Portable Scales
Brand:
A&D Weighing
YO-11111-29
More Details
A&D HT-Series Compact Scales 5100g x 1g incl carry case
Category: A&D Weighing Portable Balances
Description: A&D HT Portable Scales
Brand:
A&D Weighing
YO-11111-32
More Details
A&D HT-Series Compact Scales 510g x 0.1g incl carry case
Category: A&D Weighing Portable Balances
Description: A&D HT Portable Scales
Brand:
A&D Weighing
YO-11111-30
More Details
A&D HT-Series Compact Scales Stainless Steel Weighing Pan Option
Category: A&D Weighing Portable Balances
Description: A&D HT Portable Scales
Brand:
A&D Weighing
YO-11111-33
More Details
A&D HW Industrial Scale, 10 kg x 0.001 kg
Category: A&D Weighing Industrial Scales
Description: A&D Weighing HW Industrial Scales
Brand:
A&D Weighing
Model Number:
HW-10KGL
YO-11125-00
More Details
A&D HW Industrial Scale, 100 kg x 0.01 kg (Triple Range)
Category: A&D Weighing Industrial Scales
Description: A&D Weighing HW Industrial Scales
Brand:
A&D Weighing
Model Number:
HW-100KGL
YO-11125-20
More Details
A&D HW Industrial Scale, 15/30/60 kg x 0.1/0.005/0.01 kg (Triple Range
Category: A&D Weighing Industrial Scales
Description: A&D Weighing HW Industrial Scales
Brand:
A&D Weighing
Model Number:
HV-60KGL
YO-11125-16
More Details
A&D HW Industrial Scale, 220 kg x 0.02 kg (Triple Range)
Category: A&D Weighing Industrial Scales
Description: A&D Weighing HW Industrial Scales
Brand:
A&D Weighing
Model Number:
HW-200KGL
YO-11125-30
More Details
A&D HW Industrial Scale, 3/6/15 kg x 0.001/0.002/0.005 kg (Triple Rang
Category: A&D Weighing Industrial Scales
Description: A&D Weighing HW Industrial Scales
Brand:
A&D Weighing
Model Number:
HV-15KGL
YO-11122-04
More Details
A&D HW Industrial Scale, 60 kg x 0.005 kg
Category: A&D Weighing Industrial Scales
Description: A&D Weighing HW Industrial Scales
Brand:
A&D Weighing
Model Number:
HW-60KGL
YO-11125-10
More Details
A&D HW Industrial Scale, 60/150/220 kg x 0.02/0.05/0.1 kg (Triple Rang
Category: A&D Weighing Industrial Scales
Description: A&D Weighing HW Industrial Scales
Brand:
A&D Weighing
Model Number:
HV-200KGL
YO-11122-10
More Details
A&D HW Washdown Industrial Scale, 3/6/15 kg x 0.001/0.002/0.005 g (Tri
Category: A&D Weighing Industrial Scales
Description: A&D Weighing HW Washdown Industrial Scales
Brand:
A&D Weighing
Model Number:
HV-15K-WP
YO-11122-16
More Details
A&D HW Washdown Industrial Scale, Capacity 132/300/500lbs.
Category: A&D Weighing Industrial Scales
Description: A&D Weighing HW Washdown Industrial Scales
Brand:
A&D Weighing
Model Number:
HV-200K-WP
YO-11122-24
More Details
A&D HW Washdown Industrial Scale, Capacity 150lb.
Category: A&D Weighing Industrial Scales
Description: A&D Weighing HW Washdown Industrial Scales
Brand:
A&D Weighing
Model Number:
HW-60K-WP
YO-11122-20
More Details
A&D HW Washdown Industrial Scale, Capacity 200lbs.
Category: A&D Weighing Industrial Scales
Description: A&D Weighing HW Washdown Industrial Scales
Brand:
A&D Weighing
Model Number:
HW-100K-WP
YO-11122-22
More Details
A&D HW Washdown Industrial Scale, Capacity 30/60/150lbs.
Category: A&D Weighing Industrial Scales
Description: A&D Weighing HW Washdown Industrial Scales
Brand:
A&D Weighing
Model Number:
HV-60K-WP
YO-11122-18
More Details
A&D ION Analytical Balance, 220g x 0.1,mg IntCal, Ionizer, Env,Sensors
Category: Semi/Micro Analytical Balances
Description: A&D Weighing ION Analytical, Semi-Micro, and Micro Balances
Brand:
A&D Weighing
Model Number:
BM-200
YO-11110-92
More Details
A&D ION Analytical Balance, 320g x 0.1mg IntCal, Ionizer, Env.Sensors
Category: Semi/Micro Analytical Balances
Description: A&D Weighing ION Analytical, Semi-Micro, and Micro Balances
Brand:
A&D Weighing
Model Number:
BM-300
YO-11110-93
More Details
A&D ION Analytical Balance, 520g x 0.1mg IntCal, Ionizer, Env.Sensors
Category: Semi/Micro Analytical Balances
Description: A&D Weighing ION Analytical, Semi-Micro, and Micro Balances
Brand:
A&D Weighing
Model Number:
BM-500
YO-11110-94
More Details
A&D ION Micro Balance, 22g x 0.001mg Int.Cal/Ionizer/Environmtl Sensor
Category: Semi/Micro Analytical Balances
Description: A&D Weighing ION Analytical, Semi-Micro, and Micro Balances
Brand:
A&D Weighing
Model Number:
BM-20
YO-11110-89
More Details
A&D ION Micro Balance, 5.1g/22g x 0.001mg/0.01mg IntCal/Ionizer/Env.Se
Category: Semi/Micro Analytical Balances
Description: A&D Weighing ION Analytical, Semi-Micro, and Micro Balances
Brand:
A&D Weighing
Model Number:
BM-22
YO-11110-90
More Details
A&D ION Semi-Micro Balance, 250g x 0.01mg IntCal, Ionizer, Env.Sensors
Category: Semi/Micro Analytical Balances
Description: A&D Weighing ION Analytical, Semi-Micro, and Micro Balances
Brand:
A&D Weighing
Model Number:
BM-252
YO-11110-91
More Details
A&D Moisture Determination Balance, MF-50, 51 g x 0.002 g, 115 VAC
Category: Moisture Balances
Description: A&D Weighing Moisture Balances
Brand:
A&D Weighing
Model Number:
MF-50
YO-01041-30
More Details
A&D Moisture Determination Balance, ML-50, 51 g x 0.005 g, 115 VAC
Category: Moisture Balances
Description: A&D Weighing Moisture Balances
Brand:
A&D Weighing
Model Number:
ML-50
YO-01041-25
More Details
64114
results
<<<Previous
1
2
3
4
5
6
7
8
9
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,138
A&D Hlwp Portion Control Washdown Scale to a&D Moisture Determination Balance
>
31125329-p-toc_2 31125329ptoc2