Adapters For 250mL Nalgene Oak Ridge Bottle (1 Position), Fits Bucket
Category: Thermo Scientific Sorvall ST 40 Centrifuges
Description: High Versatility Swinging Bucket Rotor, Buckets, and Adapters
Brand: Thermo Scientific Nalgene
Model Number: 75003710