Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 800
Glass Bead
to
Glass Fiber Filter
Image
Product Detail
SKU
Glass Beads, 0.1 to 0.15 mm (YO-36270-52)
Category: Mini Bead Beater
Description: Beads
Brand:
Cole-Parmer
YO-36270-52
More Details
Glass Beads, 0.5 mm (YO-36270-51)
Category: Mini Bead Beater
Description: Beads
Brand:
Cole-Parmer
YO-36270-51
More Details
Glass Beads, 1.0 mm (YO-36270-50)
Category: Mini Bead Beater
Description: Beads
Brand:
Cole-Parmer
YO-36270-50
More Details
Glass Body, Platinum Electrodes Conductivity Cell for OAKTON Conductiv
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Oakton
YO-35608-52
More Details
Glass capillary tubes, closed at both ends, pack of 100
Category: Melting Point
Description: Stuart Analog Melting Point Apparatus
Brand:
Cole-Parmer
YO-03013-65
More Details
Glass capillary tubes, open both ends, pack of 100
Category: Melting Point
Description: Stuart Analog Melting Point Apparatus
Brand:
Cole-Parmer
YO-03013-63
More Details
Glass capillary tubes, open one end, pack of 100
Category: Digital Melting Point Apparatuses
Description: Stuart Digital Melting Point Apparatus
Brand:
Cole-Parmer
YO-03013-64
More Details
Glass conductivity dip cell, K=0.1 (384592)
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Horiba
Model Number:
384592
YO-19600-60
More Details
Glass conductivity dip cell, K=1 (384591)
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Horiba
Model Number:
384591
YO-19600-61
More Details
Glass conductivity dip cell, K=10 (384590)
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Horiba
Model Number:
384590
YO-19600-62
More Details
Glass conductivity flow-through cell, K=0.1 (384589)
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Horiba
Model Number:
384589
YO-19600-70
More Details
Glass Coplin Staining Jar with glass cover (YO-48585-30)
Category: Staining Jars and Dishes for Histology
Description: Glass Coplin Staining Jar with Cover
Brand:
Cole-Parmer
YO-48585-30
More Details
Glass cuvettes with PTFE cover, square, 10-mm pathlength, 3.5 mL capac
Category: Glass Cuvettes
Description: Standard Paired Glass Cuvettes for Visible Spectroscopy
Brand:
PTFE
Model Number:
1-G
YO-83200-20
More Details
Glass cuvettes with PTFE stopper, square, 10-mm pathlength, 3.5 mL cap
Category: Glass Cuvettes
Description: Standard Paired Glass Cuvettes for Visible Spectroscopy
Brand:
PTFE
Model Number:
21-G
YO-83200-70
More Details
Glass Disposal Boxes, Bench Style, 6 per pack
Category: Biosafety Disposal
Description: Disposal Boxes
Brand:
Cole-Parmer
Model Number:
797015
YO-81402-00
More Details
Glass Disposal Boxes, Floor Style, 6 per pack
Category: Biosafety Disposal
Description: Disposal Boxes
Brand:
Cole-Parmer
Model Number:
797005
YO-81402-02
More Details
Glass Disposal Boxes, Intermediate Style, 6 per pack
Category: Biosafety Disposal
Description: Disposal Boxes
Brand:
Cole-Parmer
Model Number:
797035
YO-81402-01
More Details
Glass electrodes with BNC fixed cable (285129155(BLUE LINE#15PH))
Category: pH/Ion Laboratory Electrodes
Description: Schott BlueLine Electrodes
Brand:
Schott
Model Number:
285129155(BLUE LINE#15PH)
YO-55503-34
More Details
Glass electrodes with BNC fixed cable (285129171(BLUE LINE#17PH))
Category: pH/Ion Laboratory Electrodes
Description: Schott BlueLine Electrodes
Brand:
Schott
Model Number:
285129171(BLUE LINE#17PH)
YO-55503-46
More Details
Glass electrodes with removable cable
Category: pH/Ion Laboratory Electrodes
Description: Schott BlueLine Electrodes
Brand:
Schott
Model Number:
285129114(BLUE LINE#11PH)
YO-55503-00
More Details
Glass Encased magnetic Stir Bars, 1 1/2"l X 3/8"d (F37101-0112)
Category: Stir Bars
Description: Glass-Encased Stir Bars
Brand:
Cole-Parmer
Model Number:
F37101-0112
YO-04769-53
More Details
Glass Encased magnetic Stir Bars, 1"l X 3/8"d (F37101-0001)
Category: Stir Bars
Description: Glass-Encased Stir Bars
Brand:
Cole-Parmer
Model Number:
F37101-0001
YO-04769-52
More Details
Glass Encased magnetic Stir Bars, 1/2"l X 3/8"d (F37101-0012)
Category: Stir Bars
Description: Glass-Encased Stir Bars
Brand:
Cole-Parmer
Model Number:
F37101-0012
YO-04769-50
More Details
Glass Encased magnetic Stir Bars, 7/8"l X 1/4"d (F37101-7814)
Category: Stir Bars
Description: Glass-Encased Stir Bars
Brand:
Cole-Parmer
Model Number:
F37101-7814
YO-04769-51
More Details
Glass Extension 2", Kg47 (311450)
Category: Holders
Description: Glass and Stainless Steel Microanalysis Holders
Brand:
Advantec
Model Number:
311450
YO-06645-57
More Details
Glass fiber filter, DP70, 110-mm dia; box of 50
Category: Filter Papers
Description: Advantec Glass and Quartz Fiber Filters
Brand:
Advantec
Model Number:
DP70110MM
YO-06658-05
More Details
Glass fiber filter, DP70, 55-mm dia; box of 50
Category: Filter Papers
Description: Advantec Glass and Quartz Fiber Filters
Brand:
Advantec
Model Number:
DP7055MM
YO-06658-03
More Details
Glass fiber filter, GA100, 110-mm dia; box of 100
Category: Filter Papers
Description: Advantec Glass and Quartz Fiber Filters
Brand:
Advantec
Model Number:
GA100110MM
YO-06657-55
More Details
Glass fiber filter, GA100, 42.5-mm dia; box of 100
Category: Filter Papers
Description: Advantec Glass and Quartz Fiber Filters
Brand:
Advantec
Model Number:
GA10042.5MM
YO-06657-51
More Details
Glass fiber filter, GA100, 55-mm dia; box of 100
Category: Filter Papers
Description: Advantec Glass and Quartz Fiber Filters
Brand:
Advantec
Model Number:
GA10055MM
YO-06657-53
More Details
64114
results
<<<Previous
796
797
798
799
800
801
802
803
804
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
100
200
300
400
500
600
700
710
720
730
740
750
760
770
780
785
786
787
788
789
790
791
792
793
794
795
796
797
798
799
800
801
802
803
804
805
806
807
808
809
810
811
812
813
814
820
830
840
850
860
870
880
890
900
1,000
1,100
1,200
1,300
1,400
1,500
1,600
1,700
2,000
2,138
Glass Bead to Glass Fiber Filter
>
31125329-p-toc_2 31125329ptoc2