Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 9
A760
to
Abb Commander Pr250 Strip Chart Recorder
Image
Product Detail
SKU
A760 : Replacement CO Sensor (YO-10104-13)
Category: Air Sampling and Gas Analysis
Description: Air Sampling and Gas Analysis Accessories
Brand:
Davis
YO-10104-13
More Details
A761 : Replacement O2 Sensor (YO-10104-14)
Category: Air Sampling and Gas Analysis
Description: Air Sampling and Gas Analysis Accessories
Brand:
Davis
YO-10104-14
More Details
A766 : AC Charger (YO-10104-16)
Category: Air Sampling and Gas Analysis
Description: Air Sampling and Gas Analysis Accessories
Brand:
Davis
YO-10104-16
More Details
A772 : PC Software (YO-10104-17)
Category: Air Sampling and Gas Analysis
Description: Air Sampling and Gas Analysis Accessories
Brand:
Davis
YO-10104-17
More Details
A773 : Sulfer Filter For Oil (YO-10104-18)
Category: Air Sampling and Gas Analysis
Description: Air Sampling and Gas Analysis Accessories
Brand:
Davis
YO-10104-18
More Details
A774 : Silicone Tubing, 6FT. (YO-42500-30)
Description: General Accessories
Brand:
TPI
YO-42500-30
More Details
A776 : Static Tip (YO-42500-31)
Description: General Accessories
Brand:
TPI
YO-42500-31
More Details
A784 : Flow Regulator for Model 715 (YO-10104-19)
Category: Air Sampling and Gas Analysis
Description: Air Sampling and Gas Analysis Accessories
Brand:
Davis
YO-10104-19
More Details
A791:Acc Kit Washers/ Fittings/Adapter (A791)
Category: Dickson Weather Resistant Pressure Recorders
Description: Accessory Kit
Brand:
Cole-Parmer
Model Number:
A791
YO-08410-80
More Details
A799 : Filter for Hi-Efficiency Boilers (YO-10104-20)
Category: Air Sampling and Gas Analysis
Description: Air Sampling and Gas Analysis Accessories
Brand:
Davis
YO-10104-20
More Details
A900529-00014 : Hart Test Lead Kit (YO-16104-10)
Category: General Accessories
Description: Davis General Accessories
YO-16104-10
More Details
Aalborg Stepping Motor Proportioning Valve (aluminum), 1000 sL/min (ai
Category: Aalborg Stepping Motor Proportioning Valve
Description: Aalborg Stepping Motor Proportioning Valves (aluminum)
Brand:
Aalborg
Model Number:
SMV40-A
YO-98651-08
More Details
Aalborg Stepping Motor Proportioning Valve (aluminum), 200 sL/min (air
Category: Aalborg Stepping Motor Proportioning Valve
Description: Aalborg Stepping Motor Proportioning Valves (aluminum)
Brand:
Aalborg
Model Number:
SMV20-A
YO-98651-00
More Details
Aalborg Stepping Motor Proportioning Valve (aluminum), 500 sL/min (air
Category: Aalborg Stepping Motor Proportioning Valve
Description: Aalborg Stepping Motor Proportioning Valves (aluminum)
Brand:
Aalborg
Model Number:
SMV30-A
YO-98651-04
More Details
Aalborg Stepping Motor Proportioning Valve (stainless steel), 1000 sL/
Category: Aalborg Stepping Motor Proportioning Valve
Description: Aalborg Stepping Motor Proportioning Valves (stainless steel)
Brand:
Aalborg
Model Number:
SMV40-S
YO-98651-10
More Details
Aalborg Stepping Motor Proportioning Valve (stainless steel), 200 sL/m
Category: Aalborg Stepping Motor Proportioning Valve
Description: Aalborg Stepping Motor Proportioning Valves (stainless steel)
Brand:
Aalborg
Model Number:
SMV20-S
YO-98651-02
More Details
Aalborg Stepping Motor Proportioning Valve (stainless steel), 500 sL/m
Category: Aalborg Stepping Motor Proportioning Valve
Description: Aalborg Stepping Motor Proportioning Valves (stainless steel)
Brand:
Aalborg
Model Number:
SMV30-S
YO-98651-06
More Details
ABB Commander 10" Circular Chart Recorder, 2 pens
Category: Circular Chart Recorders
Description: ABB Commander Circular Chart Recorder/Controllers
Brand:
ABB Commander
Model Number:
C1912JA001100000STD
YO-80704-10
More Details
ABB Commander 10" Circular Chart Recorder, 3 pens
Category: Circular Chart Recorders
Description: ABB Commander Circular Chart Recorder/Controllers
Brand:
ABB Commander
Model Number:
C1913JA001100000STD
YO-80704-20
More Details
ABB Commander 10" Circular Chart Recorder, 4 pens
Category: Circular Chart Recorders
Description: ABB Commander Circular Chart Recorder/Controllers
Brand:
ABB Commander
Model Number:
C1914JA001100000STD
YO-80704-30
More Details
ABB Commander Circular Chart Recorder/Controller - 1 controller, 2 pen
Category: Circular Chart Recorders
Description: ABB Commander Circular Chart Recorders
Brand:
ABB Commander
Model Number:
C1912RA001100000STD
YO-80705-10
More Details
ABB Commander Circular Chart Recorder/Controller - 1 controller, 3 pen
Category: Circular Chart Recorders
Description: ABB Commander Circular Chart Recorders
Brand:
ABB Commander
Model Number:
C1913RA001100000STD
YO-80705-20
More Details
ABB Commander Circular Chart Recorder/Controller - 1 controller, 4 pen
Category: Circular Chart Recorders
Description: ABB Commander Circular Chart Recorders
Brand:
ABB Commander
Model Number:
C1914RA001100000STD
YO-80705-30
More Details
ABB Commander Circular Chart Recorder/Controller - 2 controllers, 2 pe
Category: Circular Chart Recorders
Description: ABB Commander Circular Chart Recorders
Brand:
ABB Commander
Model Number:
C1922RA001100000STD
YO-80705-40
More Details
ABB Commander Circular Chart Recorder/Controller - 2 controllers, 3 pe
Category: Circular Chart Recorders
Description: ABB Commander Circular Chart Recorders
Brand:
ABB Commander
Model Number:
C1923RA001100000STD
YO-80705-50
More Details
ABB Commander Circular Chart Recorder/Controller - 2 controllers, 4 pe
Category: Circular Chart Recorders
Description: ABB Commander Circular Chart Recorders
Brand:
ABB Commander
Model Number:
C1924RA001100000STD
YO-80705-60
More Details
ABB Commander MR250 Strip Chart Recorder, 12 traces
Category: Strip Chart Recorder
Description: ABB Commander 250-mm Strip Chart Recorders
Brand:
ABB Commander
Model Number:
SR250B125BOO000111ST
YO-80702-30
More Details
ABB Commander MR250 Strip Chart Recorder, 3 traces
Category: Strip Chart Recorder
Description: ABB Commander 250-mm Strip Chart Recorders
Brand:
ABB Commander
Model Number:
SR250B03SB-1
YO-80702-00
More Details
ABB Commander MR250 Strip Chart Recorder, 6 traces
Category: Strip Chart Recorder
Description: ABB Commander 250-mm Strip Chart Recorders
Brand:
ABB Commander
Model Number:
SR250B065B00000112ST
YO-80702-10
More Details
ABB Commander PR250 Strip Chart Recorder, 6 traces
Category: Strip Chart Recorder
Description: ABB Commander 250-mm Strip Chart Recorders
Brand:
ABB Commander
Model Number:
SR106AB000000011ST
YO-80703-10
More Details
64114
results
<<<Previous
5
6
7
8
9
10
11
12
13
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,138
A760 to Abb Commander Pr250 Strip Chart Recorder
>
31125329-p-toc_2 31125329ptoc2