Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 90
Allegro One Man Full Faceshield Supplied Air System with 50 Foot Hose
to
Alnor Rva501
Image
Product Detail
SKU
Allegro One Man Full Faceshield Supplied Air System with 50 foot Hose
Category: Supplied Air Systems
Description: Low-Pressure Full Faceshield Supplied-Air Respirator Kits
Brand:
Cole-Parmer
YO-81520-30
More Details
Allegro One Man Tyvek Hood Supplied Air System with 100 foot Hose
Category: Supplied Air Systems
Description: Low-Pressure Tyvek-Hood Supplied-Air Respirator Kits
Brand:
Allegro
YO-81520-15
More Details
Allegro One Man Tyvek Hood Supplied Air System with 50 foot Hose
Category: Supplied Air Systems
Description: Low-Pressure Tyvek-Hood Supplied-Air Respirator Kits
Brand:
Allegro
YO-81520-13
More Details
Allegro Two Man Full Face Piece Supplied Air System with 100 foot Hose
Category: Supplied Air Systems
Description: Low-Pressure Full-Facepiece Supplied-Air Respirator Kits
Brand:
Allegro
YO-81520-09
More Details
Allegro Two Man Full Face Piece Supplied Air System with 50 foot Hose
Category: Supplied Air Systems
Description: Low-Pressure Full-Facepiece Supplied-Air Respirator Kits
Brand:
Allegro
YO-81520-07
More Details
Allegro Two Man Full Faceshield Supplied Air System with 50 foot Hose
Category: Supplied Air Systems
Description: Low-Pressure Full Faceshield Supplied-Air Respirator Kits
Brand:
Cole-Parmer
YO-81520-31
More Details
Allegro Two Man Tyvek Hood Supplied Air System with 100 Foot Hose
Category: Supplied Air Systems
Description: Low-Pressure Tyvek-Hood Supplied-Air Respirator Kits
Brand:
Allegro
YO-81520-19
More Details
Allegro Two Man Tyvek Hood Supplied Air System with 50 foot Hose
Category: Supplied Air Systems
Description: Low-Pressure Tyvek-Hood Supplied-Air Respirator Kits
Brand:
Allegro
YO-81520-17
More Details
Allen Wrench (301721)
Category: Pressure Filtration Holders
Description: Stainless steel holders with reservoir
Model Number:
301721
YO-29902-22
More Details
Alligator Clip RTD Probe (08117-89)
Category: RTD Probes and Accessories
Description: Oakton Special-purpose probes
Brand:
Oakton
Model Number:
08117-89
YO-08117-89
More Details
Alligator Clips
Category: Fluke Digital Multimeters
Description: Fluke Test Leads, Probes, and Clips
Brand:
Fluke
Model Number:
AC72
YO-20007-83
More Details
Allihn condenser (YO-34510-03)
Category: Analytical Glassware
Description: Accelerated One-Step Modular Extractor/Concentrator
Brand:
One-Step
YO-34510-03
More Details
Allsafe MonoGoggle XTR™, Blue Frame, Clear Lens
Category: Safety Goggles
Description: Allsafe/SMC MonoGoggle XTR Goggles
Brand:
Allsafe
Model Number:
3010334
YO-81657-15
More Details
Allsafe MonoGoggle, Large, Clear Lens
Category: Safety Goggles
Description: Allsafe SMC MonoGoggle Goggles
Brand:
Allsafe
Model Number:
10195
YO-81657-11
More Details
Allsafe MonoGoggle, Regular, Clear Lens
Category: Safety Goggles
Description: Allsafe SMC MonoGoggle Goggles
Brand:
Allsafe
Model Number:
10115
YO-81657-00
More Details
Allsafe Shield/MonoGoggle XTR™, Blue Frame, Clear Lens
Category: Safety Goggles
Description: Allsafe/SMC MonoGoggle XTR Goggles
Brand:
Allsafe
Model Number:
3010343
YO-81657-18
More Details
Allsafe SMC MonoShield™ Face Shield
Category: Face Shields
Description: Allsafe/SMC MonoShield Shield
Brand:
Allsafe
Model Number:
MTL (10179)
YO-81657-30
More Details
Allsafe SMC™ Nemesis™ Safety Glasses, Black frame, Clear lens
Category: Allsafe SMC Safety Glasses
Description: Allsafe SMC Nemesis Safety Glasses
Brand:
Allsafe
Model Number:
19804
YO-86475-10
More Details
Allsafe SMC™ Nemesis™ Safety Glasses, Black frame, I/O lens
Category: Allsafe SMC Safety Glasses
Description: Allsafe SMC Nemesis Safety Glasses
Brand:
Allsafe
Model Number:
19807
YO-86475-12
More Details
ALLYL ACETATE 99% 5ML (YO-88193-58)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88193-58
More Details
ALLYL BROMIDE 99% STAB 500ML (YO-88089-44)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88089-44
More Details
ALLYL ISOTHIOCYANATE 94% 5G (YO-88089-52)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88089-52
More Details
ALLYLDIBUTYLTIN CHLORIDE 250MG (YO-88274-10)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88274-10
More Details
ALLYLDIPHENYLPHOSPHINE 5G (YO-88291-79)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88291-79
More Details
ALLYLPENTAFLUOROBENZENE 97 5G (YO-88285-14)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88285-14
More Details
Alnor Digital Manometer
Category: Airflow Capture Hoods
Description: TSI/Alnor
®
Electronic Balancing Tools
Brand:
Alnor
Model Number:
EBT720
YO-05949-10
More Details
Alnor Digital Manometer EBT730
Category: Airflow Capture Hoods
Description: TSI/Alnor Electronic Balancing Tools
Brand:
Alnor
Model Number:
EBT720
YO-05949-49
More Details
Alnor Low Flow Balometer Capture Hood
Category: Airflow Capture Hoods
Description: Alnor Low Flow Balometer Capture Hood
Brand:
Alnor
Model Number:
G200D
YO-10550-22
More Details
Alnor mechanical air velocity meter, 0-1600 ft/min
Category: Mechanical Air Velocity Meters
Description: Alnor Air Velocity Meters
Brand:
Alnor
Model Number:
8100A-16
YO-05949-70
More Details
Alnor RVA501 : Data Logging Vane Anemometer Hand Held
Category: Vane Anemometers
Description: Alnor Rotating Vane Anemometers
Brand:
Alnor
Model Number:
RVA501
YO-10508-74
More Details
64114
results
<<<Previous
86
87
88
89
90
91
92
93
94
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
10
20
30
40
50
60
70
75
76
77
78
79
80
81
82
83
84
85
86
87
88
89
90
91
92
93
94
95
96
97
98
99
100
101
102
103
104
110
120
130
140
150
160
170
180
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Allegro One Man Full Faceshield Supplied Air System with 50 Foot Hose to Alnor Rva501
>
31125329-p-toc_2 31125329ptoc2