Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 10
4' Purifier Logic+ B2 Biosafety Cabinet
to
Krohne Optimass Coriolis Mass Meter
Image
Product Detail
SKU
4' Purifier Logic+ B2 Biosafety Cabinet. Energy efficient ECM motor. I (YO-33517-67)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic+ Class II Biosafety Cabinets
Brand:
Labconco
YO-33517-67
More Details
4' Purifier Logic+ B2 Biosafety Cabinet. Energy efficient ECM motor. I (YO-33517-71)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic+ Class II Biosafety Cabinets
Brand:
Labconco
YO-33517-71
More Details
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-01)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco UxF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-01
More Details
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-02)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco UxF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-02
More Details
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-03)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco UxF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-03
More Details
Tektronix DPO5054 500 MHz, 5 GS/s, 12.5M record length, 4-channel digi
Category: Benchtop Oscilloscopes
Description: Tektronix MSO5000 and DPO5000 Mixed Signal Phosphor Oscilloscopes
Brand:
Tektronix
Model Number:
DPO5054
YO-20041-21
More Details
Labconco XPert NANO Enclosures, 6-ft wide; 100/115 VOLT 60 HZ; 30 (YO-33512-45)
Category: Balance Enclosures
Description: Labconco XPert Nano Enclosures
Brand:
Labconco
YO-33512-45
More Details
Purifier Cell Logic Biosafety Cabinet; 6 ft wide; Temperature Zone w (YO-33516-31)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Cell Logic Class II Biosafety Cabinets
Brand:
Labconco
YO-33516-31
More Details
Horizontal Tube Furnace, 42-1/8" Tube Length, 1500°C, 208 / 240 V (RHTC 80-710/15/P330)
Category: Tube Furnaces
Description: Nabertherm Compact Tube Furnaces with Silicon Carbide Rod Heating
Brand:
Nabertherm
Model Number:
RHTC 80-710/15/P330
YO-33334-36
More Details
TDS5054B: 4-Channel, 500 MHz, up to 5 GS/s, 16M Record Length (TDS5054B)
Category: Oscilloscopes
Description: Tektronix TDS5000B Series Oscilloscopes
Brand:
Tektronix
Model Number:
TDS5054B
YO-20014-49
More Details
Fluke TIR4/FT-20 Thermal Imager with IR-Fusion for the Building Indus (FLK-TIR4/FT-20)
Category: Building Industry Thermal Imagers
Description: Fluke Tir2, Tir3, Tir4 Advanced FlexCam Industrial Thermal Imagers
Brand:
Fluke
Model Number:
FLK-TIR4/FT-20
YO-39715-36
More Details
Temperature Chamber 100 L, (-20 C To 120°C) 100L; 220V 60 HZ
Category: Environmental Chambers
Description: Temperature and Environmental Chambers
Brand:
Lab Companion
Model Number:
AAA86031
YO-52401-82
More Details
Temperature Chamber 100 L, (-20 C To 120°C) 100L; 230V 50 HZ
Category: Environmental Chambers
Description: Temperature and Environmental Chambers
Brand:
Lab Companion
Model Number:
AAH86032
YO-52401-85
More Details
14-1/2 ft, seven cabinet island assembly with end sink and fixtures (YO-41526-10)
Category: Fume Hoods
Description: Hamilton Lab Bench Island Assemblies
Brand:
Thermo Scientific
YO-41526-10
More Details
Refrigerator, Blood Bank 80 cu ft with Casters, 3 Glass Doors, 115V/20 (NSBR803WWG/8)
Category: Nor-Lake Scientific Refrigerators
Description: Nor-Lake Blood Bank Refrigerators
Brand:
Nor-Lake
Model Number:
NSBR803WWG/8
YO-44173-32
More Details
Flaskscrubber Vantage Series Glassware Washer With Window And Interior (4540031)
Category: Labconco Laboratory Glassware Washers & Accessories
Description: Labconco FlaskScrubber Vantage Glassware Washer
Brand:
FlaskScrubber
Model Number:
4540031
YO-98964-50
More Details
Purifier Cell Logic Biosafety Cabinet; 5 ft wide; Scope-Ready and Te (YO-33516-08)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Cell Logic Class II Biosafety Cabinets
Brand:
Labconco
YO-33516-08
More Details
FreeZone -105° C 4.5 Liter Benchtop Freeze Dry System, 230V 50Hz CHI
Category: FreeZone Freeze Dry Systems
Description: Labconco FreeZone -105°C 4.5-L Freeze Dry System
Brand:
FreeZone
YO-05953-93
More Details
FreeZone -105° C 4.5 Liter Benchtop Freeze Dry System, 230V 50Hz NA
Category: FreeZone Freeze Dry Systems
Description: Labconco FreeZone -105°C 4.5-L Freeze Dry System
Brand:
FreeZone
YO-05953-89
More Details
FreeZone -105° C 4.5 Liter Benchtop Freeze Dry System, 230V 50Hz SHUK
Category: FreeZone Freeze Dry Systems
Description: Labconco FreeZone -105°C 4.5-L Freeze Dry System
Brand:
FreeZone
YO-05953-91
More Details
Class II B2 4-ft Biosafety Cabinet with 8" sash opening and UV light, (YO-33515-35)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic Class II Biosafety Cabinets
Brand:
Labconco
YO-33515-35
More Details
Labconco Protector XL Floor-mounted Hoods, 6 ft, 30" deep
Category: Ducted Fume Hoods
Description: Labconco Protector XL Floor-Mounted Hoods
Brand:
Labconco
YO-09098-70
More Details
6 ft Purifier® Cell Logic®+, Class II A2, 115V, Scope Ready with Stand
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-13
More Details
6 ft Purifier® Cell Logic®+, Class II A2, 230V (NA), Scope Ready with
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-15
More Details
5-ft Purifier® Cell Logic®+, Class II A2, 115V, Scope and Temp Zone wi
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-41
More Details
5-ft Purifier® Cell Logic®+, Class II A2, 230V (NA), Scope and Temp Zo (YO-33518-43)
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-43
More Details
Fluke TIR3/FT-10/20 Thermal Imager with IR-Fusion for the Building Ind (FLK-TIR3/FT-10/20)
Category: Building Industry Thermal Imagers
Description: Fluke Tir2, Tir3, Tir4 Advanced FlexCam Industrial Thermal Imagers
Brand:
Fluke
Model Number:
FLK-TIR3/FT-10/20
YO-39715-33
More Details
Temperature And Humidity Chamber 20 L, (0 C To 120°C) 20L; 220V 60
Category: Environmental Chambers
Description: Temperature and Environmental Chambers
Brand:
Lab Companion
Model Number:
AAH82211
YO-52401-86
More Details
Temperature And Humidity Chamber 20 L, (0 C To 120°C) 20L; 230V 50
Category: Environmental Chambers
Description: Temperature and Environmental Chambers
Brand:
Lab Companion
Model Number:
AAH82212
YO-52401-89
More Details
Krohne Optimass Coriolis mass Meter, 3" 220V (CP VE27 VE52)
Category: Coriolis Flowmeters
Description: KROHNE Optimass Coriolis Mass Meters
Brand:
Krohne
Model Number:
CP VE27 VE52
YO-33169-38
More Details
64114
results
<<<Previous
6
7
8
9
10
11
12
13
14
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,138
4' Purifier Logic+ B2 Biosafety Cabinet to Krohne Optimass Coriolis Mass Meter
>
31125329-p-toc_2 31125329ptoc2