Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 14
Keithley 2636b Sourcemeter Smu
to
Additel 761-M-N Automated Pressure Calibrator
Image
Product Detail
SKU
Keithley 2636B Sourcemeter SMU, 2-Channel, 0.1 fA, 200V, 1.5A DC/10 A
Category: SourceMeters
Description: Keithley 2600B Series SourceMeters
Brand:
Keithley
Model Number:
2636B
YO-20045-46
More Details
4-ft Purifier® Cell Logic®+, Class II A2, 230V (NA), Scope and Temp Zo (YO-33518-39)
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-39
More Details
4-ft Purifier® Cell Logic®+, Class II A2, 115V, Scope and Temp Zone wi
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-37
More Details
Purifier Cell Logic Biosafety Cabinet; 4 ft wide; Scope-Ready and Te (YO-33516-20)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Cell Logic Class II Biosafety Cabinets
Brand:
Labconco
YO-33516-20
More Details
5 ft Purifier® Cell Logic®+, Class II A2, 115V, Scope Ready with Stand
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-09
More Details
5 ft Purifier® Cell Logic®+, Class II A2, 230V (NA), Scope Ready with
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-11
More Details
Thermo Scientific APEX 100 Metal Detector, with 700 mm width x 350 mm
Category: Metal Detectors
Description: Thermo Scientific APEX 100 Metal Detectors
Brand:
Thermo Scientific
Model Number:
APEX 100 150 x 100
YO-20040-03
More Details
Nor-Lake Scientific -86°C Select Ultra-Low Upright Freezer, 23.2 c (YO-44174-72)
Category: Nor-Lake Scientific Freezers
Description: Nor-Lake Scientific -86°C Select Ultra-Low Temperature Freezers
Brand:
Nor-Lake
YO-44174-72
More Details
Nor-Lake Scientific -86°C Select Ultra-Low Upright Freezer, 23.2 c (YO-44174-74)
Category: Nor-Lake Scientific Freezers
Description: Nor-Lake Scientific -86°C Select Ultra-Low Temperature Freezers
Brand:
Nor-Lake
YO-44174-74
More Details
Nor-Lake Scientific -86°C Select Ultra-Low Upright Freezer, 23.2 c (YO-44174-76)
Category: Nor-Lake Scientific Freezers
Description: Nor-Lake Scientific -86°C Select Ultra-Low Temperature Freezers
Brand:
Nor-Lake
YO-44174-76
More Details
Tektronix MSO4034B, 350MHz, 2.5 GS/s, 20M Record Length, 4 + 16 Channe
Category: Benchtop Oscilloscopes
Description: Tektronix MSO4000B Mixed Signal Phosphor Oscilloscopes
Brand:
Tektronix
Model Number:
MSO4034B
YO-20040-73
More Details
Fluke 1750 Three Phase Power Quality Recorder
Category: Power Meters/Analyzers
Description: Fluke 1750 Series Power Analyzer
Brand:
Fluke
Model Number:
FLUKE-1750
YO-26046-51
More Details
Nor-Lake Scientific Select™ Pass-thru Refrigerator +4C, Four door 2
Category: General-Purpose Laboratory Refrigerators
Description: Nor-Lake Scientific Select Pass-Thru Laboratory Refrigerators
Brand:
Nor-Lake
YO-44174-73
More Details
A&D ION Micro Balance, 22g x 0.001mg Int.Cal/Ionizer/Environmtl Sensor
Category: Semi/Micro Analytical Balances
Description: A&D Weighing ION Analytical, Semi-Micro, and Micro Balances
Brand:
A&D Weighing
Model Number:
BM-20
YO-11110-89
More Details
Purifier Cell Logic Biosafety Cabinet; 5 ft wide; Scope-Ready; 230 V
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Cell Logic Class II Biosafety Cabinets
Brand:
Labconco
YO-33516-25
More Details
8-ft XStream Hood, 115V, without fixtures (YO-09099-30)
Category: Ducted Fume Hoods
Description: Labconco
®
Protector
®
XStream
™
Laboratory Hoods
Brand:
Labconco
YO-09099-30
More Details
8-ft XStream Hood, 230V, without fixtures (YO-09099-34)
Category: Ducted Fume Hoods
Description: Labconco
®
Protector
®
XStream
™
Laboratory Hoods
Brand:
Labconco
YO-09099-34
More Details
Horizontal Tube Furnace, 24-3/4" Tube Length, 1500°C, 208 / 240 V (RHTC 80-450/15/P330)
Category: Tube Furnaces
Description: Nabertherm Compact Tube Furnaces with Silicon Carbide Rod Heating
Brand:
Nabertherm
Model Number:
RHTC 80-450/15/P330
YO-33334-35
More Details
Cole-ParmerTissue Embedding Station 220 VAC 50/60Hz
Category: Tissue Embedding Products and Equipment
Description: Cole-Parmer Tissue Embedding Station
Brand:
Cole-Parmer
YO-48577-09
More Details
Cole-ParmerTissue Embedding Station 120 VAC 50/60Hz
Category: Tissue Embedding Products and Equipment
Description: Cole-Parmer Tissue Embedding Station
Brand:
Cole-Parmer
YO-48577-07
More Details
5-ft Xpert Filtered Balance System With Guardian Airflow Monitor, 230V (YO-09105-65)
Category: Balance Enclosures
Description: Labconco Filtered Balance Systems
Brand:
Labconco
YO-09105-65
More Details
Thermo Scientific MaxQ 8000 Stackable Incubated Shaker, 120V
Category: Thermo Scientific Forma Stackable Shaker System
Description: Thermo Scientific Forma Stackable Incubating/Refrigerating Shakers
Brand:
Thermo Scientific Forma
YO-51709-00
More Details
Thermo Scientific MaxQ 8000 Stackable Incubated Shaker, 230V
Category: Thermo Scientific Forma Stackable Shaker System
Description: Thermo Scientific Forma Stackable Incubating/Refrigerating Shakers
Brand:
Thermo Scientific Forma
YO-51709-02
More Details
Atago Programmable Digital Refractometer, ±0.01% Brix accuracy
Category: Digital Benchtop Refractometers
Description: ATAGO Programmable Digital Refractometer
Brand:
Atago
YO-81100-36
More Details
Standing Height Wall Assembly with Fume Hood - 16ft (YO-41524-65)
Category: Workstations
Description: Thermo Scientific Hamilton Laboratory Workbench Assemblies
Brand:
Thermo Scientific
YO-41524-65
More Details
Complete 6foot Fume Hood System With Acid And Solvent Storage Cabinet (YO-41525-20)
Category: Hamilton Constant Volume Bypass Fume Hoods
Description: Hamilton Constant Volume Benchtop Fume Hoods Assemblies
Brand:
Thermo Scientific Hamilton
YO-41525-20
More Details
5-ft Purifier® Cell Logic®+, Class II A2, 115V, Temp-Zone/UV with Stan
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-25
More Details
5-ft Purifier® Cell Logic®+, Class II A2, 230V (NA), Temp-Zone/UV with
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-27
More Details
Labconco Protector XL Floor-mounted Hoods, 5 ft, 30" deep
Category: Ducted Fume Hoods
Description: Labconco Protector XL Floor-Mounted Hoods
Brand:
Labconco
YO-09098-60
More Details
Additel 761-M-N Automated Pressure Calibrator
Category: Pressure Calibrators
Description: Additel 761 Automated Pressure Calibrators
Brand:
ADDITEL
Model Number:
761-M-N
YO-16101-12
More Details
64114
results
<<<Previous
10
11
12
13
14
15
16
17
18
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Keithley 2636b Sourcemeter Smu to Additel 761-M-N Automated Pressure Calibrator
>
31125329-p-toc_2 31125329ptoc2