Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 16
Heraeus Multifuge X3 R 120v Carbon Fiber 8x50 Bundle
to
Hydraulic Press Test System
Image
Product Detail
SKU
Heraeus Multifuge X3 R 120V Carbon Fiber 8x50 Bundle
Category: Thermo Scientifc Heraeus Centrifuges
Description: Thermo Scientific Heraeus Promotion
Brand:
Thermo Scientific
YO-17710-47
More Details
TBS SHUR/Cut™ 3500 Semi-automatic retracting microtome, 100-120VAC,
Category: TBS SHUR/Cut Microtomes
Description: TBS SHUR/Cu Microtome Series
Brand:
TBS
YO-48577-03
More Details
5 ft Purifier® Cell Logic®+, Class II A2, 115V, Scope Ready
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-08
More Details
5 ft Purifier® Cell Logic®+, Class II A2, 230V (NA), Scope Ready
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-10
More Details
5-ft Xpert Filtered Balance System, 230V 50hz (YO-09105-55)
Category: Balance Enclosures
Description: Labconco Filtered Balance Systems
Brand:
Labconco
YO-09105-55
More Details
Standing Height Wall Assembly with Fume Hood - 13ft (YO-41524-50)
Category: Workstations
Description: Thermo Scientific Hamilton Laboratory Workbench Assemblies
Brand:
Thermo Scientific
YO-41524-50
More Details
Tektronix PA4000 Multi-phase Power Analyzer, Two Input Channel
Category: Power Meters/Analyzers
Description: Tektronix PA4000 Series Power Analyzers
Brand:
Tektronix
YO-20045-51
More Details
Thermo Scientific Barnstead LabTower TII 40 l/hr with 100 L tank
Category: Water Purification
Description: Thermo Scientific Barnstead LabTower TII Water Purification Systems
Brand:
Barnstead
YO-99290-54
More Details
Class II A2 6-ft Biosafety Cabinet with 8" sash opening and UV light, (YO-33515-84)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic Class II Biosafety Cabinets
Brand:
Labconco
YO-33515-84
More Details
6-ft Xpert Filtered Balance System With Digital Airflow Monitor, 230V (YO-09106-50)
Category: Balance Enclosures
Description: Labconco Filtered Balance Systems
Brand:
Labconco
YO-09106-50
More Details
Rheometer system- low viscosity range (362-0052)
Category: Viscometers
Description: Rheometer System
Brand:
Thermo Scientific
Model Number:
362-0052
YO-98941-00
More Details
Tektronix DPO4104B-L, 1 GHz, 5/5/2.5 GS/s on 1,2 and 4 channels, 5M re
Category: Benchtop Oscilloscopes
Description: Tektronix® MSO4000B Mixed Signal and DPO4000B Digital Phosphor Oscilloscopes
Brand:
Tektronix
Model Number:
DPO4104B-L
YO-20043-43
More Details
Thermo Scientific APEX 100 Metal Detector, with 550 mm width x 350 mm
Category: Metal Detectors
Description: Thermo Scientific APEX 100 Metal Detectors
Brand:
Thermo Scientific
Model Number:
APEX 100 150 x 100
YO-20039-79
More Details
Complete 8foot Fume Hood System With Acid And Base Storage Cabinet Epo (YO-41525-30)
Category: Hamilton Constant Volume Bypass Fume Hoods
Description: Hamilton Constant Volume Benchtop Fume Hoods Assemblies
Brand:
Thermo Scientific Hamilton
YO-41525-30
More Details
Thermo Scientific Barnstead LabTower TII UV 20 l/hr with 100 L tank
Category: Water Purification
Description: Thermo Scientific Barnstead LabTower TII Water Purification Systems
Brand:
Barnstead
YO-99290-62
More Details
5-ft Purifier® Cell Logic®+, Class II A2, 115V, Temp-Zone/UV
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-24
More Details
5-ft Purifier® Cell Logic®+, Class II A2, 230V (NA), Temp-Zone/UV
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-26
More Details
Paramount Filtered Enclosure 6ft Wide 32in High 230 Volt; Requires 8 F (YO-28614-50)
Category: Labconco Paramount Filtered Enclosures and Accessories
Description: Labconco Paramount Ductless Enclosures
Brand:
Labconco
YO-28614-50
More Details
Microannular Gear Pump, 1.2 microliter to 72ml/min, 435 PSI with digit
Category: Gear Pump Systems/Skids
Description: Micropump Micro-Annular Gear Pumps
Brand:
Micropump
Model Number:
L24698
YO-73100-52
More Details
Thermo Scientific APEX 100 Metal Detector, with 700 mm width x 300 mm
Category: Metal Detectors
Description: Thermo Scientific APEX 100 Metal Detectors
Brand:
Thermo Scientific
Model Number:
APEX 100 150 x 100
YO-20040-02
More Details
5-ft Xpert Filtered Balance System, 115V 60hz (YO-09105-25)
Category: Balance Enclosures
Description: Labconco Filtered Balance Systems
Brand:
Labconco
YO-09105-25
More Details
Koehler Manual Vacuum Distillation System, 115 VAC, 60 Hz
Category: Petroleum Testing
Description: Koehler Manual Vacuum Distillation System
Brand:
Koehler
YO-59876-80
More Details
Koehler Manual Vacuum Distillation System, 220-240 VAC, 50/60 Hz
Category: Petroleum Testing
Description: Koehler Manual Vacuum Distillation System
Brand:
Koehler
YO-59876-81
More Details
Meiji Epi-Fluorescent Microscope, Trinocular
Category: Specialty Microscopes
Description: Meiji Epi-Fluorescent Microscopes
Brand:
Meiji
YO-48924-65
More Details
72" Explosion-Proof Bypass Fume Hood with Blower, 115 VAC/60 Hz (YO-09054-16)
Category: Ducted Fume Hoods
Description: Labconco
®
Protector
®
Premier
®
Fiberglass Fume Hoods
Brand:
Labconco
YO-09054-16
More Details
Paramount Filtered Enclosure 6ft Wide 32in High 115 Volt; Requires 8 F (YO-28613-50)
Category: Labconco Paramount Filtered Enclosures and Accessories
Description: Labconco Paramount Ductless Enclosures
Brand:
Labconco
YO-28613-50
More Details
Tektronix MSO4102B-L, 1 GHz, 5/2.5 GS/s on 1 and 2 channels, 5M record
Category: Benchtop Oscilloscopes
Description: Tektronix MSO4000B Mixed Signal Phosphor Oscilloscopes
Brand:
Tektronix
Model Number:
MSO4102B-L
YO-20043-45
More Details
6-ft Xpert Filtered Balance System With Digital Airflow Monitor, 115V (YO-09106-20)
Category: Balance Enclosures
Description: Labconco Filtered Balance Systems
Brand:
Labconco
YO-09106-20
More Details
Labconco Protector XL Floor-mounted Hoods, 4 ft, 36" deep
Category: Ducted Fume Hoods
Description: Labconco Protector XL Floor-Mounted Hoods
Brand:
Labconco
YO-09098-52
More Details
Hydraulic Press Test System, Automatic, 115 VAC, 60 Hz
Category: Presses
Description: Hydraulic Press Test Systems, Automatic
Brand:
Cole-Parmer
YO-59620-30
More Details
64114
results
<<<Previous
12
13
14
15
16
17
18
19
20
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Heraeus Multifuge X3 R 120v Carbon Fiber 8x50 Bundle to Hydraulic Press Test System
>
31125329-p-toc_2 31125329ptoc2