Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 200

Wrist-Action Shaker   to   Sanitary 3a Centrifugal Pump

Image
  Product Detail
SKU

Wrist-Action Shaker, 8 Flask, 230VAC, 50Hz

Category: Hand Motion Shakers
Description: Wrist-Action Shakers
Brand: Burrell
Model Number: 075-775-08-36


Series Q single head, variable-speed AC drive with controller, 1800 sp (QV-0710432)

Category: Series Q Valveless Piston Pump Systems
Description: Single-Head, Variable-Speed AC Drives with Separate Speed Controller
Brand: Cole-Parmer
Model Number: QV-0710432


Ismatec™ Ecoline pump with Easy-Load head, 1 channel, 0.23 to 1600

Category: Ismatec Ecoline Pumps
Description: Single-Channel Digital Pumps
Brand: Ismatec


Thermo Scientific Orion Versa Star with pH/Conductivity Modules Kit

Category: Benchtop Multiparameter meters
Description: Thermo Scientific Orion VERSA STAR Advanced Meter
Brand: Thermo Scientific Orion
Model Number: VERSA STAR W pH/Conductivity Modules Kit


Tub assembly, non-sealed, 5 L (50035008)

Category: Laboratory Fluid Bed Dryers
Description: Non-Sealed Tub Assemblies
Brand: Cole-Parmer
Model Number: 50035008


Sanitary 3A Centrifugal Pump, 148 GPM, 3 HP, White Epoxy Motor, 3600 R

Category: Specialty Pumps - Sanitary / Petroleum / Utility
Description: Sanitary Centrifugal Pumps
Brand: Cole-Parmer


Sartorius Extend ED Toploading Balance, 420 g x 1 mg, External Calibra

Category: Sartorius Toploading Balances
Description: Sartorius Extend ED Toploading Balances
Brand: Sartorius
Model Number: ED423S


Masterflex L/S Digital Pump System with Easy-Load 3 Pump Head, 600 RPM

Category: L/S Digital Pump Systems
Description: L/S Standard Digital Pump Systems
Brand: Masterflex


Ismatec™ compact modular drive with remote control, 110 VAC, 50/60

Category: Gear Pump Drives - For Micropump and Other Pump Heads
Description: Ismatec Compact Modular Drive and Controller for Micropump A Mount Pump Heads
Brand: Ismatec
Model Number: ISM896+MI0065+MI0181


AC With Condensate Evaporator 37.4" X 15.7" X 10.2", 10amps, 230V (YO-30002-18)

Category: Wall Mount Enclosures, Cabinets, & Racks
Description: Enclosure Air Conditioners
Brand: Rittal


Burkert Type 3234 SS Diaphragm Valve, 3-Way, 1" x 1" x 1" Tri-Clamp Co

Category: Diaphragm Valves
Description: Burkert Type 3234 Stainless Steel Diaphragm Valves
Brand: Burkert


Heated Evaporator/Concentrator; single block for 96-well plates (YO-28690-30)

Category: Evaporative Sample Concentrator
Description: Cole-Parmer Evaporator/Concentrators
Brand: Cole-Parmer


Cole-Parmer Dual Syringe Pump, Infuse / Withdraw Programmable, Touchsc

Category: ColeParmer Touch Screen Syringe Pumps
Description: Cole-Parmer Entry-Level Touch Screen Syringe Pumps
Brand: Cole-Parmer


Stainless Steel Polarimeter Tube, temperature-controllable with temper (YO-81250-45)

Category: Polarimeters
Description: Accessories for Kruss Automatic Polarimeter
Brand: Cole-Parmer


Scienceware Automatic Extra-Large Desiccator Cabinet, 3.6 cu. ft.

Category: Non Vacuum Desiccators
Description: Scienceware Extra-large Desiccators
Brand: Scienceware
Model Number: F43020-1010


Thermo Scientific Immersion Circulator, AC 200 thermostat with bridge (AC 200) (YO-12135-40)

Category: Immersion Circulators
Description: Thermo Scientific Heating Immersion Circulators
Brand: Thermo Scientific
Model Number: AC 200


Stainless Steel Polarimeter Tube, temperature-controllable with temper (YO-81250-46)

Category: Polarimeters
Description: Accessories for Kruss Automatic Polarimeter
Brand: Cole-Parmer


Sartorius Signum Industrial Bench Scale, 7kg X 0.1 G (6 Button Keypad)

Category: Sartorius Industrial Scales
Description: Sartorius Signum Industrial Bench Scales
Brand: Sartorius
Model Number: SIWADCP-V3


Cole-Parmer Gear Pump System, Benchtop Digital Drive, 0.261 mL/rev, 11

Category: Gear Pump Systems/Skids
Description: Cole-Parmer Benchtop Digital Gear Pump Drive Systems
Brand: Cole-Parmer


Thermocouple,Wire,TYPE-K,20-GAUGE, Ceramic Fiber Braid Insulation, 100 (K20-2-350) (YO-08542-23)

Category: Wire - Bulk / Spooled Thermocouple Wire
Description: 20 Gauge Thermocouple Wire
Brand: Cole-Parmer
Model Number: K20-2-350


Lower Spindle Rack For Labconco Washers (4595700)

Category: Labconco Laboratory Glassware Washers & Accessories
Description: Labconco Glassware Washer Accessories
Brand: Labconco
Model Number: 4595700


Current Probe for TDS3000B Series Oscilloscopes (50 MHz, BW 15 Amp Max (TCP202)

Category: Tektronix Oscilloscopes
Description: Tektronix Oscilloscope Probes
Brand: Tektr
Model Number: TCP202


Optional Shaker platform for 14575-16; 120V

Category: Laboratory & Utility Water Baths
Description: StableTemp Digital Utility Water Bath Accessories
Brand: Cole-Parmer


Knick Conductivity 4-wire explosion-proof controller

Category: Multiparameter Control
Description: Explosion-Proof Stratos Pro Series Conductivity Analyzers
Brand: STRATOS
Model Number: CONDUCTIVITY


Scienceware Lab Companion UV Blocking Vacuum Desiccator Cabinet, 45L.

Category: Vacuum Desiccators
Description: Scienceware Lab Companion Cabinet Vacuum Desiccators
Brand: Scienceware


Knick pH/ORP 4-wire explosion-proof controller

Category: Multiparameter Control
Description: Explosion-Proof Stratos Pro Series pH Analyzers
Brand: KNICK
Model Number: STRATOS pH/ORP


Solar-Powered Feed Verification Systems, 22.8 GPD, PVDF, 94 to 264 VAC

Category: Solenoid/Electronic Diaphragm Pumps
Description: Solar-Powered Feed Verification Systems
Brand: Cole-Parmer
Model Number: 73803-50+73803-90


Air-operated Diaphragm Pump, FDA-compliant, 45 Max GPM, 1-1/2" Tri-cla

Category: Air-Powered Double-Diaphragm Pumps
Description: Air-Operated Double Diaphragm Pumps
Brand: Warren Rupp
Model Number: T1FB1SASWTS000


Summit Series Analytical Balance, 60 g x 0.1 mg, 230 VAC, with Interna (SI-64.3)

Category: Denver Instrument® Analytical Balances
Description: Denver Instrument Summit Series Analytical Balances
Brand: Denver Instrument
Model Number: SI-64.3


Sanitary 3A Centrifugal Pump, 62 GPM, 0.5 HP, Stainless Steel Motor, 1 (YO-76720-00)

Category: Specialty Pumps - Sanitary / Petroleum / Utility
Description: Sanitary Centrifugal Pumps
Brand: Cole-Parmer




Wrist-Action Shaker to Sanitary 3a Centrifugal Pump

>