Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 2000
Ansell Touch-N-Tuff™ Nitrile Glove
to
Quick-Disconnect Panel-Mount Hose Barb Fitting Body
Image
Product Detail
SKU
Ansell Touch-N-Tuff™ Nitrile Gloves, Powdered, Medium
Category: Disposable Nitrile Gloves
Description: Ansell Touch-N-Tuff Nitrile Gloves
Brand:
Touch-N-Tuff
Model Number:
92-500 585189
YO-81602-50
More Details
Swab, cotton, non sterile, wooden shaft, 6"L x 1/12" dia (YO-14001-52)
Category: Essentials and Accessories for Microbial Analysis
Description: General-Purpose Sampling Swabs
Brand:
Cole-Parmer
YO-14001-52
More Details
RS-232 cable, 9 pin type for Doran Portion Control Scales 11911-00, 11
Category: Balances / Scales Accessories
Description: Doran Scales Accessories
Brand:
Doran
YO-11911-48
More Details
Extension shaft for tachometers (YO-08203-72)
Category: General Accessories
Description: Replacement Accessories
Brand:
Cole-Parmer
YO-08203-72
More Details
Bausch & Lomb Binocular Magnifier , Lens only, 2.6x Magnification; foc
Category: General Accessories
Description: Replacement Accessories
Brand:
Bausch & Lomb
Model Number:
81-43-06
YO-03886-21
More Details
Ball Valve, 2-way, 1/4" NPT(F), PP with Buna N seals (YO-01378-83)
Category: Small Plastic Ball Valves
Description: Polypropylene Plastic Ball Valves, Two-way (straight)
Brand:
Cole-Parmer
YO-01378-83
More Details
Polypropylene conical graduate, 500 mL (212225)
Category: Plastic Graduated Cylinders
Description: Conical Graduates, PP
Brand:
Cole-Parmer
Model Number:
212225
YO-06135-35
More Details
Quick-disconnect fittings, Straight-through hose barbs , PP,3/8 (HFC17812)
Category: Hose Barbs
Description: Straight-Through Quick-Disconnect Hose Barb Bodies
Brand:
Colder
Model Number:
HFC17812
YO-31303-16
More Details
Quick-disconnect fittings, Standard John Guest valved fittings Delrin (PLCD29006)
Category: Standard John Guest Inserts
Description: Valved body
Brand:
Delrin
Model Number:
PLCD29006
YO-31304-32
More Details
Compression male pipe adapter elbow, 1/8" x 1/8", Nylon, 10 Pack (YO-06382-47)
Category: Male Threaded Adapter Elbows
Description: Nylon Compression to NPT(M) Adapter Elbows
Brand:
Cole-Parmer
YO-06382-47
More Details
Ball Valve, 2-way, 1/4" NPT(F) x 1/4" NPT(M), PP with Viton® seals
Category: Small Plastic Ball Valves
Description: Polypropylene Plastic Ball Valves, Two-way (straight)
Brand:
Guaranteed In Stock Program
YO-01377-28
More Details
Ansell Neoprene Apron, 35" x 45", black
Category: Aprons
Description: Ansell Neoprene Aprons
Brand:
Ansell
YO-86479-01
More Details
Ball Valve, 2-way, 1/4" NPT(F) x 1/4" NPT(M), PP with EPDM seals
Category: Small Plastic Ball Valves
Description: Polypropylene Plastic Ball Valves, Two-way (straight)
Brand:
Cole-Parmer
YO-01378-86
More Details
Uvex STEALTH Safety Goggles, Clear Lens
Category: Safety Goggles
Description: Uvex STEALTH Safety Goggles
Brand:
Uvex
Model Number:
S3960C
YO-86326-00
More Details
NFPA Label, 2" X 2"
Category: Warning Labels and Stickers
Description: NFPA Right-To-Know Hazard Communication Labels
Brand:
Cole-Parmer
Model Number:
HMS12L
YO-86396-03
More Details
Masterflex TEE-Connector, polypropylene, I/P 89, 10/pack
Category: Masterflex Barbed Fittings
Description: Masterflex Tee connectors
Brand:
Masterflex
YO-30613-67
More Details
Reusable Particulate Respirator, Small (YO-86458-00)
Category: Particulate Respirators
Description: North CFR-1
™
Reusable Particulate Respirators
Brand:
North
YO-86458-00
More Details
Barbed fittings, Reducing Connector, Kynar,5/32" x 1/16" ID, 1/32", 31 (YO-30703-43)
Category: Barbed to barbed connectors
Description: Straight barbed reducing connectors
Brand:
Cole-Parmer
YO-30703-43
More Details
Thermo Scientific Nalgene graduated polymethylpentene cylinder, 250 mL
Category: Plastic Graduated Cylinders
Description: Thermo Scientific Nalgene Graduated Cylinders, PMP
Brand:
Thermo Scientific Nalgene
Model Number:
3665-0250
YO-06134-55
More Details
Pump, Centrifugal Pump Flanges, Cast Iron, 1.5"
Category: Centrifugal Pump Accessories
Description: Armstrong Pumps - Flanges for centrifugal pumps
Brand:
Armstrong
YO-79730-88
More Details
Quick-disconnect fittings, Panel-mount John Guest straight-through fit (PLC11004)
Category: Panel-Mount John Guest Bodies
Description: Standard body
Brand:
Delrin
Model Number:
PLC11004
YO-31304-40
More Details
Quick-Disconnect Panel-Mount Hose Barb Fitting Body; Straight-through, (PMC1604)
Category: Panel-Mount Hose Barb Bodies
Description: Straight-through Panel-Mount Hose Barb Bodies
Brand:
Colder
Model Number:
PMC1604
YO-06361-06
More Details
Ansell Touch-N-Tuff™ Nitrile Gloves, Powdered, Large
Category: Disposable Nitrile Gloves
Description: Ansell Touch-N-Tuff Nitrile Gloves
Brand:
Touch-N-Tuff
Model Number:
92-500 585190
YO-81602-60
More Details
Ltc7pn Lithium Battery (YO-18002-81)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
YO-18002-81
More Details
Masterflex TEE-Connector, polypropylene, B/T 90, 10/pack
Category: Masterflex Barbed Fittings
Description: Masterflex Tee connectors
Brand:
Masterflex
YO-30613-71
More Details
General Purpose Aluminum Weighing Dishes, 44mm i.d., 51mm o.d. x 1/2" (D44-100 (Pack of 100))
Category: Weighing Dishes
Description: Aluminum Weighing Dishes - General Purpose
Brand:
Eagle Thermoplastic
Model Number:
D44-100 (Pack of 100)
YO-01018-60
More Details
Heavy-duty low-density polyethylene pail, 10 quart (2963-GRAY)
Category: Buckets and Pails
Description: Heavy-Duty LDPE Pails
Brand:
Cole-Parmer
Model Number:
2963-GRAY
YO-06272-10
More Details
Safety Sign, Danger Use Lockout Before Working On Equipment, 10x14 Pla (MLKT016VP)
Category: Lockout/Tagout Signs
Description: DANGER - USE LOCKOUT BEFORE WORKING ON EQUIPMENT
Brand:
Cole-Parmer
Model Number:
MLKT016VP
YO-40711-30
More Details
Cole-Parmer Single-Use Temperature Recorder; 5 days, -20 to 100°F/
Category: Temperature Dataloggers
Description: Cole-Parmer Single-Use Temperature Recorders
Brand:
Cole-Parmer
Model Number:
16000/5 DAY TR
YO-80150-40
More Details
Quick-Disconnect Panel-Mount Hose Barb Fitting Body; Straight-through, (PLC16006)
Category: Panel-Mount Hose Barb Bodies
Description: Straight-through Panel-Mount Hose Barb Bodies
Brand:
Colder
Model Number:
PLC16006
YO-06361-48
More Details
64114
results
<<<Previous
1996
1997
1998
1999
2000
2001
2002
2003
2004
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
1,000
1,100
1,200
1,300
1,400
1,500
1,600
1,700
1,800
1,900
1,910
1,920
1,930
1,940
1,950
1,960
1,970
1,980
1,985
1,986
1,987
1,988
1,989
1,990
1,991
1,992
1,993
1,994
1,995
1,996
1,997
1,998
1,999
2,000
2,001
2,002
2,003
2,004
2,005
2,006
2,007
2,008
2,009
2,010
2,011
2,012
2,013
2,014
2,020
2,030
2,040
2,050
2,060
2,070
2,080
2,090
2,100
2,138
Ansell Touch-N-Tuff™ Nitrile Glove to Quick-Disconnect Panel-Mount Hose Barb Fitting Body
>
31125329-p-toc_2 31125329ptoc2