Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 28

5-Ft Xpert Filtered Balance System with Guardian Airflow Monitor   to   Thermo Scientific Apex 100 Metal Detector

Image
  Product Detail
SKU

5-ft Xpert Filtered Balance System With Guardian Airflow Monitor, 115V (YO-09105-10)

Category: Balance Enclosures
Description: Labconco Filtered Balance Systems
Brand: Labconco


Keithley 2430 Sourcemeter, 100V, 10A, 1000W

Category: SourceMeters
Description: Keithley 2400 Series Sourcemeters
Brand: Keithley
Model Number: 2430


Purifier Cell Logic Biosafety Cabinet; 3 ft wide; Temperature Zone; (YO-33516-02)

Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Cell Logic Class II Biosafety Cabinets
Brand: Labconco


Rotary evaporator with digital display, safety-coated vertical glasswa (YO-23122-17)

Category: Buchi Rotary Evaporators and Accessories
Description: Buchi Rotavapor R-215 Digital Rotary Evaporators
Brand: Buchi


Rotary evaporator with digital display, safety-coated vertical glasswa (YO-23112-17)

Category: Buchi Rotary Evaporators and Accessories
Description: Buchi Rotavapor R-215 Digital Rotary Evaporators
Brand: Buchi


Labconco Purifier 6 ft Horizontal Clean Bench, 34" D/115V

Category: Ductless Fume Hoods
Description: Labconco Purifier Horizontal Clean Benches
Brand: Labconco


Labconco Purifier 6 ft Horizontal Clean Bench, 34" D/UK

Category: Ductless Fume Hoods
Description: Labconco Purifier Horizontal Clean Benches
Brand: Labconco


6-ft XStream Hood, 115V, with 2 fixtures (YO-09099-22)

Category: Ducted Fume Hoods
Description: Labconco® Protector® XStream Laboratory Hoods
Brand: Labconco


6-ft XStream Hood, 230V, with 2 fixtures (YO-09099-26)

Category: Ducted Fume Hoods
Description: Labconco® Protector® XStream Laboratory Hoods
Brand: Labconco


Rotary evaporator with digital display, safety-coated dry ice condense (YO-23122-27)

Category: Buchi Rotary Evaporators and Accessories
Description: Buchi Rotavapor R-215 Digital Rotary Evaporators
Brand: Buchi


Rotary evaporator with digital display, safety-coated dry ice condense (YO-23112-27)

Category: Buchi Rotary Evaporators and Accessories
Description: Buchi Rotavapor R-215 Digital Rotary Evaporators
Brand: Buchi


Self-Monitoring Ductless Fume Hood, 42", 115 VAC (YO-33713-10)

Category: Ductless Fume Hoods
Description: Self-Monitoring Ductless Fume Hoods
Brand: Cole-Parmer


Thermo Scientific APEX 100 Metal Detector, with 500 mm width x 200 mm

Category: Metal Detectors
Description: Thermo Scientific APEX 100 Metal Detectors
Brand: Thermo Scientific
Model Number: APEX 100 150 x 100


Rotary Evaporator, Safety-coated Dry Ice Reflux Condenser, Professiona (YO-23022-47)

Category: Buchi Rotary Evaporators and Accessories
Description: Buchi Rotavapor R-210 Analog Rotary Evaporators
Brand: Buchi


Additel 761-L-N Automated Pressure Calibrator

Category: Pressure Calibrators
Description: Additel 761 Automated Pressure Calibrators
Brand: ADDITEL
Model Number: 761-L-N


Rotary Evaporator, Safety-coated Dry Ice Reflux Condenser, Professiona (YO-23012-47)

Category: Buchi Rotary Evaporators and Accessories
Description: Buchi Rotavapor R-210 Analog Rotary Evaporators
Brand: Buchi


48" Explosion-Proof Bypass Fume Hood, 115V/60Hz (YO-09012-16)

Category: Ducted Fume Hoods
Description: Labconco® Protector® Premier® Fiberglass Fume Hoods
Brand: Labconco


Thermo Scientific APEX 100 Metal Detector, with 650 mm width x 150 mm

Category: Metal Detectors
Description: Thermo Scientific APEX 100 Metal Detectors
Brand: Thermo Scientific
Model Number: APEX 100 150 x 100


Purifier Cell Logic Biosafety Cabinet; 3 ft wide; Scope-Ready; 115 V

Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Cell Logic Class II Biosafety Cabinets
Brand: Labconco


3-ft Purifier® Cell Logic®+, Class II A2, 115V, Temp-Zone/UV

Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier® Cell Logic+® Class II Biosafety Cabinets
Brand: Labconco


3-ft Purifier® Cell Logic®+, Class II A2, 230V (NA), Temp-Zone/UV

Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier® Cell Logic+® Class II Biosafety Cabinets
Brand: Labconco


Rotary evaporator with digital display, safety-coated dry ice reflux c (YO-23122-46)

Category: Buchi Rotary Evaporators and Accessories
Description: Buchi Rotavapor R-215 Digital Rotary Evaporators
Brand: Buchi


Rotary evaporator with digital display, safety-coated dry ice reflux c (YO-23112-46)

Category: Buchi Rotary Evaporators and Accessories
Description: Buchi Rotavapor R-215 Digital Rotary Evaporators
Brand: Buchi


Complete 5foot Fume Hood System With Acid And Base Storage Cabinets Ep (YO-41525-10)

Category: Hamilton Constant Volume Bypass Fume Hoods
Description: Hamilton Constant Volume Benchtop Fume Hoods Assemblies
Brand: Thermo Scientific Hamilton


TBS SHUR/Cut™ 2500 Semi-automatic non-retracting microtome, 100-120

Category: TBS SHUR/Cut Microtomes
Description: TBS SHUR/Cu Microtome Series
Brand: TBS


Protector Premier, 4', No Services, With Blower, Explosion Proof, 115V

Category: Ducted Fume Hoods
Description: Labconco Protector Premier Fume Hoods
Brand: LABCONCO


Protector Premier, 5', Two Services, With Blower, 230V, 60Hz

Category: Ducted Fume Hoods
Description: Labconco Protector Premier Fume Hoods
Brand: LABCONCO


Three Zone Single Program, Multiple Segment Programmable Controller fo

Category: Thermo Scientific Lindberg Blue M Split-Hinge Tube Furnaces and Controllers
Description: Thermo Scientific Lindberg Blue M Split-Hinge Tube Furnace Controllers
Brand: Thermo Scientific Lindberg
Model Number: CC584343PC-1


Thermo Scientific APEX 100 Metal Detector, with 700 mm width x 125 mm

Category: Metal Detectors
Description: Thermo Scientific APEX 100 Metal Detectors
Brand: Thermo Scientific
Model Number: APEX 100 150 x 100


Thermo Scientific APEX 100 Metal Detector, with 450 mm width x 200 mm

Category: Metal Detectors
Description: Thermo Scientific APEX 100 Metal Detectors
Brand: Thermo Scientific
Model Number: APEX 100 150 x 100




5-Ft Xpert Filtered Balance System with Guardian Airflow Monitor to Thermo Scientific Apex 100 Metal Detector

>