Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 4
Vertical Tube Furnace
to
Thermo Scientific Revco
Image
Product Detail
SKU
Vertical Tube Furnace, 24-3/4" Tube Length, 1600°C, 208 / 240 VAC (RHTV 120/300/16/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTV 120/300/16/C42
YO-33334-49
More Details
Calorimeter C 2000 Basic V1 230V
Category: Calorimeters
Description: IKA C 2000 Calorimeter Systems
Brand:
IKA
YO-50905-25
More Details
Temperature and Humidity Controlled glove box; 2 port; 230 VAC, 50 Hz (YO-34750-45)
Category: Glove Boxes
Description: Plas-Labs Controlled Atmosphere Glove Boxes
Brand:
Plas-Labs
YO-34750-45
More Details
Thermo Scientific NESLAB 15.1 L -90°C Circulating Bath 220-240 V, (YO-13400-02)
Category: Thermo Scientific Refrigerated Circulators
Description: Thermo Scientific NESLAB Ultra Low Temperature Circulating Baths
Brand:
Thermo Scientific
YO-13400-02
More Details
Koehler Automatic Density Meter
Category: Petroleum Testing
Description: Koehler Automatic Density Meter
Brand:
Koehler
YO-59876-83
More Details
Temperature and Humidity Controlled glove box; 2 port; 110 VAC, 60 Hz (YO-34750-40)
Category: Glove Boxes
Description: Plas-Labs Controlled Atmosphere Glove Boxes
Brand:
Plas-Labs
YO-34750-40
More Details
Fluke Ti55FT Thermal Imager with IR-Fusion (FLK-TI55FT-20)
Category: Industrial Thermal Imager
Description: Fluke Ti50, Ti55 Advanced FlexCam Industrial Thermal Imagers
Brand:
Fluke
Model Number:
FLK-TI55FT-20
YO-39715-30
More Details
Vertical Tube Furnace, 18-7/8" Tube Length, 1700°C, 208 / 240 VAC (RHTV 120/150/17/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTV 120/150/17/C42
YO-33334-47
More Details
Hypoxia Glove Box Chamber, 230v
Category: Glove Boxes
Description: Plas-Labs Hypoxia Chamber Glove Box
Brand:
Plas-Labs
Model Number:
856-HYPO/EXP
YO-34755-02
More Details
14-1/2 ft, nine cabinet island assembly with reagent shelf, sink, and (YO-41526-15)
Category: Fume Hoods
Description: Hamilton Lab Bench Island Assemblies
Brand:
Thermo Scientific
YO-41526-15
More Details
Horizontal Tube Furnace, 24-3/8" Tube Length, 1600°C, 208 / 240 V (RHTH 120/300/16/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTH 120/300/16/C42
YO-33334-40
More Details
Horizontal Tube Furnace, 18-1/2" Tube Length, 1700°C, 208 / 240 V (RHTH 120/150/17/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTH 120/150/17/C42
YO-33334-38
More Details
Thermo Scientific Focus Gc Automated Bundle With 8-position Auto-injec (YO-34010-60)
Category: Gas Chromatography (GC)
Description: Thermo Scientific FOCUS Gas Chromatographs
Brand:
Thermo Scientific
YO-34010-60
More Details
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 33.5 c (YO-44146-14)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco UxF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-14
More Details
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 33.5 c (YO-44146-13)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco UxF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-13
More Details
Hypoxia Glove Box Chamber, 115v
Category: Glove Boxes
Description: Plas-Labs Hypoxia Chamber Glove Box
Brand:
Plas-Labs
Model Number:
856-HYPO
YO-34755-00
More Details
Systems for cell culture applications, 115 VAC (YO-29200-32)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-32
More Details
Tektronix MSO4104B, 1 GHz, 5 GS/s, 20M Record Length, 4 + 16 Channels
Category: Benchtop Oscilloscopes
Description: Tektronix MSO4000B Mixed Signal Phosphor Oscilloscopes
Brand:
Tektronix
Model Number:
MSO4104B
YO-20040-75
More Details
Systems for cell culture applications, 220 VAC (YO-29200-33)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-33
More Details
Tintometer PFXi-995 General Purpose Long Pathlength Color Meter with H
Category: Water Quality Test Strips, Indicators, and Kits
Description: Tintometer PFXi Series Long-Pathlength Color Meters
Brand:
LOVIBOND
YO-99562-26
More Details
Mettler Toledo Excellence Plus XP Micro Balance 5.1/22G X .001MG/.01MG
Category: Semi/Micro Analytical Balances
Description: Mettler Toledo Excellence Plus XP Analytical, Semi-Micro, and Micro Balances
Brand:
Mettler Toledo
Model Number:
XP26DR
YO-11332-72
More Details
Vertical Tube Furnace, 18-7/8" Tube Length, 1600°C, 208 / 240 VAC (RHTV 120/150/16/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTV 120/150/16/C42
YO-33334-46
More Details
APPLIKON RAMbio Acoustic Mixer
Category: Incubating Shakers
Description: Applikon RAMbio Benchtop Acoustic Mixer
Brand:
Applikon
Model Number:
Z361000010
YO-29206-83
More Details
Tektronix MDO4054-3 Mixed Domain Oscilloscope with (4) 500 MHz analog
Category: Benchtop Oscilloscopes
Description: Tektronix MDO4000 Series Mixed Domain Oscilloscopes
Brand:
Tektronix
YO-20042-70
More Details
TDS5104B: 4-Channel, 1 GHz, up to 5 GS/s, 16M Record Length (TDS5104B)
Category: Benchtop Oscilloscopes
Description: Tektronix TDS5000B Series Oscilloscopes
Brand:
Tektronix
Model Number:
TDS5104B
YO-20014-50
More Details
FLIR T620 Thermal Imaging Infrared Camera for Industrial w/25 and 45 d
Category: Industrial Thermal Imager
Description: Flir T620, T640 Advanced Industrial Thermal Imagers
Brand:
Flir
Model Number:
T620
YO-39754-13
More Details
FLIR T620 Thermal Imaging Infrared Camera for Industrial w/25 deg lens
Category: Industrial Thermal Imager
Description: Flir T620, T640 Advanced Industrial Thermal Imagers
Brand:
Flir
Model Number:
T620
YO-39754-18
More Details
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 28.8 c (YO-44146-11)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco UxF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-11
More Details
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 28.8 c (YO-44146-12)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco UxF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-12
More Details
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 28.8 c (YO-44146-10)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco UxF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-10
More Details
64114
results
<<<Previous
1
2
3
4
5
6
7
8
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Vertical Tube Furnace to Thermo Scientific Revco
>
31125329-p-toc_2 31125329ptoc2