Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 5
Thermo Scientific Neslab 15
to
Fluke 1760 Us
Image
Product Detail
SKU
Thermo Scientific NESLAB 15.1 L -90°C Circulating Bath 208-230 V, (YO-13400-00)
Category: Thermo Scientific Refrigerated Circulators
Description: Thermo Scientific NESLAB Ultra Low Temperature Circulating Baths
Brand:
Thermo Scientific
YO-13400-00
More Details
Temperature And Humidity Chamber 100 L, (-20 C To 150°C) 100L; 220
Category: Environmental Chambers
Description: Temperature and Environmental Chambers
Brand:
Lab Companion
Model Number:
AAH82131
YO-52401-94
More Details
Temperature And Humidity Chamber 100 L, (-20 C To 150°C) 100L; 230
Category: Environmental Chambers
Description: Temperature and Environmental Chambers
Brand:
Lab Companion
Model Number:
AAH82132
YO-52401-97
More Details
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital 2.8 cu ft, 2 (23210606)
Category: Sterilizers
Description: Heidolph Tabletop Autoclaves
Brand:
Brinkmann
Model Number:
23210606
YO-10780-10
More Details
Tektronix MSO4104, 1 GHz, 2.5 GS/s, 4-channel analog, 16-digital chann (YO-20028-06)
Category: Oscilloscopes
Description: Tektronix MSO4000 Mixed Signal Digital Phosphor Oscilloscopes
Brand:
TEKTRONIX
YO-20028-06
More Details
Tektronix DPO5104 1 GHz, 10/5 GS/s(2/4 CH), 12.5M record length, 4-cha
Category: Benchtop Oscilloscopes
Description: Tektronix MSO5000 and DPO5000 Mixed Signal Phosphor Oscilloscopes
Brand:
Tektronix
Model Number:
DPO5104
YO-20041-22
More Details
Tintometer PFXi-995 General Purpose Long Pathlength Color Meter
Category: Water Quality Test Strips, Indicators, and Kits
Description: Tintometer PFXi Series Long-Pathlength Color Meters
Brand:
LOVIBOND
YO-99562-24
More Details
Cell fusion system (YO-02894-02)
Category: Electroporation Systems
Description: BTX Electro Cell Fusion Systems
Brand:
BTX
YO-02894-02
More Details
300-liter Plant Growth Chamber With Humidity Control, 230VAC (AAHA1011) (YO-39353-15)
Category: Incubators
Description: Lab Companion Plant Growth Chambers
Brand:
Lab Companion
Model Number:
AAHA1011
YO-39353-15
More Details
300-liter Plant Growth Chamber With Humidity Control, 230VAC (AAHA1011) (YO-39353-10)
Category: Incubators
Description: Lab Companion Plant Growth Chambers
Brand:
Lab Companion
Model Number:
AAHA1011
YO-39353-10
More Details
Systems for cell culture applications, water-jacketed, 115 VAC (YO-29200-14)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-14
More Details
Systems for cell culture applications, water-jacketed, 220 VAC (YO-29200-15)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-15
More Details
10-Liter Rotary Vacuum Evaporator
Category: Rotary Evaporator Systems
Description: 10-Liter Rotary Vacuum Evaporator System
Brand:
Cole-Parmer
YO-28615-11
More Details
Thermo Scientific Barnstead LabTower EDI Water Purification System 30
Category: EASYpure Water Purification Systems
Description: Thermo Scientific Barnstead LabTower EDI Water Purification Systems
Brand:
Barnstead
YO-99281-30
More Details
Tektronix MSO5054 500 MHz, 5 GS/s, 12.5M record length, 4+16 channel m
Category: Benchtop Oscilloscopes
Description: Tektronix MSO5000 and DPO5000 Mixed Signal Phosphor Oscilloscopes
Brand:
Tektronix
Model Number:
MSO5054
YO-20041-25
More Details
Nor-Lake Scientific Select™ Pass-thru Refrigerator +4C, Six Glass D
Category: General-Purpose Laboratory Refrigerators
Description: Nor-Lake Scientific Select Pass-Thru Laboratory Refrigerators
Brand:
Nor-Lake
YO-44174-97
More Details
Horizontal Tube Furnace, 18-1/2" Tube Length, 1600°C, 208 / 240 V (RHTH 120/150/16/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTH 120/150/16/C42
YO-33334-37
More Details
TBS Minotome PLUS™ Cryostat,115-100VAC,50/60Hz
Category: Histology Equipment
Description: TBS Rotary Minotome PLUS Cryostat
Brand:
TBS
YO-48578-36
More Details
Minotome PLUS™ Cryostat, 200-240 VAC, 50Hz
Category: Histology Equipment
Description: TBS Rotary Minotome PLUS Cryostat
Brand:
TBS
YO-48578-37
More Details
Microbial systems (YO-29200-10)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Microbial and Bacterial Applications
Brand:
Cole-Parmer
YO-29200-10
More Details
Microbial systems (YO-29200-11)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Microbial and Bacterial Applications
Brand:
Cole-Parmer
YO-29200-11
More Details
Nor-Lake Scientific Select™ Pass-thru Refrigerator +4C, Six Solid D
Category: General-Purpose Laboratory Refrigerators
Description: Nor-Lake Scientific Select Pass-Thru Laboratory Refrigerators
Brand:
Nor-Lake
YO-44175-17
More Details
12L Console Freeze Dryer With Purge Valve And Mini Chamber 220vac 50hz (YO-03336-63)
Category: Freeze Dryers
Description: Labconco 12-liter FreeZone Freeze Dry Systems
Brand:
Labconco
YO-03336-63
More Details
12L Console Freeze Dryer With Purge Valve And Mini Chamber 230vac 60hz (YO-03336-73)
Category: Freeze Dryers
Description: Labconco 12-liter FreeZone Freeze Dry Systems
Brand:
Labconco
YO-03336-73
More Details
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 24.1 c (YO-44146-07)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco UxF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-07
More Details
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 24.1 c (YO-44146-08)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco UxF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-08
More Details
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 24.1 c (YO-44146-09)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco UxF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-09
More Details
Tektronix PA4000 Multi-phase Power Analyzer, Four Input Channel
Category: Power Meters/Analyzers
Description: Tektronix PA4000 Series Power Analyzers
Brand:
Tektronix
YO-20045-53
More Details
Fluke TIR4/FT-10/20 Thermal Imager with IR-Fusion for the Building In (FLK-TIR4/FT-10/20)
Category: Building Industry Thermal Imagers
Description: Fluke Tir2, Tir3, Tir4 Advanced FlexCam Industrial Thermal Imagers
Brand:
Fluke
Model Number:
FLK-TIR4/FT-10/20
YO-39715-35
More Details
Fluke 1760 US, Power Quality Recorder, no Trans
Category: Power Meters/Analyzers
Description: Fluke 1760 Three Phase Power Quality Recorder
Brand:
Fluke
Model Number:
FLUKE-1760 US
YO-20035-77
More Details
64114
results
<<<Previous
1
2
3
4
5
6
7
8
9
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Thermo Scientific Neslab 15 to Fluke 1760 Us
>
31125329-p-toc_2 31125329ptoc2