Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 6
Temperature and Humidity Chamber 65 L
to
Cole-Parmer Lc2 System 5ml
Image
Product Detail
SKU
Temperature And Humidity Chamber 65 L, (-20 C To 150°C) 65L; 220V
Category: Environmental Chambers
Description: Temperature and Environmental Chambers
Brand:
Lab Companion
Model Number:
AAH82121
YO-52401-93
More Details
Temperature And Humidity Chamber 65 L, (-20 C To 150°C) 65L; 230V
Category: Environmental Chambers
Description: Temperature and Environmental Chambers
Brand:
Lab Companion
Model Number:
AAH82122
YO-52401-96
More Details
Large-Hose Peristaltic Pump, Dry Pump Cavity, Hypalon Hose, 57 GPM, co
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Hypalon
Model Number:
2009-HF-BB-D2
YO-75800-44
More Details
Large-Hose Peristaltic Pump, Dry Pump Cavity, Hypalon Hose, 97 GPM, in
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Hypalon
Model Number:
2009-HF-BB-J2
YO-75800-49
More Details
Keithley 2657A Sourcemeter, High Power, 180W, 120 mA
Category: SourceMeters
Description: Keithley 2657A High Power SourceMeter
Brand:
Keithley
Model Number:
2657A
YO-20045-43
More Details
Fluke Ti50FT Thermal Imager with IR-Fusion (FLK-TI50FT-20)
Category: Industrial Thermal Imager
Description: Fluke Ti50, Ti55 Advanced FlexCam Industrial Thermal Imagers
Brand:
Fluke
Model Number:
FLK-TI50FT-20
YO-39715-28
More Details
Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 28 cub (YO-44146-29)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco ExF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-29
More Details
Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 28 cub (YO-44146-30)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco ExF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-30
More Details
Systems for cell culture applications, 115 VAC (YO-29200-12)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-12
More Details
Systems for cell culture applications, 220 VAC (YO-29200-13)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-13
More Details
Large-Hose Peristaltic Pump, Dry Pump Cavity, EPDM Hose, 97 GPM, inter
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2009-EF-BB-J2
YO-75800-48
More Details
Labconco XPert NANO Enclosures, 6-ft wide; 230V 50 HZ W/IONIZER; 30 (YO-33512-49)
Category: Balance Enclosures
Description: Labconco XPert Nano Enclosures
Brand:
Labconco
YO-33512-49
More Details
Labconco XPert NANO Enclosures, 6-ft wide; 100V 60 HZ W/IONIZER; 30 (YO-33512-47)
Category: Balance Enclosures
Description: Labconco XPert Nano Enclosures
Brand:
Labconco
YO-33512-47
More Details
Labconco XPert NANO Enclosures, 6-ft wide; 115V 60 HZ W/IONIZER; 30 (YO-33512-48)
Category: Balance Enclosures
Description: Labconco XPert Nano Enclosures
Brand:
Labconco
YO-33512-48
More Details
Large-Hose Peristaltic Pump, Dry Pump Cavity, Heavy Duty Natural Rubbe (2009-MF-BB-J2)
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2009-MF-BB-J2
YO-75800-47
More Details
6' Purifier Logic+ B2 Biosafety Cabinet. Energy efficient ECM motor. I (YO-33517-75)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic+ Class II Biosafety Cabinets
Brand:
Labconco
YO-33517-75
More Details
6' Purifier Logic+ B2 Biosafety Cabinet. Energy efficient ECM motor. I (YO-33517-79)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic+ Class II Biosafety Cabinets
Brand:
Labconco
YO-33517-79
More Details
Labconco Protector XL Floor-mounted Hoods, 8 ft, 30" deep
Category: Ducted Fume Hoods
Description: Labconco Protector XL Floor-Mounted Hoods
Brand:
Labconco
YO-09098-80
More Details
industrial Hose Pump, Natural Rubber Hose, 97 GPM, intermittent duty,
Category: Positive Displacement Pumps
Description: Large-Hose, Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2009-NF-BB-J2
YO-75800-46
More Details
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital with Printer (23210568)
Category: Sterilizing Equipment
Description: Heidolph Tabletop Autoclaves
Brand:
Heidolph
Model Number:
23210568
YO-10780-13
More Details
Temperature And Humidity Chamber 100 L, (0 C To 120°C) 100L; 230V
Category: Environmental Chambers
Description: Temperature and Environmental Chambers
Brand:
Lab Companion
Model Number:
AAH82232
YO-52401-91
More Details
Temperature And Humidity Chamber 100 L, (0 C To 120°C) 100L; 220V
Category: Environmental Chambers
Description: Temperature and Environmental Chambers
Brand:
Lab Companion
Model Number:
AAH82231
YO-52401-88
More Details
Class II B2 6-ft Biosafety Cabinet with 8" sash opening and UV light, (YO-33515-75)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic Class II Biosafety Cabinets
Brand:
Labconco
YO-33515-75
More Details
Medium Capacity Steam Generator; 542 Lbs/hr, 480 VAC (ES180100F3)
Category: Steam Generators
Description: Sussman Medium Capacity Electric Steam Generators
Brand:
Sussman
Model Number:
ES180100F3
YO-07601-39
More Details
Large-Hose Peristaltic Pump, Dry Pump Cavity, EPDM Hose, 57 GPM, conti
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2009-EF-BB-D2
YO-75800-43
More Details
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 19.4 c (YO-44146-04)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco UxF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-04
More Details
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 19.4 c (YO-44146-05)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco UxF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-05
More Details
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 19.4 c (YO-44146-06)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco UxF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-06
More Details
Tektronix DPO4104B, 1 GHz, 5 GS/s, 20M Record Length, 4 channel Digita
Category: Benchtop Oscilloscopes
Description: Tektronix DPO4000B Digital Phosphor Oscilloscopes
Brand:
Tektronix
Model Number:
DPO4104B
YO-20040-72
More Details
Cole-Parmer LC2 System 5ml, PEEK, Clarity
Category: High Pressure Liquid Chromatography (HPLC) Systems
Description: Cole-Parmer Economical Binary Gradient System
Brand:
Cole-Parmer
Model Number:
B2305CLP
YO-42662-51
More Details
64114
results
<<<Previous
2
3
4
5
6
7
8
9
10
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Temperature and Humidity Chamber 65 L to Cole-Parmer Lc2 System 5ml
>
31125329-p-toc_2 31125329ptoc2