Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 7
Cole-Parmer Lc2 System 5ml
to
11-1
Image
Product Detail
SKU
Cole-Parmer LC2 System 5ml, SS, Clarity
Category: High Pressure Liquid Chromatography (HPLC) Systems
Description: Cole-Parmer Economical Binary Gradient System
Brand:
Cole-Parmer
Model Number:
B2305CLS
YO-42662-53
More Details
Cole-Parmer LC2 System 10ml, PEEK, Clarity
Category: High Pressure Liquid Chromatography (HPLC) Systems
Description: Cole-Parmer Economical Binary Gradient System
Brand:
Cole-Parmer
Model Number:
B2310CLP
YO-42662-55
More Details
Cole-Parmer LC2 System 10ml, SS, Clarity
Category: High Pressure Liquid Chromatography (HPLC) Systems
Description: Cole-Parmer Economical Binary Gradient System
Brand:
Cole-Parmer
Model Number:
B2310CLS
YO-42662-57
More Details
Labconco Protector XL Floor-mounted Hoods, 6 ft, 48" deep
Category: Ducted Fume Hoods
Description: Labconco Protector XL Floor-Mounted Hoods
Brand:
Labconco
YO-09098-74
More Details
Parker Balston Zero Air Generator; 30 LPM; 220VAC/50Hz
Category: Gas Generators
Description: Parker Balston Zero Air Generators
Brand:
Cole-Parmer
YO-86442-21
More Details
Parker Balston Zero Air Generator; 30 LPM; 120VAC/60Hz
Category: Gas Generators
Description: Parker Balston Zero Air Generators
Brand:
Cole-Parmer
YO-86442-11
More Details
Large-Hose Peristaltic Pump, Dry Pump Cavity, Heavy Duty Natural Rubbe (2009-MF-BB-D2)
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2009-MF-BB-D2
YO-75800-42
More Details
Class II B2 6-ft Biosafety Cabinet with 8" sash opening and UV light, (YO-33515-55)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic Class II Biosafety Cabinets
Brand:
Labconco
YO-33515-55
More Details
6' Purifier Logic+ B2 Biosafety Cabinet. Energy efficient ECM motor. I (YO-33517-74)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic+ Class II Biosafety Cabinets
Brand:
Labconco
YO-33517-74
More Details
6' Purifier Logic+ B2 Biosafety Cabinet. Energy efficient ECM motor. I (YO-33517-78)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic+ Class II Biosafety Cabinets
Brand:
Labconco
YO-33517-78
More Details
Tektronix MSO4054B, 500MHz, 2.5 GS/s, 20M Record Length, 4 + 16 Channe
Category: Benchtop Oscilloscopes
Description: Tektronix MSO4000B Mixed Signal Phosphor Oscilloscopes
Brand:
Tektronix
Model Number:
MSO4054B
YO-20040-74
More Details
6' Purifier Logic+ B2 Biosafety Cabinet. Energy efficient ECM motor. I (YO-33517-73)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic+ Class II Biosafety Cabinets
Brand:
Labconco
YO-33517-73
More Details
6' Purifier Logic+ B2 Biosafety Cabinet. Energy efficient ECM motor. I (YO-33517-77)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic+ Class II Biosafety Cabinets
Brand:
Labconco
YO-33517-77
More Details
Temperature And Humidity Chamber 20 L, (-20 C To 150°C) 20L; 220V
Category: Environmental Chambers
Description: Temperature and Environmental Chambers
Brand:
Lab Companion
Model Number:
AAH82111
YO-52401-92
More Details
Temperature And Humidity Chamber 20 L, (-20 C To 150°C) 20L; 230V
Category: Environmental Chambers
Description: Temperature and Environmental Chambers
Brand:
Lab Companion
Model Number:
AAH82112
YO-52401-95
More Details
TBS SHUR/Cut™ 4500 Fully-automated retracting microtome w/ adjustab
Category: TBS SHUR/Cut Microtomes
Description: TBS SHUR/Cu Microtome Series
Brand:
TBS
YO-48577-04
More Details
industrial Hose Pump, Natural Rubber Hose, 57 GPM, continuous duty, 7.
Category: Positive Displacement Pumps
Description: Large-Hose, Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2009-NF-BB-D2
YO-75800-41
More Details
Thermo Scientific NESLAB 15.1 L -80°C Circulating Bath 208-230 V, (YO-13400-06)
Category: Thermo Scientific Refrigerated Circulators
Description: Thermo Scientific NESLAB Ultra Low Temperature Circulating Baths
Brand:
Thermo Scientific
YO-13400-06
More Details
Class II B2 6-ft Biosafety Cabinet with 8" sash opening and fluorescen (YO-33515-65)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic Class II Biosafety Cabinets
Brand:
Labconco
YO-33515-65
More Details
Koehler Cold Filter Plugging Point Bath, 220 to 240 VAC, 50 Hz
Category: Petroleum Testing
Description: Koehler Mechanically Refrigerated Cold Filter Plugging Point Bath
Brand:
Koehler
YO-59876-91
More Details
12L Cnsl Freez Dry W/prge 230 (YO-03336-70)
Category: Freeze Dryers
Description: Labconco 12-liter FreeZone Freeze Dry Systems
Brand:
Labconco
YO-03336-70
More Details
12L Cnsl Freez Dry W/prge 230 (YO-03336-65)
Category: Freeze Dryers
Description: Labconco 12-liter FreeZone Freeze Dry Systems
Brand:
Labconco
YO-03336-65
More Details
Embryo manipulation system (YO-02894-00)
Category: Electroporation Systems
Description: BTX Electro Cell Fusion Systems
Brand:
BTX
YO-02894-00
More Details
Thermo Scientific Focus Gc Basic Bundle 115VAC (YO-34010-50)
Category: Gas Chromatography (GC)
Description: Thermo Scientific FOCUS Gas Chromatographs
Brand:
Thermo Scientific
YO-34010-50
More Details
Labconco XPert NANO Enclosures, 6-ft wide; 230V 50 HZ W/IONIZER; 30 (YO-33512-44)
Category: Balance Enclosures
Description: Labconco XPert Nano Enclosures
Brand:
Labconco
YO-33512-44
More Details
Thermo Scientific Barnstead LabTower EDI Water Purification System 15
Category: EASYpure Water Purification Systems
Description: Thermo Scientific Barnstead LabTower EDI Water Purification Systems
Brand:
Barnstead
YO-99281-15
More Details
Labconco XPert NANO Enclosures, 6-ft wide; 100V 60 HZ W/IONIZER; 30 (YO-33512-42)
Category: Balance Enclosures
Description: Labconco XPert Nano Enclosures
Brand:
Labconco
YO-33512-42
More Details
Labconco XPert NANO Enclosures, 6-ft wide; 115V 60 HZ W/IONIZER; 30 (YO-33512-43)
Category: Balance Enclosures
Description: Labconco XPert Nano Enclosures
Brand:
Labconco
YO-33512-43
More Details
Class II B2 6-ft Biosafety Cabinet with 8" sash opening and fluorescen (YO-33515-45)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic Class II Biosafety Cabinets
Brand:
Labconco
YO-33515-45
More Details
11-1/2 ft, seven cabinet island assembly with reagent shelf, sink, and (YO-41526-20)
Category: Fume Hoods
Description: Hamilton Lab Bench Island Assemblies
Brand:
Thermo Scientific
YO-41526-20
More Details
64114
results
<<<Previous
3
4
5
6
7
8
9
10
11
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Cole-Parmer Lc2 System 5ml to 11-1
>
31125329-p-toc_2 31125329ptoc2