Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 70
Fluke Calibration 7526a Precision Process Calibrator
to
Polyscience Performance Programmable 28l -30° Refrigerated Bath Ci (Yo-12210-41)
Image
Product Detail
SKU
Fluke Calibration 7526A Precision Process Calibrator
Category: Calibration Equipment
Description: Fluke Calibration (formerly Hart Scientific) Precision Process Calibrator
Brand:
Fluke Calibration
YO-16100-93
More Details
Atago Standard Abbe refractometer, 1.4700 to 1.8700 RI
Category: Abbe Refractometers
Description: Atago Abbe Refractometers
Brand:
ATAGO
Model Number:
1240 4T
YO-02940-06
More Details
Cole-Parmer Vacuum Pump, 20LM 2MB 115/230V
Category: Standard-Duty Vacuum Pumps
Description: Cole-Parmer Diaphragm Vacuum Pumps
Brand:
Cole-Parmer
YO-72226-55
More Details
Standard low-temperature cabinets; 6.8 cu ft; -85°C; 230V (85-6.8 50 HZ)
Category: Chest-style Freezers
Description: ScienTemp Standard Low-Temperature Cabinets
Brand:
ScienTemp
Model Number:
85-6.8 50 HZ
YO-03815-16
More Details
Flir B50 Thermal Imaging Infrared Camera With Laser For Building Indus (FLIR B50)
Category: Temperature Infrared Thermal Imagers
Description: Flir i and b-Series Compact Industrial and Building Thermal Imagers (i40, i50, i60, b40, b50, b60)
Brand:
Flir
Model Number:
FLIR B50
YO-39754-29
More Details
Mettler Toledo Deluxe Halogen Moisture Analyzer HG63, 61 g x 0.001 g (HG63)
Category: Moisture Balances
Description: METTLER TOLEDO Moisture Analyzers
Brand:
Mettler Toledo
Model Number:
HG63
YO-11334-51
More Details
Mettler Toledo Excellence Plus XP Precision Toploading Balance, 32,100
Category: Mettler Toledo Toploading Balances
Description: Mettler Toledo Excellence Plus XP Precision Toploading Balances
Brand:
Mettler Toledo
Model Number:
XP32000L
YO-11332-92
More Details
Standard Gear Pump System With Calibration Column, Relief Valve, Stati (CP-G24-1-1-1-1)
Category: Gear Pump Systems/Skids
Description: Standard Gear Pump Skid Systems - With Calibration Column, Relief Valve, and Static Mixer
Brand:
Cole-Parmer
Model Number:
CP-G24-1-1-1-1
YO-70787-30
More Details
Hermle High-Capacity Refrigerated Microcentrifuge; 120VAC, 50/60Hz
Category: Refrigerated Microcentrifuges
Description: Hermle High-Capacity Refrigerated Microcentrifuge
Brand:
Hermle Labnet
YO-17405-20
More Details
Hermle High-Capacity Refrigerated Microcentrifuge; 230VAC, 50/60Hz
Category: Refrigerated Microcentrifuges
Description: Hermle High-Capacity Refrigerated Microcentrifuge
Brand:
Hermle Labnet
YO-17405-25
More Details
XPert TXE Equipment Enclosure with Guardian Airflow Monitor, 115 volts (YO-33512-02)
Category: Balance Enclosures
Description: Labconco XPert TXE Equipment Equipment Enclosure
Brand:
Labconco
YO-33512-02
More Details
Mini glove box; 31 1/2 glove box 110v (815-PGB)
Category: Glove Boxes
Description: Plas-Labs La Petite Glove Box
Brand:
Plas-Labs
Model Number:
815-PGB
YO-34750-20
More Details
1 L Benchtop Freeze Dry System, -54° with SS interior, 220V (YO-03337-10)
Category: Freeze Dryers
Description: Freezone Personal Freeze Dry Systems
Brand:
Labconco
YO-03337-10
More Details
MUFFLE FURNACE, 910 CU.IN., 100-1200°C, 208 / 240 VAC (LT 15/12/P330)
Description: Nabertherm Muffle Furnaces
Brand:
Nabertherm
Model Number:
LT 15/12/P330
YO-33334-17
More Details
Sartorius Cubis Semi-Micro Balance, 120g by 0.01mg,iso Calibration
Category: Semi/Micro Analytical Balances
Description: Sartorius Cubis Analytical, Semi-Micro, and Micro Balances
Brand:
Sartorius
YO-11800-61
More Details
Sartorius Cubis Analytical Balance, 320 G X 0.1 MG, 120VAC (MSU324S-000-DU)
Category: Semi/Micro Analytical Balances
Description: Sartorius Cubis Analytical, Semi-Micro, and Micro Balances
Brand:
Sartorius
Model Number:
MSU324S-000-DU
YO-11228-37
More Details
Seward Stomacher 400C Lab Blenders, programmable, 110VAC (YO-36850-04)
Category: Reciprocating Paddle Blenders
Description: Seward Stomacher Lab Blenders
Brand:
Seward
YO-36850-04
More Details
Seward Stomacher 400C Lab Blenders, programmable, 230VAC (YO-36850-06)
Category: Reciprocating Paddle Blenders
Description: Seward Stomacher Lab Blenders
Brand:
Seward
YO-36850-06
More Details
Blender motor base for one-liter container; explosion proof; 115V
Category: Blenders and Paddle Blenders
Description: Explosion-Proof Laboratory Blenders
Brand:
Waring
YO-04257-50
More Details
uVISC™ Portable Viscometer Control Unit, Advanced
Category: uVISC Portable Viscometers
Description: uVISC Portable Viscometers Control Units
Brand:
Cole-Parmer
YO-98946-41
More Details
PolyScience Performance Programmable 20L -30° Refrigerated Bath Ci (YO-12210-34)
Category: PolyScience Refrigerated Circulators
Description: PolyScience Refrigerated Circulating Baths
Brand:
PolyScience
YO-12210-34
More Details
PolyScience Performance Programmable 20L -30° Refrigerated Bath Ci (YO-12210-35)
Category: PolyScience Refrigerated Circulators
Description: PolyScience Refrigerated Circulating Baths
Brand:
PolyScience
YO-12210-35
More Details
IKA T 65 D Homogenizers; 230VAC; 3 phase (YO-04737-01)
Category: IKA Homogenizers
Description: IKA T 65 D Batch Process Homogenizer
Brand:
IKA
YO-04737-01
More Details
StableTemp refrigerator, 12 cu ft, glass door, chart recorder, 115V
Category: StableTemp Laboratory Refrigerators
Description: StableTemp General-Purpose Laboratory Refrigerators
Brand:
StableTemp
YO-44261-02
More Details
Benchtop hood; 96" (YO-41520-30)
Category: Hamilton Constant Volume Bypass Fume Hoods
Description: Hamilton Constant Volume Benchtop Fume Hoods
Brand:
Thermo Scientific Hamilton
YO-41520-30
More Details
Sartorius Cubis Analytical Balance, 120 G X 0.1 Mg, 120VAC (MSA124S-000-DU)
Category: Semi/Micro Analytical Balances
Description: Sartorius Cubis Analytical, Semi-Micro, and Micro Balances
Brand:
Sartorius
Model Number:
MSA124S-000-DU
YO-11228-25
More Details
Labconco Purifier Class I Filtered Enclosure, 3'W, 115V
Category: Ductless Fume Hoods
Description: Labconco Purifier Class I Biosafety Enclosures
Brand:
Labconco
Model Number:
3980303
YO-33507-04
More Details
Horiba U-53G Multiparameter Meter, 6-ft (2-m) cable
Category: Handheld Multiparameter Meters
Description: Horiba U-53G Multiparameter Meter
Brand:
Horiba
YO-99600-31
More Details
PolyScience Performance Programmable 28L -30° Refrigerated Bath Ci (YO-12210-40)
Category: PolyScience Refrigerated Circulators
Description: PolyScience Refrigerated Circulating Baths
Brand:
PolyScience
YO-12210-40
More Details
PolyScience Performance Programmable 28L -30° Refrigerated Bath Ci (YO-12210-41)
Category: PolyScience Refrigerated Circulators
Description: PolyScience Refrigerated Circulating Baths
Brand:
PolyScience
YO-12210-41
More Details
64114
results
<<<Previous
66
67
68
69
70
71
72
73
74
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
10
20
30
40
50
55
56
57
58
59
60
61
62
63
64
65
66
67
68
69
70
71
72
73
74
75
76
77
78
79
80
81
82
83
84
90
100
110
120
130
140
150
160
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Fluke Calibration 7526a Precision Process Calibrator to Polyscience Performance Programmable 28l -30° Refrigerated Bath Ci (Yo-12210-41)
>
31125329-p-toc_2 31125329ptoc2