Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 8
6' Purifier Logic+ B2 Biosafety Cabinet
to
Koehler Cold Filter Plugging Point Bath
Image
Product Detail
SKU
6' Purifier Logic+ B2 Biosafety Cabinet. Energy efficient ECM motor. I (YO-33517-72)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic+ Class II Biosafety Cabinets
Brand:
Labconco
YO-33517-72
More Details
6' Purifier Logic+ B2 Biosafety Cabinet. Energy efficient ECM motor. I (YO-33517-76)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic+ Class II Biosafety Cabinets
Brand:
Labconco
YO-33517-76
More Details
Purifier Cell Logic Biosafety Cabinet; 6 ft wide; Scope-Ready and Te (YO-33516-28)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Cell Logic Class II Biosafety Cabinets
Brand:
Labconco
YO-33516-28
More Details
Nor-Lake Scientific Select™ Pass-thru Refrigerator +4C, Six door 3
Category: General-Purpose Laboratory Refrigerators
Description: Nor-Lake Scientific Select Pass-Thru Laboratory Refrigerators
Brand:
Nor-Lake
YO-44174-79
More Details
Tektronix MSO4104B-L, 1 GHz, 5/5/2.5 GS/s on 1,2, and 4 channels, 5M r
Category: Benchtop Oscilloscopes
Description: Tektronix MSO4000B Mixed Signal Phosphor Oscilloscopes
Brand:
Tektronix
Model Number:
MSO4104B-L
YO-20043-46
More Details
Tektronix MDO4034-3 Mixed Domain Oscilloscope with (4) 350 MHz analog
Category: Benchtop Oscilloscopes
Description: Tektronix MDO4000 Series Mixed Domain Oscilloscopes
Brand:
Tektronix
YO-20044-45
More Details
Tektronix MSO4102B, 1 GHz, 5/5 GS/s on 1 and 2 channels, 20M record le
Category: Benchtop Oscilloscopes
Description: Tektronix MSO4000B Mixed Signal Phosphor Oscilloscopes
Brand:
Tektronix
Model Number:
MSO4102B
YO-20043-44
More Details
6-ft Purifier® Cell Logic®+, Class II A2, 115V, Scope and Temp Zone wi
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-45
More Details
6-ft Purifier® Cell Logic®+, Class II A2, 230V (NA), Scope and Temp Zo (YO-33518-47)
Category: Biosafety Cabinets (Class II)
Description: Labconco
Purifier
®
Cell Logic+
®
Class II Biosafety Cabinets
Brand:
Labconco
YO-33518-47
More Details
Refrigerator, Blood Bank 80 cu ft with Casters, 3 Glass Doors, 230V/50 (NSBR803WWG/5)
Category: Blood Bank Refrigerators
Description: Nor-Lake Blood Bank Refrigerators
Brand:
Nor-Lake
Model Number:
NSBR803WWG/5
YO-44173-34
More Details
FreeZone -105° C PTFE 4.5 Liter Benchtop Freeze Dry System, 230V 50Hz (YO-05953-92)
Category: FreeZone Freeze Dry Systems
Description: Labconco FreeZone -105°C 4.5-L Freeze Dry System
Brand:
PTFE
YO-05953-92
More Details
FreeZone -105° C PTFE 4.5 Liter Benchtop Freeze Dry System, 230V 50Hz (YO-05953-94)
Category: FreeZone Freeze Dry Systems
Description: Labconco FreeZone -105°C 4.5-L Freeze Dry System
Brand:
PTFE
YO-05953-94
More Details
FreeZone -105° C 4.5 Liter Benchtop Freeze Dry System, PTFE, 230V 50Hz
Category: FreeZone Freeze Dry Systems
Description: Labconco FreeZone -105°C 4.5-L Freeze Dry System
Brand:
FreeZone
YO-05953-90
More Details
Electroporation system (YO-02894-04)
Category: Electroporation Systems
Description: BTX Electro Cell Fusion Systems
Brand:
BTX
YO-02894-04
More Details
Climatic Simulation Chamber, 3.6 cu ft., 200-240V
Category: Environmental Chambers
Description: Binder Climatic Simulation and Photostability Test Chambers
Brand:
Binder
Model Number:
9020-0139
YO-37756-20
More Details
Temperature And Humidity Chamber 65 L, (0 C To 120°C) 65L; 220V 60
Category: Environmental Chambers
Description: Temperature and Environmental Chambers
Brand:
Lab Companion
Model Number:
AAH82221
YO-52401-87
More Details
Temperature And Humidity Chamber 65 L, (0 C To 120°C) 65L; 230V 50
Category: Environmental Chambers
Description: Temperature and Environmental Chambers
Brand:
Lab Companion
Model Number:
AAH82222
YO-52401-90
More Details
Sanitary Wash-down Rotary Lobe Pump With Vertical Connections, 58 GPM, (LU440-YVSW1)
Category: Rotary Pumps
Description: Sanitary Washdown Rotary Lobe Pump Systems
Brand:
Cole-Parmer
Model Number:
LU440-YVSW1
YO-75201-70
More Details
Sanitary Wash-down Rotary Lobe Pump With Vertical Connections, 58 GPM, (LU440-YVSW1-230)
Category: Rotary Pumps
Description: Sanitary Washdown Rotary Lobe Pump Systems
Brand:
Cole-Parmer
Model Number:
LU440-YVSW1-230
YO-75201-72
More Details
Labconco XPert NANO Enclosures, 5-ft wide; 230 VOLT 50 HZ W/IONIZER (YO-33512-39)
Category: Balance Enclosures
Description: Labconco XPert Nano Enclosures
Brand:
Labconco
YO-33512-39
More Details
FreeZone -105° C 4.5 Liter Benchtop Freeze Dry System, PTFE, 115V 50Hz
Category: FreeZone Freeze Dry Systems
Description: Labconco FreeZone -105°C 4.5-L Freeze Dry System
Brand:
FreeZone
YO-05953-88
More Details
Purifier Cell Logic Biosafety Cabinet; 6 ft wide; Scope-Ready and Te (YO-33516-12)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Cell Logic Class II Biosafety Cabinets
Brand:
Labconco
YO-33516-12
More Details
Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 23 cub (YO-44146-26)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco ExF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-26
More Details
Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 23 cub (YO-44146-27)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco ExF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-27
More Details
Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 23 cub (YO-44146-28)
Category: Thermo Scientific Revco Ultra-Low Temperature Freezers
Description: Thermo Scientific Revco ExF Ultra-Low Temperature Freezers
Brand:
Thermo Scientific Revco
YO-44146-28
More Details
Labconco XPert NANO Enclosures, 5-ft wide; 100 VOLT 60 HZ W/IONIZER (YO-33512-37)
Category: Balance Enclosures
Description: Labconco XPert Nano Enclosures
Brand:
Labconco
YO-33512-37
More Details
Labconco XPert NANO Enclosures, 5-ft wide; 115 VOLT 60 HZ W/IONIZER (YO-33512-38)
Category: Balance Enclosures
Description: Labconco XPert Nano Enclosures
Brand:
Labconco
YO-33512-38
More Details
Medium Capacity Steam Generator; 475 Lbs/hr, 480 VAC (ES160100F3)
Category: Steam Generators
Description: Sussman Medium Capacity Electric Steam Generators
Brand:
Sussman
Model Number:
ES160100F3
YO-07601-37
More Details
Fluke 1760 Basic, Power Quality Recorder, No V or I Probes, No Trans
Category: Power Meters/Analyzers
Description: Fluke 1760 Three Phase Power Quality Recorder
Brand:
Fluke
Model Number:
FLUKE-1760 BASIC
YO-20035-75
More Details
Koehler Cold Filter Plugging Point Bath, 115 VAC, 60 Hz
Category: Petroleum Testing
Description: Koehler Mechanically Refrigerated Cold Filter Plugging Point Bath
Brand:
Koehler
YO-59876-90
More Details
64114
results
<<<Previous
4
5
6
7
8
9
10
11
12
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,138
6' Purifier Logic+ B2 Biosafety Cabinet to Koehler Cold Filter Plugging Point Bath
>
31125329-p-toc_2 31125329ptoc2