Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 90

Mettler Toledo Excellence Plus Xp Precision Toploading Balance   to   Vortex Flowmeter

Image
  Product Detail
SKU

Mettler Toledo Excellence Plus XP Precision Toploading Balance, 12,100

Category: Mettler Toledo Toploading Balances
Description: Mettler Toledo Excellence Plus XP Precision Toploading Balances
Brand: Mettler Toledo
Model Number: XP12001M


Thermo Scientific NESLAB ThermoFlex™ 1400 EPR Recirculating Chiller (YO-13042-32)

Category: Recirculators & Chillers
Description: Thermo Scientific NESLAB ThermoFlex Recirculating Chillers
Brand: Thermo Scientific


Thermo Scientific NESLAB ThermoFlex™ 1400 IPR Recirculating Chiller (YO-13042-40)

Category: Recirculators & Chillers
Description: Thermo Scientific NESLAB ThermoFlex Recirculating Chillers
Brand: Thermo Scientific


Laboratory undercounter refrigerator (single door); 5.4 CU. FT; 115V (REL-404A)

Category: Thermo Scientific Revco Laboratory Refrigerators
Description: Thermo Scientific Revco Undercounter Laboratory Refrigerators
Brand: Thermo Scientific Revco
Model Number: REL-404A


Portable Pressure Calibrator, complete hydraulic pump kit PM305-0060-K

Category: Pressure Calibrators
Description: Pressure Hand Pump Calibrator Kits
Brand: Cole-Parmer
Model Number: PM305-0010-K


Portable Pressure Calibrator, complete hydraulic pump kit PM305-0100-K

Category: Pressure Calibrators
Description: Pressure Hand Pump Calibrator Kits
Brand: Cole-Parmer
Model Number: PM305-0010-K


Portable Pressure Calibrator, complete hydraulic pump kit PM305-0150-K

Category: Pressure Calibrators
Description: Pressure Hand Pump Calibrator Kits
Brand: Cole-Parmer
Model Number: PM305-0010-K


Portable Pressure Calibrator, complete hydraulic pump kit PM305-0200-K

Category: Pressure Calibrators
Description: Pressure Hand Pump Calibrator Kits
Brand: Cole-Parmer
Model Number: PM305-0010-K


Portable Pressure Calibrator, complete hydraulic pump kit PM305-0250-K

Category: Pressure Calibrators
Description: Pressure Hand Pump Calibrator Kits
Brand: Cole-Parmer
Model Number: PM305-0010-K


Portable Pressure Calibrator, complete hydraulic pump kit PM305-0300-K

Category: Pressure Calibrators
Description: Pressure Hand Pump Calibrator Kits
Brand: Cole-Parmer
Model Number: PM305-0010-K


Portable Pressure Calibrator, complete hydraulic pump kit PM305-0350-K

Category: Pressure Calibrators
Description: Pressure Hand Pump Calibrator Kits
Brand: Cole-Parmer
Model Number: PM305-0010-K


Portable Pressure Calibrator, complete hydraulic pump kit PM305-0400-K

Category: Pressure Calibrators
Description: Pressure Hand Pump Calibrator Kits
Brand: Cole-Parmer
Model Number: PM305-0010-K


Portable Pressure Calibrator, complete hydraulic pump kit PM305-0450-K

Category: Pressure Calibrators
Description: Pressure Hand Pump Calibrator Kits
Brand: Cole-Parmer
Model Number: PM305-0010-K


Portable Pressure Calibrator, complete hydraulic pump kit PM305-0500-K

Category: Pressure Calibrators
Description: Pressure Hand Pump Calibrator Kits
Brand: Cole-Parmer
Model Number: PM305-0010-K


Portable Pressure Calibrator, complete hydraulic pump kit PM305-0550-K

Category: Pressure Calibrators
Description: Pressure Hand Pump Calibrator Kits
Brand: Cole-Parmer
Model Number: PM305-0010-K


Portable Pressure Calibrator, complete hydraulic pump kit PM305-0600-K

Category: Pressure Calibrators
Description: Pressure Hand Pump Calibrator Kits
Brand: Cole-Parmer
Model Number: PM305-0010-K


Portable Pressure Calibrator, complete hydraulic pump kit PM305-0650-K

Category: Pressure Calibrators
Description: Pressure Hand Pump Calibrator Kits
Brand: Cole-Parmer
Model Number: PM305-0010-K


Portable Pressure Calibrator, complete hydraulic pump kit PM305-0700-K

Category: Pressure Calibrators
Description: Pressure Hand Pump Calibrator Kits
Brand: Cole-Parmer
Model Number: PM305-0010-K


Portable Pressure Calibrator, complete hydraulic pump kit PM305-0050-K

Category: Pressure Calibrators
Description: Pressure Hand Pump Calibrator Kits
Brand: Cole-Parmer
Model Number: PM305-0010-K


Thermo Scientific Barnstead Smart2Pure 3 l/h UV

Category: Water Purification
Description: Thermo Scientific Barnstead Smart2Pure Water Purification System
Brand: Barnstead


Thermo Scientific Barnstead MicroPure UV/UF

Category: EASYpure Water Purification Systems
Description: Thermo Scientific Barnstead MicroPure Water Purification Systems
Brand: Thermo Scientific Barnstead


Labconco Purifier HEPA Filtered Enclosure, 2'W, 230V

Category: Ductless Fume Hoods
Description: Labconco Purifier HEPA-filtered Enclosures
Brand: Labconco
Model Number: ENCLOSURE HEPA 2W 230V


Thermo Scientific Thermolyne Benchtop Muffle Furnace with 350 cu in ch (F48020-33)

Category: Thermo Scientific Muffle Furnaces
Description: Thermo Scientific Thermolyne Benchtop Muffle Furnaces
Brand: Thermo Scientific Thermolyne
Model Number: F48020-33


Front View Distillation Apparatus, Group 4, right side, 220 VAC

Category: Koehler Front View Distillation Apparatus
Description: Front View Distillation Apparatus, Group 4
Brand: Koehler
Model Number: K45290


GE (Druck) DPI 620 Multifuction Calibrator (Now capable of HART Commun

Category: Calibration Equipment
Description: GE (Druck) DPI 620 Multifunction Calibrator
Brand: Druck
Model Number: DPI 620


Casella CEL-630.C1/K1 Type 1 Third Octave Sound Meter Kit

Category: Sound Meters
Description: Casella Wide-Range Sound Level Meters
Brand: Casella
Model Number: CEL-630.C1/K1


A&D AD-4212A Precision Weighing Sensor, 110g X 0.1mg 230 VAC

Category: A&D Weighing Industrial Scales
Description: A&D Weighing AD-4212A Precision Weighing Sensor
Brand: A&D Weighing
Model Number: AD-4212-100+230VAC


A&D AD-4212A Precision Weighing Sensor, 1100g X 0.001g 230 VAC

Category: A&D Weighing Industrial Scales
Description: A&D Weighing AD-4212A Precision Weighing Sensor
Brand: A&D Weighing
Model Number: AD-4212-1000+230VAC


Babcock 220V, 50Hz centrifuge; 24 bottle capacity (YO-17360-25)

Category: Babcock System for Fat Analysis
Description: Centrifuges for Babcock system
Brand: Cole-Parmer


Vortex Flowmeter, 4" Flange Connection, for Steam

Category: Vortex Flowmeters
Description: ABB Vortex Flowmeters
Brand: ABB
Model Number: VT4.3.1.3.1.1H.Q.A.2.3.A.2.E.X.0.A.B




Mettler Toledo Excellence Plus Xp Precision Toploading Balance to Vortex Flowmeter

>