Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 900

Replacement Sample Chamber for Spindles 98945-81   to   Cole-Parmer Tuff-Tip

Image
  Product Detail
SKU

Replacement sample chamber for spindles 98945-81, -82, -83, -84, -85, (SC4-13R)

Category: Accessories for Brookfield Rotational Viscometers
Description: Sample Chambers for Spindles for Temperature Control Unit and Small Sample Adapters
Brand: Lab Products
Model Number: SC4-13R


Lightnin® (A-310) 316 SS high-efficiency axial flow impeller, 3.8" (871581PSP)

Category: Accessories for Industrial Mixers
Description: High-Efficiency Axial Flow Impellers (Lightnin® A-310™)
Brand: Lightnin
Model Number: 871581PSP


Convoluted FEP-lined hose assembly, 3/4" hex male, 1-ft length (YO-96550-03)

Category: Hose
Description: Chemfluor Convoluted FEP-Lined Hose Assemblies
Brand: Chemfluor


ISE electrode, fluoride, for controller 56105-00 (YO-27077-06)

Category: Cole-Parmer® Ion Concentration Controller
Description: Cole-Parmer® Ion-Selective Electrodes
Brand: Cole-Parmer


Cylinder, 2000 ml, Class A , To Deliver, Certificate of Traceability (CP08557H)

Category: Glass Graduated Cylinders
Description: Cole-Parmer Serialized Class A Graduated Glass Cylinders
Brand: Cole-Parmer
Model Number: CP08557H


Pyrex Brand 1000 Griffin Low-Form Beaker, 1000 mL, 24/cs (1000-1L)

Category: Glass Beakers
Description: PYREX Brand 1000 Glass Beakers - Griffin, low form
Brand: Corning
Model Number: 1000-1L


Starter Kit, Includes FL1E-H12RCE base unit, software, programming cab

Category: Automation Control
Description: idec Programmable Smart Relay
Brand: Idec
Model Number: SMARTSTART-HDC-E


Easy-read NIST Certified Thermometer, -10/225c, 355mm Length, Total Im (30520C)

Category: 100% Non-Hazardous Liquid Filled Glass Thermometers
Description: H-B Instrument Easy-Read Enviro-Safe Nonhazardous Glass Thermometers Traceable to NIST, Celsius-scale
Brand: H-B Instrument Company
Model Number: 30520C


Latex test for Salmonella (YO-36151-02)

Category: Bacteria test kits
Description: Rapid Bacterial Test Kits
Brand: Cole-Parmer


Smooth PTFE-lined hose assembly, 1/2" JIC female x 1/2" male, 10-ft le (YO-96554-13)

Category: Hose
Description: Chemfluor Smooth PTFE-Lined Hose Assemblies
Brand: Chemfluor


15" (380 mm) X 24" (600 mm) X 8.3" (210 mm) compact enclosure.

Category: Wall Mount Enclosures, Cabinets, & Racks
Description: Compact Wall-mount Enclosures
Brand: Cole-Parmer
Model Number: 1038.500


6x6x4 Type 316L Junction Box, Stainless Steel (JB060604H6)

Category: Junction Boxes
Description: Junction Boxes -Type 316L Stainless Steel (Sizes ranging from: 6x6x4 to 16x14x6)
Brand: Automation Control
Model Number: JB060604H6


Digital multimeter/infrared thermometer with laser sighting Dot/Circle (HHM290)

Category: Newport Digital Multimeters
Description: Newport Supermeter Multimeter
Brand: Newport
Model Number: HHM290


PAXT0000, Temperature Panel Meter Thermocouples- T, E, J, K, R, S, B,

Category: Panel Meters
Description: Red Lion PAX Panel Meters
Brand: Red Lion
Model Number: PAXT0000


Masterflex Norprene tubing (A 60 G), I/P 88, 25 ft.

Category: Masterflex Norprene Pump Tubing Formulation
Description: I/P High-Performance Precision Norprene (A 60 G) Pump Tubing for Masterflex I/P Pumps
Brand: Masterflex


Mini containers; 12 to 37 mL capacity

Category: Waring Blender Accessories
Description: Waring One-Liter Blenders Special Containers
Brand: Waring Products
Model Number: MC-1


96-Well Caps for Thermo Scientific Nunc™ 96-Well Microplates, Steri (YO-01929-54)

Category: General Accessories
Description: Replacement Accessories
Brand: Thermo Scientific Nunc


Polysulfone Ultrapure Flowmeter for Water, 0.1 to 1.2 GPM (F-46010LX-08P)

Category: Polysulfone In-Line Direct Reading Flowmeters
Description: Ultrapure In-Line Flowmeters for Liquids
Brand: Cole-Parmer
Model Number: F-46010LX-08P


Smooth PTFE-lined hose assembly, 1" JIC female x 1" JIC female, 1-ft l (YO-96553-05)

Category: Hose
Description: Chemfluor Smooth PTFE-Lined Hose Assemblies
Brand: Chemfluor


Particle Tolerant Solenoid Valve, three-way, 12 VDC, 2.0 mm (MTV-3K.25U-G-12V)

Category: Solenoid Valves
Description: Particle Tolerant Solenoid Valves
Brand: Cole-Parmer
Model Number: MTV-3K.25U-G-12V


Cole-Parmer 65-mm Correlated Flowmeter , 316 SS Glass float; For Liqui (YO-03218-37)

Category: Cole-Parmer 65-mm Correlated Flowmeters
Description: High-Flow Correlated Flowmeters with Standard Valves
Brand: Cole-Parmer


Polysulfone Ultrapure Flowmeter for Water, 0.2 to 2 GPM (F-46020LX-08P)

Category: Polysulfone In-Line Direct Reading Flowmeters
Description: Ultrapure In-Line Flowmeters for Liquids
Brand: Cole-Parmer
Model Number: F-46020LX-08P


Particle Tolerant Solenoid Valve, three-way, 24 VDC, 2.0 mm (MTV-3K.25U-G-24V)

Category: Solenoid Valves
Description: Particle Tolerant Solenoid Valves
Brand: Cole-Parmer
Model Number: MTV-3K.25U-G-24V


Cole-Parmer Tuff-Tip, Semi-Domed 1" In-Line or Submersible Graphite Ti (YO-27020-62)

Category: Cole-Parmer In-Line or Submersible, Tuff-Tip, Semi-Domed, pH Electrodes with Ceramic Junctions
Description: Cole-Parmer Tuff-Tip, Semi-Domed 1
Brand: Cole-Parmer


Cole-Parmer 65-mm Correlated Flowmeter , 316 SS SS float; For Liquids (YO-03218-39)

Category: Cole-Parmer 65-mm Correlated Flowmeters
Description: High-Flow Correlated Flowmeters with Standard Valves
Brand: Cole-Parmer


A&D Portable Balance, 610 g x 0.1 g (SV-610)

Category: AND Engineering Portable Balances
Description: AND Weighing Portable Balances
Brand: A&D Weighing
Model Number: SV-610


PAXP0000, Process Input Panel Meter 0 to 20mA, 0 to 10V DC 85-250VAC

Category: Panel Meters
Description: Red Lion PAX Panel Meters
Brand: Red Lion
Model Number: PAXP0000


Cole-Parmer 65-mm Correlated Flowmeter , 316 SS Carboloy float; For L (YO-03218-41)

Category: Cole-Parmer 65-mm Correlated Flowmeters
Description: High-Flow Correlated Flowmeters with Standard Valves
Brand: Cole-Parmer


Hyrdronic Specialty, 7.6 Gallon volume, 2.4 gallon acceptance Diaphrag

Category: Centrifugal Pump Accessories
Description: Armstrong Hydronic Specialties
Brand: Armstrong Hydronic
Model Number: 572018-003


Cole-Parmer Tuff-Tip, Semi-Domed 1" In-Line or Submersible Graphite Ti (YO-27020-66)

Category: Cole-Parmer In-Line or Submersible, Tuff-Tip, Semi-Domed, pH Electrodes with Ceramic Junctions
Description: Cole-Parmer Tuff-Tip, Semi-Domed 1
Brand: Cole-Parmer




Replacement Sample Chamber for Spindles 98945-81 to Cole-Parmer Tuff-Tip

>