Subtotal =  
Items =  
View Cart
Welcome

Product Index Page 1

10.0   to   Thermo Scientific Nalgene® Straight-Sided Polypropylene Wide-Mouth (2118-0002)

Image
  Product Detail
SKU

10.0 mL Measuring Pipettes (7070-10)

Category: Glass Pipettes
Description: PYREX® Serialized/Certified, Class A, Measuring (Mohr) Pipettes
Brand: Pyrex
Model Number: 7070-10


C-FLEX® tubing, 1/8"ID x 1/4"OD, 25 ft/pack (YO-06424-67)

Category: Flexible Tubing
Description: C-FLEX® Tubing
Brand: C-FLEX


Flowmeter,316SS,150mm (YO-32466-14)

Category: Cole-Parmer Easy-View Correlated Flowmeters
Description: Cole-Parmer Easy-View Correlated Flowmeters - 150-mm
Brand: Cole-Parmer


Oakton® conductivity solution, 1413 µS

Category: Conductivity Calibration Solutions
Description: Oakton® Conductivity Calibration Solutions
Brand: Oakton


Thermo Scientific Nalgene Quick Filling/Venting Closures, PP, 83b 2-po (2158-0021)

Category: Carboys
Description: Thermo Scientific Nalgene Quick Filling/Venting Closures, PP
Brand: Thermo Scientific
Model Number: 2158-0021


50mm in-line filter, PTFE, 0.2µm, 10/pk (6720-5002)

Category: In-Line Filters
Description: Whatman 50 mm In-Line Filters
Brand: Cole-Parmer
Model Number: 6720-5002


Cole-Parmer Polypropylene Stir Rods, 10" x 10mm, 12/pk (YO-06369-42)

Category: Spatulas & Stir Rods
Description: Cole-Parmer Stir Rods, PP
Brand: Cole-Parmer


316 Stainless Steel three-blade propeller, 3" dia x 5/16" bore dia (YO-04552-60)

Category: Laboratory Mixer Accessories
Description: Stir-Pak and Servodyne Mixer Accessories
Brand: Stir-Pak


LaMotte® Dissolved Oxygen Test Kit Refill; Winkler Titration Metho

Category: General Accessories
Description: Replacement Accessories
Brand: LaMotte
Model Number: R-5860


Male luer lock x 1/4" NPT, 316 SS, each (YO-31507-45)

Category: Metal Luer Fittings
Description: 316 SS Fittings
Brand: Cole-Parmer


Oakton® pH 11 meter with "All-in-One" electrode (35811-71)

Category: pH/Ion Handheld Meters
Description: Oakton® pH 11 Handheld pH Meters
Brand: Oakton
Model Number: pH 11


CAB-O-SIL® M-5, scintillation grade (150g) (YO-88341-32)

Category: Ca to Calycot
Description: CAB-O-SIL ®  M-5
Brand: CAB-O-SIL


Deep depth backsaver soil sampling kit (PN134)

Category: Soil Auger and Sampling Kits
Description: Sampling Kits
Brand: Cole-Parmer
Model Number: PN134


303/304 SS Three-blade Paddle Assembly; 1/4 (YO-04350-00)

Category: 303/304 Stainless Steel Mixing Paddle Assemblies
Description: Three-Blade Paddle Assemblies
Brand: Cole-Parmer


Thermo Scientific Nalgene Autoclavable plastic desiccator; 140mm plate (5315-0150)

Category: Non Vacuum Desiccators
Description: Thermo Scientific Nalgene Autoclavable Plastic Desiccator
Brand: NALGE NUNC INTERNATIONAL
Model Number: 5315-0150


Tetrabutylammonium hydroxide, 0.1N solution in toluene/methanol for no (YO-88122-32)

Category: Tertraa to Tetrae
Description: Tetrabutylammonium hydroxide
Brand: Hydroxide


PVC Check Valve, 1/4" NPT(F) connections (CKD025V-PV)

Category: Check Valves
Description: Check Valves with Pipe Thread Connections
Brand: Cole-Parmer
Model Number: CKD025V-PV


Calibration fluid for Thermo Scientific NanoDrop (CHEM-CF1)

Category: Accessories for Spectroscopy
Description: Thermo Scientific NanoDrop Spectrophotometer and Fluorometer Accessories
Brand: Thermo Scientific NanoDrop
Model Number: CHEM-CF1


Thermo Scientific SPECTRONIC™ standards, set of 10 (333150-000)

Category: General Accessories
Description: Replacement Accessories
Brand: Spectronic
Model Number: 333150-000


Masterflex® L/S® Easy-Load® II pump head for precision tub (YO-77201-60)

Category: L/S® Easy-Load® II Pump Heads
Description: L/S® Easy-L
Brand: Masterflex


Qorpak® Kaptclean™ pre-cleaned sample bottle, 1 liter

Category: Environmental Bottles
Description: Qorpak® Kaptclean Sample Bottles, HDPE
Brand: Qorpak
Model Number: 0300-0020


Hach Total Chlorine (USEPA) Test Kit (21056-69)

Category: Colorimetry Reagents
Description: Hach® Colorimetry Test Kits
Brand: Hach
Model Number: 21056-69


Thermo Scientific Lindberg® Blue M® Muffle Furnace, 2592 cu in (BF51828C)

Category: Thermo Scientific Muffle Furnaces
Description: Thermo Scientific Lindberg Blue M Muffle Furnaces
Brand: Thermo Scientific Lindberg Blue M
Model Number: BF51828C


Pyrex® Brand 9825 culture tube with screw cap; 50 mL, pack of 48 (9825-22)

Category: Test Tubes
Description: Pyrex® Brand 9825 Culture Tubes
Brand: Pyrex
Model Number: 9825-22


Masterflex PharMed BPT Tubing, L/S #15, 25' (YO-06508-15)

Category: Tubing
Description: Masterflex
Brand: Masterflex


Thermo Scientific Barnstead EASYpure® II UV Water Purification Sys (D7401)

Category: EASYpure® Water Purification Systems
Description: Thermo Scientific Barnstead EASYpure® II UV Water Purification System
Brand: EASYPure
Model Number: D7401


Thermo Scientific Nalgene® economy low-density polyethylene wash b (2401-0250)

Category: Wash Bottles
Description: Thermo Scientific Nalgene® Economy LDPE Wash Bottles
Brand: Thermo Scientific
Model Number: 2401-0250


Vanadate-Molybdate Reagent, for Phosphorus Analysis (500 mL) (YO-86977-09)

Category: Chemicals Va to Vz
Description: Vanadate-Molybdate Reagent, for Phosphorus Analysis
Brand: Cole-Parmer


Thermo Scientific Nalgene ®  Unitary Dropper, LDPE, 100/bag (6219-0068)

Category: Plastic Pipettes
Description: Thermo Scientific Nalgene® Unitary Dropper
Brand: Nalgene
Model Number: 6219-0068


Thermo Scientific Nalgene® straight-sided polypropylene wide-mouth (2118-0002)

Category: Jars
Description: Polypropylene jars
Brand: Thermo Scientific
Model Number: 2118-0002


80836 results


10.0 to Thermo Scientific Nalgene® Straight-Sided Polypropylene Wide-Mouth (2118-0002)