Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 100
Iris Tissue Forcep
to
Rectangular Tank with Cover
Image
Product Detail
SKU
Iris Tissue Forcep, Fully Curved, Precision-grade
Category: Miltex Tissue and Dressing Forceps
Description: Miltext Precision-grade Forceps
Brand:
Miltex
YO-10923-06
More Details
Handle for crucible racks, stainless steel with insulated grip. Measur
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
50495
YO-17803-70
More Details
Whatman™ 50 mm In-Line Filters, PTFE, 0.2µm, 10/pk
Category: In-Line Filters
Description: Whatman 50 mm In-Line Filters
Brand:
Whatman
YO-29700-72
More Details
Compression male pipe adapter elbow, 1/2" NPT(M) x 1/2" OD, brass
Category: Male Threaded Adapter Elbows
Description: Brass Compression to NPT(M) Adapter Elbows
Brand:
Cole-Parmer
YO-31412-88
More Details
Thermo Scientific Nalgene Narrow-Mouth Economy Bottle, HDPE, 60 mL, 12 (YO-06035-11)
Category: Thermo Scientific Nalgene Narrow-Mouth Bottles
Description: Thermo Scientific Nalgene Narrow-Mouth Economy Bottles, HDPE
Brand:
Thermo Scientific Nalgene
YO-06035-11
More Details
Label for centrifuge and Eppendorf tubes (YO-65543-12)
Category: Label Printers
Description: Labels for TLS PC Link and IdentiLab Software
Brand:
Eppendorf
YO-65543-12
More Details
5180 Gaussmeter W/4in Probe Ac Adapter Hard Carry Case (5180)
Category: Gauss Meters
Description: Pacific Scientific OECO Gauss - Tesla Meters
Brand:
FW Bell
Model Number:
5180
YO-20001-29
More Details
High-Intensity, long-wave UV lamp exposure box (76-0055-01)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
76-0055-01
YO-09819-29
More Details
Thermo-Dynamic steam trap, 3/4"NPT(F) (TD52 3/4" 54531C)
Category: Spirax Sarco Thermo-Dynamic Steam Traps
Description: Thermo-Dynamic steam traps
Brand:
Spirax Sarco
Model Number:
TD52 3/4" 54531C
YO-07600-06
More Details
Heated Evaporator/Concentrator; single block with heated manifold gas (YO-28690-35)
Category: Evaporative Sample Concentrator
Description: Cole-Parmer Evaporator/Concentrators
Brand:
Cole-Parmer
YO-28690-35
More Details
Scienceware test tube rack for 10- to 13-mm tubes
Category: Tube Racks
Description: Scienceware Test Tube Racks
Brand:
Scienceware
YO-06737-10
More Details
Immersion heater, short rod, 5.5" L heated area, 500 watts, 115 VAC (324-114)
Category: 316 Stainless Steel Heaters
Description: Heat-O-Matic Immersion Heaters
Brand:
Heat-O-Matic
Model Number:
324-114
YO-03046-00
More Details
Fluke CNX C3002 Soft Case for Wireless Digital Multimeter
Category: Fluke Digital Multimeters
Description: Fluke CNX Series Wireless Systems Accessories
Brand:
Fluke
YO-20044-08
More Details
Push/Pull Force Gauge 2 X .020 Lbs Capacity
Category: Force Gauges
Description: Wagner Force Dials
Brand:
WAGNER INSTRUMENTS
Model Number:
FDK-2
YO-93949-54
More Details
Thermo Scientific Orion pH/ATC Triode, epoxy body, sealed, single-junc (9107BN)
Category: pH/Ion Laboratory Electrodes
Description: Thermo Scientific Orion Standard Ag/AgCl pH Electrodes
Brand:
Thermo Scientific Orion
Model Number:
9107BN
YO-58819-91
More Details
Square polycarbonate container with handles, 12 quart
Category: Plastic Containers
Description: Square Containers with Handles
Brand:
Cole-Parmer
Model Number:
1072407
YO-63570-50
More Details
Pipette filler, safety bulb
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Hach
Model Number:
14651-00
YO-99574-53
More Details
Low-Cost Utility Pressure Gauge, 0-30" Hg (KCCP7211-2.5-4-30/0)
Category: Mechanical Pressure Gauges
Description: Quality Utility Gauges
Brand:
Cole-Parmer
Model Number:
KCCP7211-2.5-4-30/0
YO-68356-00
More Details
Air Compressor hose, 1/4 in x 100' PU braid air hose with swivel fitti (AA1064-blue)
Category: Accessories for Compressors - Air Equipment
Description: Air Hose Assemblies with Swivel Fitting and Grip
Brand:
Cole-Parmer
Model Number:
AA1064-blue
YO-07123-28
More Details
H-B Instrument Battery Hydrometer with Siphon Set
Category: Hydrometers Special Applications
Description: H-B Instrument Battery Hydrometer with Siphon Set
Brand:
H-B Instrument Company
Model Number:
50860
YO-08299-43
More Details
Millipore Steriflip Vacuum-Driven Filter Device with Millipore Express
Category: Centrifugal Devices and Filters
Description: Millipore Steriflip Vacuum-Driven Filter Devices
Brand:
Millipore
YO-29969-20
More Details
Sieve Calibration Standard, mesh 10
Description: Sieve Calibration Standards
Brand:
Cole-Parmer
YO-59945-01
More Details
Barbed Reducing Y-connector, 3/4 X 1, Nylon, Pack Of 5 (RY12-16NN (5 PACK))
Category: Barbed to barbed connectors
Description: Barbed reducing Y connectors
Brand:
Cole-Parmer
Model Number:
RY12-16NN (5 PACK)
YO-30726-37
More Details
1/2" NPT brass female/male mini manual ball valve (MV608-8)
Category: Parker Metal Industrial Ball Valves
Description: Brass Female/Male Mini Manual Ball Valves
Brand:
Cole-Parmer
Model Number:
MV608-8
YO-30533-26
More Details
Wildco Sedwick Rafter Counting Cell (1801-A10)
Category: Open Water Samplers
Description: Wildco Sedgewick Rafter Counting Cells
Brand:
Cole-Parmer
Model Number:
1801-A10
YO-05491-60
More Details
Microscope/telescopes with built-in reticle; magnification, 35x/8x; fi (M820-C)
Category: Microscope/Telescopes (Dual-Purpose)
Description: Microscope/Telescopes with Built-In Reticles
Brand:
Specwell Corporation
Model Number:
M820-C
YO-03098-00
More Details
Tag, Out Of Service, 5 7/8" X 3 1/8", PF-Cardstock (TRS243CTP)
Category: Status Tags
Description: Accident Prevention Tags; Status Tags; Out Of Service
Brand:
Accuform
Model Number:
TRS243CTP
YO-50503-28
More Details
Leeson NEMA Type 56C-face TEFC motor, 1/3 hp, 1750 rpm, 90 VDC
Category: DC Motors
Description: Leeson NEMA Type 56C-Face DC Motors
Brand:
Leeson
Model Number:
098004
YO-70071-00
More Details
Stainless steel perforated tray, 4-1/8" D, fits tank 07240-10, 07240-9 (YO-07277-54)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
YO-07277-54
More Details
Rectangular Tank with Cover, HDPE, 90 Gal. (YO-06323-63)
Category: Plastic Tanks and Drums
Description: Rectangular Tanks with Covers
Brand:
Food and Beverage
YO-06323-63
More Details
64114
results
<<<Previous
96
97
98
99
100
101
102
103
104
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
10
20
30
40
50
60
70
80
85
86
87
88
89
90
91
92
93
94
95
96
97
98
99
100
101
102
103
104
105
106
107
108
109
110
111
112
113
114
120
130
140
150
160
170
180
190
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Iris Tissue Forcep to Rectangular Tank with Cover
>
31125329-p-toc_2 31125329ptoc2