Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 12
Male Luer Lock X 1
to
Hach Cod Test Kit
Image
Product Detail
SKU
Male luer lock x 1/4" NPT, 316 SS, each (YO-31507-45)
Category: Metal Luer Fittings
Description: 316 SS Fittings
Brand:
Cole-Parmer
YO-31507-45
More Details
Filter Bags for Stomacher 400C, round-bottom bag with filter, 200 Per
Category: Paddle Blender Accessories
Description: Seward Stomacher Sterile Blending Bags
Brand:
Seward Laboratory Systems Inc
YO-36860-01
More Details
Ionic strength and pH adjustors, TISAB II (940909)
Category: Ion-Selective Electrodes Solutions
Description: Ionic Strength and pH Adjusters for Orion Ion-Selective Electrodes
Brand:
Thermo Scientific Orion
Model Number:
940909
YO-05730-15
More Details
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 10 ml, 12 (99502-10)
Category: Centrifuge Tubes
Description: Corning Conical-Bottom Disposable Glass Centrifuge Tubes
Brand:
Corning
Model Number:
99502-10
YO-34534-08
More Details
Glass Vials PTFE Lined Cap Clear 8ml 200/pk (B7800-3)
Category: Sample Glass Vials
Description: Class A Sample Glass Vials
Brand:
PTFE
Model Number:
B7800-3
YO-08922-23
More Details
Thermo Scientific Nalgene Centrifuge Bottle; 250-mL; PPCO; 60.7 mm dia
Category: Centrifuge Bottles
Description: Thermo Scientific Nalgene 250-mL Centrifuge Bottles
Brand:
Thermo Scientific Nalgene
Model Number:
3141-0250
YO-06106-14
More Details
Fluke 116/322 HVAC Multimeter and Clamp Meter Combo Kit [FL116322]
Category: Fluke Digital Multimeters
Description: Fluke 116 HVAC Service Digital Multimeter with Temperature
Brand:
Fluke
Model Number:
FLUKE-116/322
YO-26048-85
More Details
Pipe fittings, Male reducers NPT x NPT, Nylon, 3/4" x 3/8", 1-3/8", 1- (YO-31902-00)
Category: Reducer nipples
Description: Reducer nipples, NPT(M) x NPT(M)
Brand:
Cole-Parmer
YO-31902-00
More Details
Marble Balance Table (TABLE)
Category: Antivibration Platforms for Balances
Description: Balance Table with Marble
Brand:
Cole-Parmer
Model Number:
TABLE
YO-11500-00
More Details
Gas sampling bulb, glass, 250 ml (653150-0250)
Category: Sampling Accessories
Description: Glass Gas Sampling Bulb
Brand:
Cole-Parmer
Model Number:
653150-0250
YO-10370-01
More Details
Ansell Neoprene Apron, 35" x 45", yellow
Category: Aprons
Description: Ansell Neoprene Aprons
Brand:
Ansell
YO-86479-03
More Details
0 to 3000 psi and 0 to 200 kg/cm2 Dual-Scale Gauge, Back Co
Category: Mechanical Pressure Gauges
Description: Ashcroft Dual-Scale Gauges
Brand:
Ashcroft
Model Number:
1008SL2.5
YO-68007-31
More Details
Millipore Amicon Centrifugal Filter Units, 100k NMWL, 24/pk, blue
Category: Centrifugal Devices and Filters
Description: Millipore Amicon Centrifugal Filter Units
Brand:
Millipore Amicon
Model Number:
42412
YO-29950-54
More Details
Biotest ErgoTouch Airborne Particle Counter
Category: Air Sampling
Description: Biotest Ergotouch Airborne Particle Counters
Brand:
Cole-Parmer
YO-18450-20
More Details
Glass Fiber Filters; 55mm dia; pore size, 1.6µ; thickness, 0.22mm; (GA5555MM)
Category: Filter Papers
Description: Glass Fiber Filters
Brand:
Advantec
Model Number:
GA5555MM
YO-06646-03
More Details
Pipe fittings, Nipple, 3/4" NPT(M), PVDF; 5/pack
Category: Threaded nipples
Description: Nipples, NPT(M)
Brand:
Cole-Parmer
YO-30706-04
More Details
Thermo Scientific SC 100 Digital Immersion Circulator, 100°C w/cla (SC 100) (YO-12135-00)
Category: Immersion Circulators
Description: Thermo Scientific Heating Immersion Circulators
Brand:
Thermo Scientific
Model Number:
SC 100
YO-12135-00
More Details
WTW ProfiLine pH 3210 meter only
Category: pH/Ion Handheld Meters
Description: WTW ProfiLine pH 3210 Handheld pH Meters
Brand:
WTW
Model Number:
pH 3210
YO-58890-10
More Details
IKA RCT Basic IKAMAG Digital Round-Top Stirring Hot Plate, 115V
Category: Digital Stirrers
Description: IKA RCT and RET basic IKAMAG Round Top Digital Stirring Hot Plates
Brand:
IKA
Model Number:
3810001
YO-04671-21
More Details
AC Variable-Speed Controller, 1/3 hp, 115/230 VAC, 1.7 amp, 120-240VAC
Category: AC Controllers
Description: Single-Phase Input Variable-Speed Controllers
Brand:
Automation Control
Model Number:
ESV251N01SXB
YO-70101-01
More Details
Thermo Scientific Barnstead Final Filter compatible with E-pure™, N (D3750)
Category: Deionization Filters
Description: Thermo Scientific Barnstead Final Filters for Deionization Systems
Brand:
E-pure
Model Number:
D3750
YO-99256-00
More Details
Hycon RCS Air Sampler
Category: Air Sampling
Description: Biotest Hycon RCS Air Sampler
Brand:
RCS
Model Number:
940015
YO-86509-00
More Details
WTW ProfiLine pH 3210 meter with Sentix 41 probe
Category: pH/Ion Handheld Meters
Description: WTW ProfiLine pH 3210 Handheld pH Meters
Brand:
WTW
YO-58890-12
More Details
Temperature Controller, TC, 1/16-DIN, Relay Output, 2 alarms (DC1010CT-102-000-E)
Category: Controllers-Panel Mount
Description: Honeywell DC 1000-Series Temperature/Process Controllers
Brand:
Honeywell
Model Number:
DC1010CT-102-000-E
YO-89556-00
More Details
Cannon Cannon-Fenske routine viscometer, size 150
Category: Cannon Glass Capillary Viscometers
Description: Cannon-Fenske Routine Viscometers
Brand:
Cannon
Model Number:
9721-B62
YO-98934-54
More Details
Sanitary Pressure Gauge, 0-100 PSI, 3/4 Tri-clamp
Category: Sanitary Gauges, Transducers, and In-Line Seals
Description: Wika 3A Sanitary Gauge
Brand:
Wika
Model Number:
M932.25
YO-68056-76
More Details
Threaded adapter, stainless steel, 3/4" female NPT to 3/4" male BSPP (YO-31830-54)
Category: British Standard Pipe Tapered Threaded (BSPT) Adapters
Description: British Standard Pipe Threaded (BSPP) Adapters
Brand:
Parker Hannifin
YO-31830-54
More Details
Quest Technologies Type 2 Sound Meter, basic (2100)
Category: Sound Meters
Description: Quest Technologies
®
Intrinsically Safe Sound Meters
Brand:
Quest Technologies
Model Number:
2100
YO-86346-00
More Details
Threaded adapter, stainless steel, 1/4" female BSPT to 1/4" male NPT (YO-31830-61)
Category: BSP Adapters
Description: British Standard Pipe Tapered Threaded (BSPT) Adapters
Brand:
Parker Hannifin
YO-31830-61
More Details
Hach COD Test Kit, Low Range, 0 to 150 ppm
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Hach
YO-99574-25
More Details
64114
results
<<<Previous
8
9
10
11
12
13
14
15
16
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Male Luer Lock X 1 to Hach Cod Test Kit
>
31125329-p-toc_2 31125329ptoc2