Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 13
Thermo Scientific Heraeus Fresco™ 21 Refrigerated Microcentrifuge W (75002426)
to
Thermo Scientific Finntip 10 Ml Sterile Pipette Tip
Image
Product Detail
SKU
Thermo Scientific Heraeus Fresco™ 21 Refrigerated Microcentrifuge w (75002426)
Category: Microcentrifuges
Description: Thermo Scientific Heraeus Pico and Fresco Microcentrifuges
Brand:
Thermo Scientific Heraeus
Model Number:
75002426
YO-17703-30
More Details
Activate™ High-Level Chlorine Test Strips 50/PK (77085)
Category: Water Quality Test Strips
Description: Activate High-Level Chlorine Test Strips
Brand:
Cole-Parmer
Model Number:
77085
YO-18105-01
More Details
Gems Sensors Flow Indicator for Liquids; Brass, 1/4" NPT(F), 0.5 to 5.
Category: Cole-Parmer Flow Indicators for Liquids
Description: Gems Sensors Flow Indicators
Brand:
Gems
YO-32779-04
More Details
Labpette Discovery Pipette 100- 1000 uL (P3940-1000)
Category: Single-Channel Pipettors
Description: Labpette Discovery Pipettes
Brand:
LABPETTE
Model Number:
P3940-1000
YO-24601-10
More Details
Barbed fittings, T connector, Clear PP, 1/16" ID, 1/32", 25/32", 15/32 (YO-30506-15)
Category: Barbed to barbed connectors
Description: Barbed T connectors
Brand:
Cole-Parmer
YO-30506-15
More Details
Professional series air compressor, 7.1 cfm, 20 gallon portable tank,
Category: Air Compressors
Description: Single-Stage Air Compressors with Storage Tanks
Brand:
Cole-Parmer
Model Number:
PK5020
YO-07121-05
More Details
SalvisLab Thermocenter Oven, 40 L capacity, TC40, 115 VAC
Category: Mechanical Convection Ovens
Description: Salvis Thermocenter Mechanical Convection Ovens
Brand:
Salvis
Model Number:
31A04100
YO-52403-00
More Details
Contec Prosat Presaturated cleanroom wipes, 300/pack
Category: Clean Room Wipes
Description: Contec Prosat Presaturated Cleanroom Wipes
Brand:
Contec
Model Number:
PS-911, PROSAT
YO-33675-00
More Details
Ohaus Triple-Beam Balance with Tare and Deep Stainless Steel Pan, 610
Category: Mechanical Balances
Description: Ohaus Triple-Beam Mechanical Balances
Brand:
Ohaus
Model Number:
710-TO
YO-01006-01
More Details
Advantec Disposable Ultrafiltration Unit, 200K MWCO, pack of 24
Category: Centrifugal Devices and Filters
Description: Advantec Disposable Ultrafiltration Units
Brand:
Advantec
YO-02910-54
More Details
Thermo Scientific GENESYS™ 10S Bio UV/Visible Spectrophotometer, 6
Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10S Bio UV/Visible Scanning Spectrophotometers
Brand:
Thermo Scientific Genesys
YO-02654-00
More Details
Wearwell Sanitizing Floor Mat, Tall Wall, 32" x 39", Black
Category: Wearwell Mats
Description: Wearwell Footwear Sanitizing Mats
Brand:
Wearwell
YO-84580-02
More Details
Thermo Scientific ERTCO® Non-Mercury ASTM Glass Thermometer, ASTM (063FBLSFC)
Category: Blue Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO
®
Blue Sprirt ASTM Equivalent Glass Thermometers
Brand:
Ertco
Model Number:
063FBLSFC
YO-08007-13
More Details
Hach SensION™+ Wastewater pH Meter Kit
Category: pH/Ion Handheld Meters
Description: Hach sensION + Waterproof pH/mV Meter
Brand:
Hach
Model Number:
PH1 5051T
YO-59004-03
More Details
Safety Sign, Emergency Exit Only - Door Alarm Will Sound, 10" X 14", A (MEXT16VS)
Category: Admittance & Exit Signs
Description: Safety: Emergency Exit Only - Door Alarm Will Sound Signs
Brand:
Accuform
Model Number:
MEXT16VS
YO-61014-40
More Details
Economical temperature controller, 110 VAC, °F (TS-13010)
Category: Controllers-Panel Mount
Description: Love Economical On/Off Temperature Controllers
Brand:
Love
Model Number:
TS-13010
YO-93520-00
More Details
GasAlertClip™ Extreme Personal Monitor, Hydrogen sulfide, 3-year op
Category: Single Gas Detection
Description: GasAlertClip Extreme Personal Monitors
Brand:
GasAlertClip
Model Number:
GA36XT-H
YO-86383-09
More Details
Type K High-temperature Flexible thermocouple probe; 12"L (050797B-K)
Category: Probes for Digi-Sense Temperature Controllers
Description: Thermocouple Probes
Brand:
Cole-Parmer
Model Number:
050797B-K
YO-08516-96
More Details
Female threaded tee fittings, nylon, 1/4" NPT(F), pack of 6 (YO-06349-51)
Category: Standard threaded tees
Description: Tees, NPT(F)
Brand:
Cole-Parmer
YO-06349-51
More Details
Calibrated loop, platinum, 10 microliter, 4 mm ID, 25 mm length (YO-01850-02)
Category: Inoculation Loops and Needles
Description: Oxford Platinum Inoculation Loops
Brand:
Oxford
YO-01850-02
More Details
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 48-well Multi (YO-01930-24)
Category: Essentials and Accessories for Microbial Analysis
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Delta MultiDishes
Brand:
Thermo Scientific Nunc
YO-01930-24
More Details
Safety Sign, Safety First Report All Accidents No Matter How Small To (MGNF984VS)
Category: First Aid/Emergency Eyewash & Shower Signs
Description: Signs; Safety First, Report All Accidents No Matter How Small To Your Supervisor
Brand:
Accuform
Model Number:
MGNF984VS
YO-60710-76
More Details
Cole-parmer Stainless Steel Syringe, 100 mL, Viton O-ring, Luer Lock
Category: Syringes and Accessories for Syringe Pumps
Description: Cole-Parmer Stainless Steel Syringes
Brand:
Cole-Parmer
YO-74044-46
More Details
PYREX Brand 5642 Volumetric Flask; 100 mL, pack of 6
Category: Glass Volumetric Flasks
Description: PYREX Brand 5642 Glass Volumetric Flasks
Brand:
Pyrex
YO-34561-06
More Details
Hach Fluoride (SPADNS) Reagent
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
25060-25
YO-99574-08
More Details
Halsted mosquito forceps, Dissecting-grade, Straight (YO-10918-14)
Category: Hemostatic Forceps
Description: Miltex Dissecting-grade Hemostatic Forceps
Brand:
Miltex
YO-10918-14
More Details
316 stainless steel 90° full-cone spray nozzle, 0.084" orifice
Category: Spray Nozzles
Description: Full-Cone Spray Nozzles, 316 Stainless Steel
Brand:
Cole-Parmer
Model Number:
460.646.17.BC.00.0
YO-83250-30
More Details
Tygon Silicone Tubing, 1/32"ID x 3/32"OD, 50 ft/pack
Category: Tygon Tubing
Description: Tygon Silicone Tubing
Brand:
Tygon
YO-95702-00
More Details
Pipe fittings, Nipple, PP, 10-32 UNF, 3/8", 1/4", 1/16"; 25/pack (YO-31503-10)
Category: Threaded nipples
Description: Nipples, UNF(M)
Brand:
Cole-Parmer
YO-31503-10
More Details
Thermo Scientific Finntip 10 mL Sterile Pipette Tips, 2 to 10 mL, 5x24
Category: Thermo Scientific Finntip Pipette Tips
Description: Thermo Scientific Finntip Pipette Tips (2000 to 10,000 uL)
Brand:
Thermo Scientific Finntip
Model Number:
9402163
YO-25001-83
More Details
64114
results
<<<Previous
9
10
11
12
13
14
15
16
17
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Thermo Scientific Heraeus Fresco™ 21 Refrigerated Microcentrifuge W (75002426) to Thermo Scientific Finntip 10 Ml Sterile Pipette Tip
>
31125329-p-toc_2 31125329ptoc2