Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 16
Ika T18 Ultra-Turrax Homogenizer 115vac
to
Pyrex Brand 1595 Solution Bottle
Image
Product Detail
SKU
IKA T18 ULTRA-TURRAX Homogenizer 115VAC
Category: IKA Homogenizers
Description: IKA T-18 Ultra Turrax Digital Homogenizer
Brand:
ULTRA-TURRAX
YO-04737-21
More Details
Baritainer Jerry Can, Natural, HDPE/Quoral 10 L
Category: Square/Rectangular Carboys and Jerricans
Description: Baritainer Jerry Can, Natural, HDPE/Quoral
Brand:
Cole-Parmer
YO-06069-03
More Details
Fluke VT02 Visual Infrared Thermometer
Category: Visual Infrared Thermometers
Description: Fluke VT02 Visual Infrared Thermometer
Brand:
Fluke
Model Number:
VT02
YO-39750-14
More Details
accumet AB150 pH Benchtop Meter Only
Category: pH/Ion Benchtop Meters
Description: accumet AB Series Benchtop Meters
Brand:
FISHER
Model Number:
AB150
YO-59331-52
More Details
Micro Scoop PP 3-7 mg
Category: Scoops and Spoons
Description: Micro Scoops
Brand:
Cole-Parmer
Model Number:
ASPS-02
YO-06277-54
More Details
ChloroSense ® Chlorine Meter (CS 100)
Category: pH/Ion Benchtop Meters
Description: Palintest ChloroSense Chlorine Meter
Brand:
Palintest
Model Number:
CS 100
YO-99359-10
More Details
DOP Test Kit For HEPA Filters (5242400)
Category: Labconco Glove Boxes and Balance Enclosures
Description: Accessories for Labconco PRECISE Basic and HEPA-Filtered Glove Boxes
Brand:
Labconco
Model Number:
5242400
YO-34765-65
More Details
PYREX Brand 5340 filter flask; 4000 mL, pack of 1
Category: Glass Filtering Flasks
Description: PYREX Brand 5340 Glass Flasks
Brand:
Pyrex
YO-34557-05
More Details
Polypropylene fittings for 1/16" tubing, 1/4-28 UNF(M), pack of 10 (YO-06471-58)
Category: Couplings and tube end fittings
Description: Inert Tube End Fittings
Brand:
Diba
YO-06471-58
More Details
RS-232 to phone jack adapter. Use with 00071-CL RJ-11 cable.
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
01X338901
YO-00123-UU
More Details
Millipore Syringe Filters, Millipore Express PES, .22um 50pk
Category: Syringe Filters
Description: Millipore Syringe Filters
Brand:
Millipore
YO-81053-02
More Details
Radiation Alert Inspector EXP Digital Radiation Monitor
Category: Radiation Detectors
Description: Radiation Alert Digital Radiation Monitor
Brand:
Radiation Alert
Model Number:
INSPECTOR-EXP
YO-81910-20
More Details
accumet® AB15 Basic pH/mV benchtop meter, 110/220 VAC
Category: pH/Ion Benchtop Meters
Description: accumet
®
AB15 Basic pH/mV Benchtop Meters
Brand:
Accumet
Model Number:
AB 15
YO-59330-00
More Details
Digital 2300 CCD microscope camera with software (SP100382D MOTICAM 2300)
Category: Cameras for Microscope Mounting
Description: Motic CCD Digital Microscope Cameras with Software
Brand:
Motic Instruments Inc
Model Number:
SP100382D MOTICAM 2300
YO-03908-20
More Details
Stage Micrometer; Scale 1 mm/0.01 mm (MA285)
Category: Meiji Modular Stereomicroscopes
Description: Meiji Micrometers
Brand:
Meiji
Model Number:
MA285
YO-48404-80
More Details
Safety Sign, Danger - Safety Glasses Required In This Area, 7" X 10", (MPPED04BVS)
Category: Danger Signs
Description: Danger: Safety Glasses Required In This Area Signs
Brand:
Accuform
Model Number:
MPPED04BVS
YO-50710-17
More Details
IKA RW 20 Digital Dual-Range Mixer, 60 to 500/240 to 2000, 230 VAC
Category: IKA Mixers
Description: IKA RW 20 Digital Dual-Range Mixers
Brand:
IKA
YO-50705-10
More Details
Digital Density / Specific Gravity Meter
Category: Density/Specific Gravity
Description: Digital Density/Specific Gravity Meter
Brand:
Cole-Parmer
YO-25750-60
More Details
Flexible PVC 180° U-bend, 5/8" Tubing OD (YO-31545-14)
Category: Fittings
Description: Flexible PVC Fittings
Brand:
Cole-Parmer
YO-31545-14
More Details
Nujol mineral oil
Category: Infrared spectroscopy accessories
Description: Sample Prep Materials
Brand:
Cole-Parmer
Model Number:
6109
YO-83001-15
More Details
Gas line for bunsen burners, flexible steel covered with rubber/cloth, (H-2087)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
H-2087
YO-36130-70
More Details
Hose Barb Adapter, PTFE, 1/2" NPT(M) X 1/4" ID (F-MH0402)
Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
Model Number:
F-MH0402
YO-34030-81
More Details
SPILL-X-A Acid Spill Treatment Kit
Category: Spill-X Spill Control Products
Description: Spill Treatment Kits
Brand:
SPILL-X-A
Model Number:
78776
YO-86292-00
More Details
Thermo Scientific Barnstead Disposable Ion Exchange Cartridge, two-bed (D8901)
Category: Water Purification Cartridges
Description: Thermo Scientific Barnstead Disposable Ion Exchange Cartridges
Brand:
Thermo Scientific Barnstead
Model Number:
D8901
YO-99260-02
More Details
Thermo Scientific Nunc Lab-Tek™ Petri Dish, 150 x 25 mm, 72/Case (YO-14007-14)
Category: Petri Dishes and Contact Plates
Description: Thermo Scientific Nunc Lab-Tek Sterile Petri Dishes, PS
Brand:
Thermo Scientific Nunc
YO-14007-14
More Details
Oxygen Electrode, microcathode
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Cole-Parmer
Model Number:
SI130
YO-53002-50
More Details
Fluke 50-Series II (Model 54) Thermocouple Thermometer Dual Input wit
Category: Handheld Thermocouple Meters
Description: Fluke 50-Series II Thermocouple Thermometers
Brand:
Fluke
Model Number:
FLUKE-54-2 60HZ
YO-91500-10
More Details
pHydrion plastic pH indicator strips, 0.0 to 14.0, clear plastic case
Category: pH Test Strips and Indicators
Description: pHydrion Plastic pH Strips in Clear Plastic Cases
Brand:
pHydrion
Model Number:
7800
YO-59200-10
More Details
NFPA Notice Chemical Hazard, Sign Only, 14" X 10", aluminum
Category: Safety Signs & Labels
Description: Right-To-Know NFPA Notice Chemical Hazard Signs and Kits
Brand:
Accuform
Model Number:
ZFD806VA
YO-41010-08
More Details
Pyrex Brand 1595 solution bottle; carboy shape, 19 L
Category: Solution Bottles
Description: Pyrex Brand 1595 Bottles
Brand:
Corning
Model Number:
1595-5
YO-34542-23
More Details
64114
results
<<<Previous
12
13
14
15
16
17
18
19
20
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Ika T18 Ultra-Turrax Homogenizer 115vac to Pyrex Brand 1595 Solution Bottle
>
31125329-p-toc_2 31125329ptoc2