Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 19
Thermocouple
to
Compression Straight Union
Image
Product Detail
SKU
Thermocouple,Wire,TYPE-K,20-GAUGE, Ceramic Fiber Braid Insulation, 100 (K20-2-350) (YO-08541-24)
Category: Wire - Bulk / Spooled Thermocouple Wire
Description: 20 Gauge Thermocouple Wire
Brand:
Cole-Parmer
Model Number:
K20-2-350
YO-08541-24
More Details
Thermo Scientific GENESYS™ 10S Visible Spectrophotometer USB; Singl
Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10S Visible and UV/Visible Scanning Spectrophotometers
Brand:
Thermo Scientific Genesys
YO-02654-10
More Details
Cole-Parmer Combination Ion Selective Electrodes, Copper (Cu+2)
Category: Ion Selective Electrodes
Description: Cole-Parmer Combination Ion-Selective Electrodes
Brand:
Cole-Parmer
YO-27504-10
More Details
Millipore Amicon Ultra Centrifugal Filters, 15ML 10K 24PK
Category: Centrifugal Devices and Filters
Description: Millipore Amicon Ultra Centrifugal Filters
Brand:
Millipore
YO-29969-82
More Details
Polycarbonate Membrane, 47mm diamter, 0.2um pore size, 100/pack (YO-36230-04)
Category: Membranes
Description: Polycarbonate Membranes
Brand:
Cole-Parmer
YO-36230-04
More Details
Bulkhead connectors, nylon, 1/4" NPT(F) x 3/4" OD, pack of 6 (YO-06349-11)
Category: Bulkhead unions
Description: Bulkhead connectors
Brand:
Cole-Parmer
YO-06349-11
More Details
Dry-Keeper™ Desiccator Cabinets, Vertical; Electronic (H420561003)
Category: Dry Keeper Desiccator Cabinets
Description: Dry-Keeper Dessicator Cabinets—Electric
Brand:
Bel-Art
Model Number:
H420561003
YO-08909-62
More Details
Cole-Parmer Gel-Loading Tips, 1 to 200 µL, 5 Racks, 204/pk
Category: Pipette Tips
Description: Cole-Parmer Gel-Loading Tips
Brand:
Cole-Parmer
Model Number:
010-R204-PRX
YO-25713-12
More Details
Thermo Scientific Orion Economy pH electrode, epoxy body, sealed, sing (9106BNWP)
Category: pH/Ion Laboratory Electrodes
Description: Thermo Scientific Orion Economy pH Electrodes
Brand:
Thermo Scientific Orion
Model Number:
9106BNWP
YO-05712-41
More Details
Cole-Parmer Wide-Form Crucible with Cover, Porcelain; 15 mL
Category: Porcelain Crucibles
Description: Cole-Parmer Wide-Form Crucibles with Covers, Porcelain
Brand:
Cole-Parmer
YO-63100-42
More Details
Variac Variable Autotransformer, Model W2. Single Phase, 120V, 50/60 H (3010-5110)
Category: Power/Voltage Controllers and Transformers
Description: Technipower Variac Variable Autotransformers
Brand:
Variac
Model Number:
3010-5110
YO-25751-50
More Details
Flir AC Adapter Charger with 110-240V with U.S. Plug (T910750)
Category: Thermal Imaging Windows and Accessories
Description: Flir Thermal Imager Accessories
Brand:
Flir
YO-39754-25
More Details
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 4-well MultiD (YO-01930-25)
Category: Essentials and Accessories for Microbial Analysis
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Delta MultiDishes
Brand:
Thermo Scientific Nunc
YO-01930-25
More Details
Oxygen flowmeter, 1/2 to 15 LPM, 1/8" NPT(F) Inlet
Category: Variable Area Flowmeters
Description: High-Accuracy Air and Oxygen Flowmeters
Brand:
Cole-Parmer
Model Number:
F026M
YO-32900-66
More Details
NIST-Traceable Calibration; Micrometers/Calipers, 0-24" (YO-17050-13)
Category: Calibration Services
Description: Dimensional Calibrations
Brand:
Cole-Parmer
YO-17050-13
More Details
Thermo Scientific Orion ROSS Ultra pH/ATC Triode, epoxy body, sealed, (8107UWMMD)
Category: pH/Ion Laboratory Electrodes
Description: Thermo Scientific Orion ROSS Ultra pH Electrodes
Brand:
Thermo Scientific Orion
Model Number:
8107UWMMD
YO-05718-48
More Details
Thermo Scientific Low Cost Orbital Shaker for 125 mL Erlenmeyer flasks (2345Q)
Category: Orbital Shakers
Description: Thermo Scientific Orbital Shaker
Brand:
Thermo Scientific
Model Number:
2345Q
YO-04730-20
More Details
B-D™ Disposable Syringes, Luer-Lock Tips, 5 mL (YO-07940-06)
Category: Chromatography Syringes
Description: BD Disposable Sterilized Syringes
Brand:
B-D
YO-07940-06
More Details
3M BX™ Dual Reader Safety Glasses, 2.0X top and bottom diopters
Category: Safety Glasses
Description: 3M BX Dual Reader Safety Glasses
Brand:
3M
Model Number:
11458-00000-20
YO-86474-33
More Details
Inlet/Outlet fitting, PP1-1/2" NPT(M) x 1 1/2" ID (YO-06450-84)
Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand:
Banjo
YO-06450-84
More Details
Low-Cost Digital Force Gauge
Category: Force Gauges
Description: Digital Force Gauges
Brand:
Extech
YO-59845-02
More Details
GasAlertClip™ Extreme Personal Monitor, Carbon monoxide, 3-year ope
Category: Single Gas Detection
Description: GasAlertClip Extreme Personal Monitors
Brand:
GasAlertClip
Model Number:
GA36XT-M
YO-86383-11
More Details
Styrofoam storage box for 100 (1.5 to 2.0 mL) microcentrifuge tubes
Category: Cryogenic Storage Boxes and Racks
Description: Styrofoam Storage Box
Brand:
Styrofoam
Model Number:
R-8000
YO-44350-20
More Details
Polycarbonate Filter Holder, 25 mm, 12/pk
Category: In-Line and Syringe Filter Holders
Description: Polycarbonate Filter Holders
Brand:
Cole-Parmer
YO-29550-42
More Details
Hach Sulfate (USEPA) Test Kit (21067-69)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
21067-69
YO-99574-21
More Details
Safety Sign, Warning - Do Not Enter Authorized Personnel Only, 10" X 1 (MADMW19VS)
Category: Warning Signs
Description: Warning: Do Not Enter - Authorized Personnel Only Signs
Brand:
Accuform
Model Number:
MADMW19VS
YO-51014-80
More Details
PVC static mixer without injection port; 4" dia (YO-04668-71)
Category: Static In-Line Mixers
Description: Koflo PVC Mixers Without Injection Port
Brand:
KoFlo
YO-04668-71
More Details
Enpac Secondary Containment Dolly for one 55-gallon drum
Category: Secondary Containment
Description: Enpac Secondary Containment Dolly
Brand:
Cole-Parmer
Model Number:
5300-YE
YO-86109-00
More Details
Hach Sulfide (USEPA) Test Kit (22445-00)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
22445-00
YO-99574-22
More Details
Compression Straight Union, Stainless Steel, 6mm OD Tubing (YO-31528-33)
Category: Compression Fittings
Description: Metric size stainless steel compression fittings
Brand:
metric fitting
YO-31528-33
More Details
64114
results
<<<Previous
15
16
17
18
19
20
21
22
23
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Thermocouple to Compression Straight Union
>
31125329-p-toc_2 31125329ptoc2