Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 2

Hach Free Chlorine (Usepa) Test Kit (21055-69)   to   Thermo Scientific Orion Ph

Image
  Product Detail
SKU

Hach Free Chlorine (USEPA) Test Kit (21055-69)

Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand: Hach
Model Number: 21055-69


Adapters with 1/4-28 UNF thread; female luer fitting; ETFE (YO-06471-91)

Category: Luer Fittings and Adapters
Description: ETFE Luer Adapters
Brand: Diba


Thermo Scientific Orion ROSS Ultra pH/ATC Triode, epoxy body, refillab (8157BNUMD)

Category: pH/Ion Laboratory Electrodes
Description: Thermo Scientific Orion ROSS Ultra pH Electrodes
Brand: Thermo Scientific Orion
Model Number: 8157BNUMD


Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyle (PB.25,TT3PP.)

Category: Air-Powered Double-Diaphragm Pumps
Description: Sandpiper Air-Powered Double-Diaphragm Polypropylene Pumps
Brand: Sandpiper
Model Number: PB.25,TT3PP.


Thermo Scientific Nunc CryoTube Starfooted Cryogenic Vials with intern (377267)

Category: Cryogenic Vials and Caps
Description: Thermo Scientific Nunc CryoTube Internal Thread Cryogenic Vials
Brand: Thermo Scientific Nunc
Model Number: 377267


Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 6-well MultiD (YO-01930-21)

Category: Essentials and Accessories for Microbial Analysis
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Delta MultiDishes
Brand: Thermo Scientific Nunc


Thermo Scientific SPECTRONIC™ standards, set of 10 (333150-000)

Category: Calibration Products and Standards
Description: Thermo Scientific SPECTRONIC™ Standards Set for Visible Light Spectroscopy
Brand: Thermo Scientific
Model Number: 333150-000


Cooper-Atkins DFP450W Waterproof digital thermometer with reduced tip

Category: Pocket Thermometers Digital - Food Industry
Description: COOPER-ATKINS DFP450W Waterproof digital thermometer with reduced tip and temperature alarm.
Brand: Cooper-Atkins
Model Number: DFP450W-0-8


PYREX Brand 2103 Burette; capacity, 50 mL; graduation interval, 0.10mL

Category: Glass and Plastic Burettes
Description: PYREX Brand 2103 Class A Burettes
Brand: Pyrex
Model Number: 2103-50


Hypersil BDS C18 Column - Length 250mm, 4.6mm ID, 5µm particle size (YO-01885-79)

Category: Thermo Scientific Hypersil BDS Columns and Accessories
Description: Thermo Scientific Hypersil BDS C18 Columns
Brand: Thermo Scientific Hypersil


NIST-Traceable Calibration; Rulers, 0-24" (YO-17050-16)

Category: Calibration Services
Description: Dimensional Calibrations
Brand: Cole-Parmer


Thermo Scientific Orion 4.01 pH buffer solution, 475-mL bottle

Category: Buffers and Solutions for pH/Ion Measurement
Description: Thermo Scientific Orion pH Buffer Solutions
Brand: Thermo Scientific Orion


Hach Total Nitrogen (TNT) Test Kit (26722-45)

Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand: Hach
Model Number: 26722-45


Ansell Edmont Sol-Vex Nitrile Gloves; Unlined; size 10; 11-mil; 13"L; (37-185 117301)

Category: Reusable Nitrile Gloves
Description: Ansell Sol-Vex Nitrile Gloves
Brand: Sol-Vex
Model Number: 37-185 117301


Isolated RS-232 to RS-422/485 Converter (ADAM-4520-D2E)

Category: Data Acquisition
Description: Advantech Data Acquisition I/O Modules
Brand: Advantech
Model Number: ADAM-4520-D2E


Free Chlorine Reagent Tablets, DPD #1, 100/BX (6903A-J)

Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand: Lab Products
Model Number: 6903A-J


DPD Free Chlorine Reagent Powder Pillow Packs, 10 mL, 100 tests (RP010-0)

Category: Colorimetry Reagents
Description: Powder Pack and Tube Test Reagents
Brand: Newest Products
Model Number: RP010-0


304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)

Category: Accessories for Gas Chromatography (GC)
Description: Stainless Steel Tubing
Brand: Cole-Parmer


Hayward double-length bag filter housing, 2" NPT(F)

Category: Bag Filter Systems
Description: Hayward Single- and Double-Length Bag Filters
Brand: Hayward
Model Number: FLT4202


Hach digital titrator (16900-01)

Category: Digital Burette
Description: Hach Digital Titrator
Brand: Hach
Model Number: 16900-01


C18 SPE cartridges, 6 mL (YO-10937-15)

Category: Solid phase extraction (SPE)
Description: Cole-Parmer SPE cartridges
Brand: Cole-Parmer


Fluke® 199C ScopeMeter® with Color Display, 200 MHz (FLUKE-199C/003)

Category: Oscilloscopes
Description: Fluke® 190-Series ScopeMeter® Oscilloscopes
Brand: Fluke
Model Number: FLUKE-199C/003


Barbed fittings, NPT female pipe adapters, Clear PP,1/4" NPT x 1/8" ID (YO-31501-41)

Category: Barbed to threaded adapters
Description: Straight barbed to female NPT threaded adapters
Brand: Cole-Parmer


inoLab 7110 basic pH/mV/T benchtop meter

Category: pH/Ion Benchtop Meters
Description: WTW inoLab pH 7110, pH 7310, and Multi 9310 Benchtop Meters
Brand: WTW
Model Number: PH 7110


Sugar/honey refractometer, 58 to 90% Brix

Category: Handheld Manual Refractometers
Description: Brix Refractometers
Brand: Cole-Parmer
Model Number: RHB-90


Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 10 psi and 0 to 70 kPa

Category: Mechanical Pressure Gauges
Description: Ashcroft Low-Pressure and Low-Vacuum Gauges
Brand: Ashcroft
Model Number: 14902.5


Burette, Class B, 50mL (2122A-50)

Category: Glass and Plastic Burettes
Description: Pyrex Brand 2122 Economy Grade Burettes
Brand: Pyrex
Model Number: 2122A-50


Handheld Mini 365 nm UV Lamp (95-0125-05(UVL-4))

Category: Ultraviolet Lamps
Description: Handheld mini ultraviolet lamps
Brand: Cole-Parmer
Model Number: 95-0125-05(UVL-4)


Thermo Scientific Orion pH/ATC Triode, epoxy body, sealed, single-junc (9107BNMD)

Category: pH/Ion Laboratory Electrodes
Description: Thermo Scientific Orion Standard Ag/AgCl pH Electrodes
Brand: Thermo Scientific Orion
Model Number: 9107BNMD


64114 results


Hach Free Chlorine (Usepa) Test Kit (21055-69) to Thermo Scientific Orion Ph

>