Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 2000

I/P   to   Cole-Parmer Black Nylon Hose Clamp

Image
  Product Detail
SKU

I/P 73 GORE Style 100SC Tubing for Masterflex Pumps, 24" per pack

Category: Masterflex GORE High-Resilience Style 100SC Pump Tubing Formulation
Description: I/P Precision GORE High-Resilience Style 100SC Pump Tubing for Masterflex I/P Pumps
Brand: Masterflex


I/P 82 GORE Style 100SC Tubing for Masterflex Pumps, 24" per pack

Category: Masterflex GORE High-Resilience Style 100SC Pump Tubing Formulation
Description: I/P Precision GORE High-Resilience Style 100SC Pump Tubing for Masterflex I/P Pumps
Brand: Masterflex


I/P 88 GORE Style 100SC Tubing for Masterflex Pumps, 24" per pack

Category: Masterflex GORE High-Resilience Style 100SC Pump Tubing Formulation
Description: I/P High-Performance Precision GORE High-Resilience Style 100SC Pump Tubing for Masterflex I/P Pumps
Brand: Masterflex


I/P 89 GORE Style 100SC Tubing for Masterflex Pumps, 24" per pack

Category: Masterflex GORE High-Resilience Style 100SC Pump Tubing Formulation
Description: I/P High-Performance Precision GORE High-Resilience Style 100SC Pump Tubing for Masterflex I/P Pumps
Brand: Masterflex


B/T 91 GORE Style 100SC Tubing for Masterflex Pumps, 27" per pack

Category: Masterflex GORE High-Resilience Style 100SC Pump Tubing Formulation
Description: B/T PerfectPosition GORE High-Resilience Style 100SC Pump Tubing for Masterflex B/T Pumps
Brand: Masterflex


B/T 87 GORE Style 100SC Tubing for Masterflex Pumps, 27" per pack

Category: Masterflex GORE High-Resilience Style 100SC Pump Tubing Formulation
Description: B/T PerfectPosition GORE High-Resilience Style 100SC Pump Tubing for Masterflex B/T Pumps
Brand: Masterflex


L/S 25 GORE Style 500 Tubing for Masterflex Pumps, 12" per pack

Category: Masterflex GORE High-Resilience Style 500 Pump Tubing Formulation
Description: L/S Precision GORE High-Resilience Style 500 Pump Tubing for Masterflex L/S Pumps
Brand: Masterflex


L/S 17 GORE Style 500 Tubing for Masterflex Pumps, 12" per pack

Category: Masterflex GORE High-Resilience Style 500 Pump Tubing Formulation
Description: L/S Precision GORE High-Resilience Style 500 Pump Tubing for Masterflex L/S Pumps
Brand: Masterflex


L/S 18 GORE Style 500 Tubing for Masterflex Pumps, 12" per pack

Category: Masterflex GORE High-Resilience Style 500 Pump Tubing Formulation
Description: L/S Precision GORE High-Resilience Style 500 Pump Tubing for Masterflex L/S Pumps
Brand: Masterflex


L/S 35 GORE Style 500 Tubing for Masterflex Pumps, 14" per pack

Category: Masterflex GORE High-Resilience Style 500 Pump Tubing Formulation
Description: L/S High-Performance Precision GORE High-Resilience Style 500 Pump Tubing for Masterflex L/S Pumps
Brand: Masterflex


Carrying Case, Star A Meters

Category: Thermo Scientific Orion Star A100 Series Portable pH/mV/Temperature Meter
Description: Star A Accessories
Brand: Thermo Scientific


Armor for Star A Meters

Category: Thermo Scientific Orion Star A100 Series Portable pH/mV/Temperature Meter
Description: Star A Accessories
Brand: Thermo Scientific


pH Electrode, Double-junction for Portable meter

Category: Hach sensION + Waterproof pH/mV Meter
Description: Replacement Electrodes
Brand: Hach


L/S 24 GORE Style 500 Tubing for Masterflex Pumps, 14" per pack

Category: Masterflex GORE High-Resilience Style 500 Pump Tubing Formulation
Description: L/S High-Performance Precision GORE High-Resilience Style 500 Pump Tubing for Masterflex L/S Pumps
Brand: Masterflex


Masterflex L/S Four-Channel Pump System for L/S Precision Pump Tubing;

Category: L/S Multichannel Pump Systems
Description: L/S Multichannel Pump Systems for L/S Pump Tubing
Brand: Masterflex


Extech MO265: Pocket Moisture Meter with Pin/Pinless Combination with

Category: Compact Moisture Meters
Description: Extech Pocket Size Moisture Meters
Brand: Extech
Model Number: MO265


Cole-Parmer Black Nylon Hose Clamps, 0.283" x 0.312" 100/pk

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Nylon Hose Clamps and Kits
Brand: Cole-Parmer


Cole-Parmer Black Nylon Hose Clamps, 0.350" x 0.395" 100/pk

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Nylon Hose Clamps and Kits
Brand: Cole-Parmer


Cole-Parmer Black Nylon Hose Clamps, 0.246" x 0.290" 100/pk

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Nylon Hose Clamps and Kits
Brand: Cole-Parmer


Cole-Parmer Black Nylon Hose Clamps, 0.360" x 0.410" 100/pk

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Nylon Hose Clamps and Kits
Brand: Cole-Parmer


Cole-Parmer Black Nylon Hose Clamps, 0.430" x 0.480" 100/pk

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Nylon Hose Clamps and Kits
Brand: Cole-Parmer


Cole-Parmer Black Nylon Hose Clamps, 0.500" x 0.545" 100/pk

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Nylon Hose Clamps and Kits
Brand: Cole-Parmer


Cole-Parmer Black Nylon Hose Clamps, 0.525" x 0.605" 100/pk

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Nylon Hose Clamps and Kits
Brand: Cole-Parmer


Cole-Parmer Black Nylon Hose Clamps, 0.650" x 0.750" 100/pk

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Nylon Hose Clamps and Kits
Brand: Cole-Parmer


Cole-Parmer Black Nylon Hose Clamps, 0.779" x 0.899" 100/pk

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Nylon Hose Clamps and Kits
Brand: Cole-Parmer


Cole-Parmer Black Nylon Hose Clamps, 0.900" x 0.989" 100/pk

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Nylon Hose Clamps and Kits
Brand: Cole-Parmer


Cole-Parmer Black Nylon Hose Clamps, 0.990" x 1.135" 100/pk

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Nylon Hose Clamps and Kits
Brand: Cole-Parmer


Cole-Parmer Black Nylon Hose Clamps, 1.123" x 1.296" 50/pk

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Nylon Hose Clamps and Kits
Brand: Cole-Parmer


Cole-Parmer Black Nylon Hose Clamps, 1.1250" x 1.425" 50/pk

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Nylon Hose Clamps and Kits
Brand: Cole-Parmer


Cole-Parmer Black Nylon Hose Clamps, 1.406" x 1.610" 50/pk

Category: Tubing Clamps, and Accessories
Description: Cole-Parmer Nylon Hose Clamps and Kits
Brand: Cole-Parmer




I/P to Cole-Parmer Black Nylon Hose Clamp

>