Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 21
Pyrex Brand 5640 Volumetric Flask
to
Thermo Scientific Orion Ise Fluoride Electrode
Image
Product Detail
SKU
PYREX Brand 5640 Volumetric Flask; 4000 mL, case of 1
Category: Glass Volumetric Flasks
Description: PYREX Brand 5640 Glass Volumetric Flasks
Brand:
Pyrex
YO-34505-34
More Details
Texwipe AlphaWipe Low-particulate wipes; 9" x 9", 150/bag
Category: Texwipe Low Particulate Wipes
Description: TEXWIPE Low-Particulate Cleanroom Wipes
Brand:
TEXWIPE
Model Number:
TX1009
YO-33677-00
More Details
DRAGER TUBES, Petroleum Hydrocarbons, 10 TO 300 PPM
Category: Gas Detection-Tubes
Description: Drager accuro Pump Kit and Gas Detection Tubes
Brand:
Drager
YO-86514-56
More Details
Cole-Parmer Symmetry PA-Analytical Balance, 220g x 0.0001g -- 220 VAC
Category: Cole-Parmer Symmetry Analytical Balances
Description: Cole-Parmer Symmetry PA Analytical Balances
Brand:
Symmetry
YO-10000-26
More Details
Barbed reducer, nylon, 3/8" x 1/4", pack of 10 (YO-30703-99)
Category: Barbed to barbed connectors
Description: Straight barbed reducing connectors
Brand:
Cole-Parmer
YO-30703-99
More Details
SPM VC100 VibChecker Vibration meter
Category: Vibration Measurement
Description: VibChecker Vibration Meter
Brand:
SPM
Model Number:
VC100
YO-65700-13
More Details
Rotary Evaporator, LABOROTA 4000 Efficient, Vertical Glassware, Manual (YO-28702-05)
Category: Heidolph Rotary Evaporators
Description: Heidolph® LABOROTA Efficient Rotary Evaporators
Brand:
Heidolph
YO-28702-05
More Details
CHECK VALVE, ACETAL BODY, push-to-connect, 3/8'' tube OD (A6VC6-MG)
Category: Check Valves
Description: Parker Plastic Check Valves
Brand:
Parker
Model Number:
A6VC6-MG
YO-01379-93
More Details
Barbed fittings, NPT male threaded adapter, HDPE, 3/4" NPT x 1/2" ID,9 (YO-30621-20)
Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30621-20
More Details
3M™ SE-401 Sound Level Meter, Type 1
Category: Sound Meters
Description: 3M
™
Sound Level Meters
Brand:
Quest Technologies - 3M Company
Model Number:
SE-401
YO-50536-35
More Details
Botulinum toxin detection kit (YO-36108-14)
Category: Bacteria test kits
Description: Biowarfare Agent Detection Devices (BADD)
Brand:
Cole-Parmer
YO-36108-14
More Details
Thermo Scientific Nalgene filling/venting cap for 1/4" tubing, 83B
Category: Bottle Accessories
Description: Thermo Scientific Nalgene Filling/Venting Caps
Brand:
Thermo Scientific Nalgene
Model Number:
2162-0831
YO-06258-20
More Details
Cole-Parmer Inline Bubble trap, PTFE and PEEK
Category: Microfluidic Syringes, Filtration, Accessories
Description: Bubble Trap
Brand:
PTFE
YO-75955-90
More Details
Three-port switching valve (YO-02012-02)
Category: UPCHURCH HPLC Valves and Filters
Description: Switching Valves
Brand:
UPCHURCH
YO-02012-02
More Details
Compression male pipe adapter elbow, 5/8" NPT(M) x 1/2" OD, brass
Category: Male Threaded Adapter Elbows
Description: Brass Compression to NPT(M) Adapter Elbows
Brand:
Cole-Parmer
YO-31412-89
More Details
Female threaded reducers, PVC, 3/8 NPT(F) x 1/4 NPT(F), pack of 6 (YO-06349-92)
Category: Female to female reducers
Description: Female reducers, NPT(F) x NPT(F)
Brand:
Cole-Parmer
YO-06349-92
More Details
Glass Coplin Staining Jar with glass cover (YO-48585-30)
Category: Staining Jars and Dishes for Histology
Description: Glass Coplin Staining Jar with Cover
Brand:
Cole-Parmer
YO-48585-30
More Details
316 SS Female Reducer, 3/4" x 1/4" NPT(F) (YO-31410-16)
Category: Female to female reducers
Description: Female reducers, NPT(F) x NPT(F)
Brand:
Cole-Parmer
YO-31410-16
More Details
Masterflex L/S Easy-Load II pump head for high-performance precision t (YO-77200-62)
Category: L/S Easy-Load II Pump Heads
Description: L/S Easy-Load II Pump Heads for High-Performance Precision Tubing
Brand:
Masterflex
YO-77200-62
More Details
Heidolph RZR 2020 Mixer System with propeller, clamp, and stand; 115VA
Category: Heidolph Laboratory Mixers
Description: Heidolph Laboratory Mixers and Accessories
Brand:
Heidolph
YO-54401-25
More Details
Co2 Gas Regulator 2 Stage
Category: CO
2
Incubators
Description: Thermo Scientific Series 8000 Incubators
Brand:
Thermo Scientific
YO-39320-30
More Details
Dc Transducer 4-20MA Dc And Ac/Dc Current Transducers
Category: Automation Control
Description: CR Magnetics Solid Core Current Voltage or Power Transducers
Brand:
CR Magnetics
Model Number:
CR5220-10
YO-68480-19
More Details
Regenerative Blower, 355/420 cfm (R7100A-3)
Description: Regenerative Blowers
Brand:
Gast
Model Number:
R7100A-3
YO-07047-75
More Details
Ashcroft Test Gauge, 0.5% Accuracy, 316 SS Bourdon Tube, 3" Dial, 30 t
Category: Mechanical Pressure Gauges
Description: Ashcroft Test Gauge
Brand:
Ashcroft
Model Number:
10843.0
YO-68034-01
More Details
Compression pipe adapter, Brass, 5/8" x 1/2", Each (YO-31412-78)
Category: Male Threaded Adapters
Description: Brass Compression to NPT(M) Adapters
Brand:
Cole-Parmer
YO-31412-78
More Details
Thermo Scientific Nalgene Polycarbonate CryoBox storage boxes for 81 ( (5026-0909)
Category: Cryogenic Storage Boxes and Racks
Description: Thermo Scientific Nalgene CryoBox Storage Boxes
Brand:
Thermo Scientific Nalgene
Model Number:
5026-0909
YO-06754-55
More Details
NIST-Traceable Calibration; pH Meter (YO-17106-20)
Category: Calibration Services
Description: pH Calibrations
Brand:
Cole-Parmer
YO-17106-20
More Details
Solenoid-Operated Three-Way Pinch Valve, 12 VDC (PM-0815W 12VDC N OPEN)
Category: Solenoid-Operated 2- and 3-Way Pinch Valves
Description: Three-Way Pinch Valves
Brand:
Cole-Parmer
Model Number:
PM-0815W 12VDC N OPEN
YO-98305-00
More Details
Fluke 337 Clamp Meter (YO-26036-07)
Category: Clamp Meters
Description: Fluke® 330 Series Clamp Meter
Brand:
FLUKE
YO-26036-07
More Details
Thermo Scientific Orion ISE fluoride electrode, solid-state, BNC conne (9409BN)
Category: Thermo Scientific Orion Ion-Selective Electrodes and Reference Electrodes
Description: Thermo Scientific Orion Half-Cell Ion-Selective Electrodes and Solutions
Brand:
Thermo Scientific Orion
Model Number:
9409BN
YO-05721-11
More Details
64114
results
<<<Previous
17
18
19
20
21
22
23
24
25
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Pyrex Brand 5640 Volumetric Flask to Thermo Scientific Orion Ise Fluoride Electrode
>
31125329-p-toc_2 31125329ptoc2