Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 22
Inductive
to
Contec Prewetted Cleanroom Wipe
Image
Product Detail
SKU
Inductive, M12, 2mm sensing distance, NPN-NO, shielded proximity senso
Category: Automation Control
Description: Omron Long Barrel Inductive Proximity Sensors
Brand:
Omron
Model Number:
E2E2-X2C1
YO-68513-17
More Details
Barbed fittings, NPT male pipe adapter elbows, Kynar,1/8" NPT x 1/8" I (YO-30704-30)
Category: Barbed to threaded adapters
Description: Barbed to male NPT threaded elbow adapters
Brand:
Cole-Parmer
YO-30704-30
More Details
Fluke 325 TRMS Clamp Meter, 400 A, with resistance, capacitance, and f
Category: Fluke Clamp Meters
Description: Fluke 323, 324, and 325 True RMS Clamp Meters
Brand:
Fluke
Model Number:
FLUKE-325
YO-20043-76
More Details
Electrode fill solution, 4 M KCl with Ag+. Case of five 60-mL bottles (900011)
Category: Buffers and Solutions for pH/Ion Measurement
Description: Thermo Scientific Orion Reference Fill Solutions
Brand:
Thermo Scientific Orion
Model Number:
900011
YO-05730-42
More Details
Female luer x 1/16" hose barb adapter, Nylon, 25/pk (YO-45502-00)
Category: Nylon Luer Fittings and Adapters
Description: Nylon Barbed Luer Adapters
Brand:
Cole-Parmer
YO-45502-00
More Details
Two-hole Rubber Stopper Size 6 1/2 21/pk
Category: Stoppers
Description: Cole-Parmer Two-Hole Stoppers, Rubber
Brand:
Cole-Parmer
YO-62991-78
More Details
Large Bottle Brush Volumetric 14"; Each
Category: Laboratory Brushes
Description: Beaker/Bottle/Flask Brushes
Brand:
Cole-Parmer
Model Number:
710380
YO-84554-02
More Details
Donaldson Ultrafilter Sterile Vent Filter, 41.4 cfm flow rate
Category: Ultrafilter Process Filtration
Description: Donaldson Ultrafilter Sterile Vent Filters
Brand:
Ultrafilter
Model Number:
P-BE-0072
YO-29582-02
More Details
Thermo Scientific Nalgene Centrifuge bottle; 500-mL; PPCO; 4/pack
Category: Centrifuge Bottles
Description: Thermo Scientific Nalgene 450- and 500-mL Centrifuge Bottles
Brand:
Thermo Scientific Nalgene
Model Number:
3120-0500
YO-06105-50
More Details
Pump, Centrifugal Pump, Hot water recirculation pump, 6 GPM, 1/2" Swea
Category: Centrifugal Pumps that are Mechanically Coupled (have a Mechanical Seal)
Description: Armstrong Fluid-Lubricated In-Line Circulators
Brand:
Armstrong
YO-79731-60
More Details
Temperature controller with timer (YO-94460-45)
Category: Temperature Controllers
Description: Cole-Parmer Controllers-with Timer
Brand:
Cole-Parmer
YO-94460-45
More Details
Whatman Quartz Filters, QM-A, 4.7 cm Dia
Category: Syringe Filters
Description: Whatman Quartz Filters
Brand:
Whatman
Model Number:
1851-047
YO-29706-12
More Details
Evaporator/Concentrator for 96-well Plates; stand-alone (non-heated) (YO-28690-50)
Category: Evaporative Sample Concentrator
Description: Cole-Parmer Evaporator/Concentrators
Brand:
Cole-Parmer
YO-28690-50
More Details
Temperature strip chart recorder
Category: Chart Recorders
Description: Temperature Strip Chart Recorder
Brand:
Cole-Parmer
Model Number:
SRJ
YO-80008-50
More Details
High-purity silicone stopper, size 00 (15D)
Category: Stoppers
Description: High-Purity Solid Stoppers, Silicone
Brand:
Lab Products
YO-62994-08
More Details
Precision Premium Mechanical Oven, 5.0 Cu. Ft. (141.6 L), 50°C To (PR305060M)
Category: Mechanical Convection Ovens
Description: Thermo Scientific Precision Premium Mechanical Convection Ovens
Brand:
Thermo Scientific Precision
Model Number:
PR305060M
YO-05014-31
More Details
Sandpiper Air-Powered Double-Diaphragm Stainless Steel Pump, 316 SS, 4 (SB1,SGN4SS.)
Category: Air-Powered Double-Diaphragm Pumps
Description: Sandpiper Air-Powered Double-Diaphragm Stainless Steel Pumps
Brand:
Sandpiper
Model Number:
SB1,SGN4SS.
YO-74007-22
More Details
Y-connector, Stainless Steel, 1/8 (YO-30620-21)
Category: Barbed to barbed connectors
Description: Barbed Y connectors
Brand:
Cole-Parmer
YO-30620-21
More Details
Elbow/Tee Kit, Polypropylene (YO-31514-58)
Category: Compression Fittings
Description: Kits of Assorted Compression Fittings
Brand:
Cole-Parmer
YO-31514-58
More Details
Aluminum test kit (3641-SC)
Category: LaMotte Smart Spectro Spectrophotometer
Description: LaMotte Reagent Kits
Brand:
LaMotte
Model Number:
3641-SC
YO-99565-10
More Details
Safety Sign, Fire Extinguisher (symbol), 7" X 10", Adhesive Vinyl (MFXG423VS)
Category: Fire Safety Signs
Description: Safety: Fire Extinguisher (symbol) Signs
Brand:
Accuform
Model Number:
MFXG423VS
YO-60710-55
More Details
SMART SPATULA™ Disposable Spatula, blue 210 mm (17211)
Category: Spatulas & Stir Rods
Description: Smart Spatula Disposable Spatulas, PP
Brand:
Smart Spatula
Model Number:
17211
YO-06265-18
More Details
Large folding cart (SBC203123)
Category: Service Carts
Description: Space-Saving Folding Carts
Brand:
Cole-Parmer
Model Number:
SBC203123
YO-47600-10
More Details
Male/female luer cap, sterile package, pack of 100 (YO-30526-08)
Category: Polycarbonate Luer Fittings and Adapters
Description: Polycarbonate Luer-to-Luer Connectors and Luer Plugs
Brand:
Cole-Parmer
YO-30526-08
More Details
Prepared agar plate, PDA (potato dextrose agar), 10/pk
Category: Prepared and Contact Media Plates
Description: Prepared Media Plates
Brand:
Cole-Parmer
YO-14201-52
More Details
Fixed-Volume Dispenser, 1 mL (4701211)
Category: Brand Dispensette III Bottletop Dispensers
Description: Dispensette III Fixed-Volume Dispensers
Brand:
Dispensette
Model Number:
4701211
YO-07912-02
More Details
Explosion-proof vacuum pump, 1.2 cfm, 27" Hg, 115 VAC (N035 STI-EX)
Category: KNF Specialty Vacuum/Pressure Pumps
Description: KNF Lab Explosion-Proof Pumps
Brand:
KNF
Model Number:
N035 STI-EX
YO-07056-50
More Details
BD™ bulk syringes, non-sterile clean, 10mL Luer-Lok™ tip, 850/ca (YO-07945-12)
Category: Syringes
Description: BD Disposable Syringes - Bulk Packages
Brand:
B-D
YO-07945-12
More Details
NSF-Certified Pressure Gauge, 0 to 30 PSI, Bottom Mount 1/4" NPT(M) Co (111.10DW)
Category: Mechanical Pressure Gauges
Description: Wika NSF-Certified Pressure Gauges
Brand:
Wika
Model Number:
111.10DW
YO-68049-01
More Details
CONTEC Prewetted Cleanroom Wipes, 100% acetone
Category: Clean Room Wipes
Description: CONTEC Prewetted Cleanroom Wipes
Brand:
Contec
Model Number:
SAT-C3-AC
YO-33675-08
More Details
64114
results
<<<Previous
18
19
20
21
22
23
24
25
26
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Inductive to Contec Prewetted Cleanroom Wipe
>
31125329-p-toc_2 31125329ptoc2