Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 29
Indenti-Plug Plastic Foam Stopper
to
Surface Thermometer
Image
Product Detail
SKU
Indenti-plug Plastic Foam Stopper, 46-50mm Openings, 80/pk
Category: Stoppers
Description: Identi-Plug Plastic Stoppers
Brand:
Indenti-Plug
YO-06298-80
More Details
PYREX Brand 2103 Burette; capacity, 10 mL; graduation interval, 0.05mL
Category: Glass and Plastic Burettes
Description: PYREX Brand 2103 Class A Burettes
Brand:
Pyrex
Model Number:
2103-10
YO-34543-21
More Details
Whatman Quartz Filters, QM-A, 8"W x 10"H
Category: Syringe Filters
Description: Whatman Quartz Filters
Brand:
Whatman
Model Number:
1851-865
YO-29706-22
More Details
Hach Low-range Nitrate Test Kit (24298-00)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
24298-00
YO-99574-12
More Details
Extech Humidity/Thermometer/Heat Index Stopwatch
Category: Digital Stopwatches
Description: Extech Humidity/Thermometer/Heat Index Stopwatch
Brand:
Extech
Model Number:
HW30
YO-94410-70
More Details
Regenerative Blower, 133/160 cfm (R5325A-2)
Description: Regenerative Blowers
Brand:
Gast
Model Number:
R5325A-2
YO-07047-45
More Details
Atago Salt refractometer, 0 to 28% salinity
Category: Salt and RI Refractometers
Description: Salt Refractometers
Brand:
Cole-Parmer
Model Number:
2483 MASTER 28M
YO-02940-43
More Details
LaMotte Dissolved Oxygen Test Kit, 50 tests/kit
Category: Water Quality Test Kits
Description: LaMotte Dissolved Oxygen Test Kit
Brand:
LaMotte
Model Number:
5860
YO-53003-00
More Details
Bergey's Manual of Determinative Bacteriology (0-683-00603-7)
Category: Biochemistry
Description: Bergey's Manual of Determinative Bacteriology
Brand:
Cole-Parmer
Model Number:
0-683-00603-7
YO-15366-01
More Details
Thermo Scientific Nalgene ® Automatic self-zeroing burette; capacity, (3640-0025)
Category: Glass and Plastic Burettes
Description: Thermo Scientific Nalgene Automatic Self-Zeroing Burettes
Brand:
Thermo Scientific Nalgene
Model Number:
3640-0025
YO-06209-20
More Details
Hose Barb Adapter, PTFE, 3/4" NPT(F) X 1/2" ID (F-FH0604)
Category: Barbed to threaded adapters
Description: Straight barbed to female NPT threaded adapters
Brand:
Cole-Parmer
Model Number:
F-FH0604
YO-34030-99
More Details
Thermo Scientific Nalgene Centrifuge bottle; 450-mL; PC; 4/pack
Category: Centrifuge Bottles
Description: Thermo Scientific Nalgene 450- and 500-mL Centrifuge Bottles
Brand:
Thermo Scientific Nalgene
Model Number:
3140-0500
YO-06106-35
More Details
Thermo Scientific Nalgene polypropylene wide-mouth bottle, 30 mL
Category: Sample Bottles
Description: Thermo Scientific Nalgene Wide-Mouth Polypropylene Bottles, Leakproof
Brand:
Thermo Scientific Nalgene
Model Number:
2105-0001
YO-06048-01
More Details
Digital Pressure Gauge, 0-3000 PSI
Category: Digital Pressure Gauges and Meters
Description: SSI MediaGauge Digital Pressure Gauge
Brand:
SSI Mediagauge
Model Number:
KCPSSDPG(3000)
YO-68349-16
More Details
MYRON L TechPro II™ TPH1 pH/Conductivity Meter
Category: Handheld Multiparameter Meters
Description: MYRON L Waterproof TechPro II TPH1 pH/Conductivity Meter
Brand:
Myron L
Model Number:
TPH1
YO-19707-07
More Details
General-purpose ultrasonic cleaning solution, 0.9 L (1 qt) bottle
Category: Ultrasonic Cleaning Solutions
Description: General-purpose cleaning solution
Brand:
Branson
YO-08801-11
More Details
Compression To Thread Adapter, Stainless Steel, 12mm OD Tubing X 1/2" (YO-31528-12)
Category: Compression Fittings
Description: Metric size stainless steel compression fittings
Brand:
metric fitting
YO-31528-12
More Details
Threaded adapter, stainless steel, 3/4" female BSPT to 3/4" male NPT (YO-31830-64)
Category: BSP Adapters
Description: British Standard Pipe Tapered Threaded (BSPT) Adapters
Brand:
Parker Hannifin
YO-31830-64
More Details
3M KCL Fill Solution, 250 mL
Category: pH/Ion Laboratory Electrodes
Description: Pinnacle pH Electrodes (formerly Corning Electrodes) and Accessories
Brand:
Corning
YO-58905-82
More Details
Sterile Multichannel Disposable Solutions Basin (HS20521C)
Category: Multichannel Pipette Reservoir
Description: Multichannel Disposable Solutions Basins
Brand:
Cole-Parmer
Model Number:
HS20521C
YO-24810-35
More Details
PVC Ball Check Valve, 3/8" NPT(F) (161.360.581)
Category: Hayward True Union Ball Check Valves
Description: Plastic Check Valves with Socket and NPT(F) Threaded Connectors
Brand:
Cole-Parmer
Model Number:
161.360.581
YO-01368-10
More Details
PYREX Brand 2103 Burette; capacity, 25 mL; graduation interval, 0.10mL
Category: Glass and Plastic Burettes
Description: PYREX Brand 2103 Class A Burettes
Brand:
Pyrex
Model Number:
2103-25
YO-34543-22
More Details
500 mL Cylindrical Flask (YO-34706-46)
Category: Multi-neck Reaction flasks
Description: Kontes Reaction Glass Flasks, Removable Four Neck Top
Brand:
Kontes
YO-34706-46
More Details
Masterflex PharMed BPT Tubing, L/S #16, 25' (YO-06508-16)
Category: Masterflex PharMed BPT Pump Tubing Formulation
Description: L/S Precision PharMed BPT Pump Tubing for Masterflex L/S Pumps
Brand:
Masterflex
YO-06508-16
More Details
Kapton heat mat kit, w/adhesive, 120 VAC
Category: Heating Tapes - Kapton Heaters and Heater Kits
Description: Kapton Heater Kits
Brand:
Kapton
Model Number:
K05711980A
YO-36067-10
More Details
Pitched blade impeller; 2-1/4" dia
Category: Mixer Accessories
Description: Laboratory Mixer Blade Impellers
Brand:
Cole-Parmer
YO-50801-62
More Details
Babcock bottle, milk test (YO-34590-06)
Category: Babcock System for Fat Analysis
Description: Babcock and Paley Bottles
Brand:
Cole-Parmer
YO-34590-06
More Details
YSI Pro20 Dissolved Oxygen Meter
Category: Dissolved Oxygen Handheld Meters
Description: YSI Pro20 Dissolved Oxygen Meter
Brand:
YSI
Model Number:
6050020
YO-59352-04
More Details
WTW Multi 3410 Multiparameter Meter w/FDO 925-3 dissolved oxygen probe
Category: Handheld Multiparameter Meters
Description: WTW MultiLine 3410 Multiparameter Meters
Brand:
WTW
YO-99575-07
More Details
Surface Thermometer , 2.5" Face, 1-Spring Pipe Mount, 0 to 250 ° F (50324373)
Category: Surface Thermometers
Description: Spring Mount Surface Thermometers
Brand:
Wika
Model Number:
50324373
YO-08107-25
More Details
64114
results
<<<Previous
25
26
27
28
29
30
31
32
33
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
10
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
42
43
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Indenti-Plug Plastic Foam Stopper to Surface Thermometer
>
31125329-p-toc_2 31125329ptoc2