Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 3

Vinyl Tubing   to   Laboratory Hydrogen Generator

Image
  Product Detail
SKU

Vinyl Tubing, 1/16"ID x 1/8"OD, 50 ft/pack (YO-06405-01)

Category: Flexible Tubing
Description: Vinyl Tubing
Brand: Cole-Parmer


316 Stainless Steel Tubing; dimensions, 3/8" ID, 1/2" OD, 1/32" wall; (YO-03300-20)

Category: Rigid Tubing
Description: 316 Stainless Steel Tubing
Brand: Cole-Parmer


Sterile Sample Bottles, 100 Pk

Category: Hach Modified Colitag Presence/Absence Test
Description: Sterile Sample Bottles, 100mL
Brand: Hach


High-purity silicone stopper, size 6D

Category: Stoppers
Description: High-Purity Solid Stoppers, Silicone
Brand: Lab Products


Pressure Gauge ~ 1/4" NPT (M) process connection;0 to 60" Water Co

Category: Mechanical Pressure Gauges
Description: Cole-Parmer Low-Pressure Mechanical Gauge
Brand: Cole-Parmer
Model Number: KC25-60"H2O


Cole-Parmer pH 7 buffer solution, 4 L bottle, NIST-traceable reference

Category: Buffers and Solutions for pH/Ion Measurement
Description: Cole-Parmer pH Buffers, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Schedule 40 PVC piping, 1.5", 9' (YO-33312-26)

Category: Piping
Description: Schedule 40 PVC Piping
Brand: Cole-Parmer


Threaded adapter, stainless steel, 1/4" female NPT to 1/4" male BSPT (YO-31830-41)

Category: BSP Adapters
Description: British Standard Pipe Tapered Threaded (BSPT) Adapters
Brand: Parker Hannifin


Standard Methods for the Examination of Water and Wastewater, 22nd edi

Category: Environmental Books
Description: Standard Methods Book
Brand: Cole-Parmer
Model Number: 9780875530130


ROSS electrode fill solution, 3 M KCl. Case of five 60-mL bottles (810007)

Category: Buffers and Solutions for pH/Ion Measurement
Description: Thermo Scientific Orion Reference Fill Solutions
Brand: Thermo Scientific Orion
Model Number: 810007


Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 3 psi and 0 to 20 kPa

Category: Mechanical Pressure Gauges
Description: Ashcroft Low-Pressure and Low-Vacuum Gauges
Brand: Ashcroft
Model Number: 14902.5


AC Variable-Speed Controller, 1/2 hp, 115/230 VAC, 2.4 amp, 120-240VAC

Category: AC Controllers
Description: Single-Phase Input Variable-Speed Controllers
Brand: Automation Control
Model Number: ESV371N01SXB


Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 24-well Multi (YO-01930-40)

Category: Essentials and Accessories for Microbial Analysis
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Delta MultiDishes
Brand: Thermo Scientific Nunc


Parafilm Wrap, 4"W

Category: Film and Foil
Description: Parafilm Wrap and Dispenser
Brand: Parafilm
Model Number: PM996


Thermo Scientific Finntip 5 mL Pipette Tips, 1000 to 5000 µL, 500/b

Category: Thermo Scientific Finntip Pipette Tips
Description: Thermo Scientific Finntip Pipette Tips (1000 to 5000uL)
Brand: Finntip
Model Number: 9402-030


Accumet pH Electrode, Combination, Double-junction, Gel-filled, Epoxy

Category: accumet Combination pH Electrodes from Cole-Parmer
Description: Cole-Parmer Rugged epoxy-body ACCUMET compatible combination electrodes
Brand: Accumet


240vac to 24 vdc power supply

Description: General Accessories
Brand: Davis


Threaded adapter, stainless steel, 1/2" female NPT to 1/2" male BSPT (YO-31830-43)

Category: BSP Adapters
Description: British Standard Pipe Tapered Threaded (BSPT) Adapters
Brand: Parker Hannifin


ATAGO Digital Pocket Refractometer, 0-53% Brix

Category: Digital Handheld Refractometers
Description: Atago Digital Handheld Pocket Refractometers
Brand: Atago
Model Number: 3810 (PAL-1)


Peptone water, 0.1% prepared;500 mL bottle, 10/pk (YO-14204-76)

Category: Prepared and Contact Media Plates
Description: Prepared Media Bottles
Brand: Cole-Parmer


Test Strips, Chlorine Dioxide, 0-500ppm, 50 strips/vial (3002)

Category: Water Quality Test Strips
Description: Lamotte Sanitizer Test Kits and Strips
Brand: Guaranteed In Stock Program
Model Number: 3002


316 Stainless Steel Tubing; dimensions, 3/16" ID, 1/4" OD, 1/32" wall; (YO-03300-10)

Category: Rigid Tubing
Description: 316 Stainless Steel Tubing
Brand: Cole-Parmer


Horiba™ D-54 pH/Conductivity Meter

Category: Handheld Multiparameter Meters
Description: Horiba D-54 pH/Conductivity Meter
Brand: Horiba
Model Number: 352734-1


B-D™ Disposable Syringes, Luer-Lock Tips, 1 mL (YO-07940-00)

Category: Chromatography Syringes
Description: BD Disposable Sterilized Syringes
Brand: B-D


Iris scissors, Precision-grade, Straight

Category: Scissors
Description: Miltex Precision-grade scissors
Brand: Miltex


Low-cost, Direct-drive Air Motor Mixer; 200-10,000 RPM (YO-04680-00)

Category: Hazardous-Duty Air-Drive Mixers
Description: Hazardous-Duty Direct-Drive Air Motor Mixers
Brand: Guaranteed In Stock Program


LaMotte Dissolved Oxygen Test Kit Refill; Winkler Titration Method, 50

Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand: LaMotte
Model Number: R-5860


Thermo Scientific NESLAB ThermoFlex™ 900 Recirculating Chiller with (YO-13042-00)

Category: Recirculators & Chillers
Description: Thermo Scientific NESLAB ThermoFlex Recirculating Chillers
Brand: Thermo Scientific


PTFE Tubing, 1/16"ID x 1/8"OD, 25 ft/pack (YO-06605-27)

Category: PTFE Tubing
Description: Cole-Parmer PTFE Tubing
Brand: Cole-Parmer


Laboratory Hydrogen Generator; 600mL/min; 115/230 VAC

Category: Gas Generators
Description: Laboratory Gas Generators for GC Applications
Brand: Cole-Parmer


64114 results


Vinyl Tubing to Laboratory Hydrogen Generator

>