Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 300
Refill Solution for Reference Electrode (362178)
to
Hypersil Silica Column - Length 50mm
Image
Product Detail
SKU
Refill solution for reference electrode (362178)
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Lab Products
Model Number:
362178
YO-99590-60
More Details
Bottle brush, X-shaped nylon bristles, green
Category: Cleaning Brushes
Description: Beaker/Bottle Brushes
Brand:
Cole-Parmer
Model Number:
1855
YO-84550-04
More Details
Thermo Scientific Nalgene straight-sided polycarbonate wide-mouth jar, (2116-0030)
Category: Jars
Description: Polycarbonate jars
Brand:
Thermo Scientific Nalgene
Model Number:
2116-0030
YO-06760-50
More Details
Inlet condensate trap
Category: Inlet Filters and Traps
Description: Inlet Condensate Trap
Brand:
Cole-Parmer
Model Number:
A441-10-000
YO-79302-00
More Details
BIOSIMPLEX CARBOY EZ TOP ASSEMBLY, 20L, 2 PORTS, 3/8 X 5/8 TUBING, inc
Category: Round Carboys
Description: Additional Cap Options and Spigot for Bio-Simplex Carboys
Brand:
Cole-Parmer
YO-62500-49
More Details
Avoirdupois Weight Hanger with Statement of Accuracy, 5 lb
Category: Troemner Class 6 Cast Iron Slotted Weights and Hangers
Description: Troemner Class 6 Cast Iron Slotted Weight Hangers
Brand:
Troemner
Model Number:
9484
YO-11966-78
More Details
Open-walled container, 24" x 16" x 12" (37-612)
Category: Plastic Storage Bins & Containers
Description: Open-Walled Containers
Brand:
Cole-Parmer
Model Number:
37-612
YO-47530-10
More Details
Extech WB200 Hygro-Thermometer Datalogger Measures Wet Bulb and Dew Po
Category: Benchtop Thermohygrometers
Description: Extech Thermohydrometer with Wet Bulb, Dew Point and Datalogging
Brand:
Extech
YO-95001-63
More Details
Smith & Wesson Pouch with Belt Clip
Category: Eyewear Accessories
Description: Smith & Wesson Glasses Pouch with Belt Clip
Brand:
Smith & Wesson
Model Number:
SWCPI
YO-86286-60
More Details
Scienceware safety labeled 500-mL wash bottle for methanol
Category: Wash Bottles
Description: Safety Labeled Wide-Mouth Wash Bottles
Brand:
Bel-Art
Model Number:
11646-0623
YO-06256-50
More Details
3-Stop Tygon® Lab Tubing, 1.65 mm ID, 12/pack (CP95608-38)
Category: Ismatec® Pump Tubing
Description: Three-Stop Color-Coded Autoanalysis Tubing for Ismatec Pumps
Brand:
Tygon
Model Number:
CP95608-38
YO-95608-38
More Details
Self-Monitoring Ductless Fume Hood, 42", 115 VAC (YO-33713-10)
Category: Ductless Fume Hoods
Description: Self-Monitoring Ductless Fume Hoods
Brand:
Cole-Parmer
YO-33713-10
More Details
Fluke® 975 AirMeter™ Calibration Kit (975)
Category: General Accessories
Description: Replacements Accessories
Brand:
Fluke
Model Number:
975
YO-86570-15
More Details
Thermo Scientific Nalgene high-density polyethylene wide-mouth bottle, (2104-0008)
Category: Sample Bottles
Description: Thermo Scientific Nalgene Wide-Mouth High-Density Polyethylene Bottles
Brand:
Thermo Scientific Nalgene
Model Number:
2104-0008
YO-06047-40
More Details
Burkert Type 2333 316L SS Diapharagm Valve, 2 Way, EPDM Wetted Materia (YO-98516-24)
Category: Diaphragm Valves
Description: Burkert Type 3233 316L Stainless Steel Diaphragm Valve
Brand:
Burkert
YO-98516-24
More Details
304 High-flow Submersible Pump, 100 GPM, Automatic, 115 VAC
Category: Submersible Pumps and Sump Pumps
Description: 304 SS High-Flow Submersible Pumps
Brand:
Cole-Parmer
YO-75505-50
More Details
Type K Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD
Category: Oakton Surface probes
Description: VELCRO Strap-On Surface Probes
Brand:
Velcro
Model Number:
112801L-K
YO-08469-82
More Details
Bleach Spray System Kit, (includes six, 11-oz. bleach cartridges, two, (77062)
Category: Lab and General Purpose Cleaners
Description: Bleach Spray System
Brand:
DEARDORFF FITZSIMMONS CORP
Model Number:
77062
YO-18105-00
More Details
Thermo Scientific Nalgene graduated polypropylene cylinder, 1000 mL
Category: Plastic Graduated Cylinders
Description: Thermo Scientific Nalgene Graduated Cylinders, PP
Brand:
Thermo Scientific Nalgene
Model Number:
3664-1000
YO-06135-72
More Details
PROPIOPHENONE 99% 2ML (YO-88134-55)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88134-55
More Details
Polypropylene Buchner funnel, 90 mm dia (YO-06128-04)
Category: Buchner Funnels
Description: Plastic Buchner Funnels
Brand:
Cole-Parmer
YO-06128-04
More Details
Solenoid Valve 2-Way, Zero Differential Pressure, Normall Closed, 1/2 (VXZ22 40L-04N-5DZ1-B)
Category: Solenoid Valves
Description: Series VXZ Solenoid Valves for Process Applications
Brand:
SMC Industries
Model Number:
VXZ22 40L-04N-5DZ1-B
YO-08617-36
More Details
YSI 5300A Biological Oxygen Monitor, 230 VAC
Category: Dissolved Oxygen Benchtop Meters
Description: YSI 5300A Biological Oxygen Monitor
Brand:
YSI
Model Number:
5300A-2
YO-05520-46
More Details
Extech Waterproof CL200 Pocket Chlorine Tester (CL200)
Category: pH/Ion Pocket Testers
Description: Extech Waterproof CL200 Pocket Chlorine Tester
Brand:
Extech
Model Number:
CL200
YO-99598-15
More Details
Multiport connector; 8-way, tube OD, 1 to 4 mm (YO-06473-05)
Category: PTFE Connectors
Description: Multiport connectors
Brand:
Diba
YO-06473-05
More Details
RVP Pressure Gauge, 0 to 100 psi
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
311-100-002
YO-59877-28
More Details
Portable Stainless Steel Cart for Laboratory Filtering Centrifuge
Category: Centrifuges
Description: Western States Laboratory Filtering Centrifuge
Brand:
Western States
YO-17400-72
More Details
Vacuum/pressure diaphragm pumps, PTFE-coated wetted parts, 0.55 cfm, 2
Category: Dry Vacuum Pressure Pumps
Description: Vacuum/Pressure Pumps Coated with PTFE
Brand:
PTFE
Model Number:
107CD18TFEL-247
YO-79200-35
More Details
Temperature Controller, RTD, 1/4-DIN, Relay Output, 1 alarms, (DC1044CR-101-000-E)
Category: Controllers-Panel Mount
Description: Honeywell DC 1000-Series Temperature/Process Controllers
Brand:
Honeywell
Model Number:
DC1044CR-101-000-E
YO-89556-29
More Details
Hypersil Silica Column - Length 50mm, 4.6mm ID, 5µm particle size (YO-01887-04)
Category: Thermo Scientific Hypersil Classical HPLC Columns and Accessories
Description: Thermo Scientific Hypersil Silica Classical HPLC Columns
Brand:
Thermo Scientific Hypersil
YO-01887-04
More Details
64114
results
<<<Previous
296
297
298
299
300
301
302
303
304
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
100
200
210
220
230
240
250
260
270
280
285
286
287
288
289
290
291
292
293
294
295
296
297
298
299
300
301
302
303
304
305
306
307
308
309
310
311
312
313
314
320
330
340
350
360
370
380
390
400
500
600
700
800
900
1,000
1,100
1,200
2,000
2,138
Refill Solution for Reference Electrode (362178) to Hypersil Silica Column - Length 50mm
>
31125329-p-toc_2 31125329ptoc2