Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 4
Horiba U-52 Multiparameter Meter
to
Thermo Scientific Snakeskin Pleated Dialysis Tubing
Image
Product Detail
SKU
Horiba U-52 Multiparameter Meter, 6-ft (2-m) cable
Category: Handheld Multiparameter Meters
Description: Horiba U-52 Multiparameter Meter
Brand:
Horiba
Model Number:
U-52
YO-99600-22
More Details
Thermo Scientific Thermolyne Benchtop Muffle Furnace with 350 cu in ch (F48015-60)
Category: Thermo Scientific Muffle Furnaces
Description: Thermo Scientific Thermolyne Benchtop Muffle Furnaces
Brand:
Thermo Scientific Thermolyne
Model Number:
F48015-60
YO-02053-12
More Details
Extraction thimble for soxhlet extraction glassware systems, 25 mL (YO-99036-50)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
YO-99036-50
More Details
Thermo Scientific Nunc CryoTube cryogenic tube with round bottom; 1.8 (368632)
Category: Cryogenic Vials and Caps
Description: Thermo Scientific Nunc CryoTube Internal Thread Cryogenic Vials
Brand:
Thermo Scientific Nunc
Model Number:
368632
YO-03755-10
More Details
Thermo Scientific Nunc Non-treated 700mL Cell Culture Triple Flask 32/
Category: Thermo Scientific Nunc Cell Culture Flasks
Description: Thermo Scientific Nunc Non-Treated Polystyrene Culture Flasks
Brand:
Thermo Scientific Nunc
YO-01928-42
More Details
GE (Druck) DPI 620 CE Multifunction Calibrator with Windows CE (Now ca
Category: Calibration Equipment
Description: GE (Druck) DPI 620 Multifunction Calibrator
Brand:
Druck
Model Number:
DPI 620 CE
YO-23000-38
More Details
WTW Multi 3430 Multiparameter Meter w/SenTix 940, TetraCon 925, and FD
Category: Handheld Multiparameter Meters
Description: WTW MultiLine 3430 Multiparameter Meters
Brand:
WTW
YO-99575-28
More Details
Thermo Scientific Orion electrode fill solution, 3 M KCl, 5 x 60mL bot
Category: Buffers and Solutions for pH/Ion Measurement
Description: Thermo Scientific Orion Reference Fill Solutions
Brand:
Thermo Scientific Orion
Model Number:
900011
YO-05719-25
More Details
Fluke 68-IS : Intrinsically Safe Infrared Thermometer (FLUKE-68IS)
Category: Handheld Infrared Thermometers
Description: Fluke 63, 66, 68, 566, 568 Infrared Thermometers
Brand:
Fluke
Model Number:
FLUKE-68IS
YO-39753-75
More Details
Leica Zoom 2000™ Stereozoom Microscope, 10x to 30x Magnification, L
Category: Stereo and Stereozoom Microscopes
Description: Leica Zoom E-Series Stereozoom Microscopes
Brand:
Leica
Model Number:
ES2 10447202
YO-49400-01
More Details
Calibration fluid for Thermo Scientific NanoDrop
Category: Calibration Products and Standards
Description: Thermo Scientific NanoDrop Spectrophotometer and Fluorometer Calibration Accessories
Brand:
Thermo Scientific NanoDrop
Model Number:
CHEM-CF1
YO-83060-90
More Details
Pressure Gauge ~ 1/4" NPT(M) process connection; 0 to 100" Water C
Category: Mechanical Pressure Gauges
Description: Cole-Parmer Low-Pressure Mechanical Gauge
Brand:
Cole-Parmer
Model Number:
KC25-100"H2O
YO-68357-08
More Details
Pyrex Desiccator, 10.5L
Category: Non Vacuum Desiccators
Description: Pyrex Desiccators, Glass
Brand:
Pyrex
Model Number:
3081-250
YO-34548-24
More Details
Cooper-Atkins DPP400W Waterproof digital pen style thermometer with re
Category: Pocket Thermometers Digital - Food Industry
Description: COOPER-ATKINS DPP400W Waterproof digital pen style thermometer with reduced tip
Brand:
Cooper-Atkins
Model Number:
DPP400W-0-8
YO-90025-02
More Details
Drager Tubes, Ammonia, 2.5 to 100 ppm
Category: Gas Detection-Tubes
Description: Drager accuro Pump Kit and Gas Detection Tubes
Brand:
Drager
YO-86514-14
More Details
Thermo Scientific Heraeus Multifuge X3R Refrigerated Centrifuge, 120VA
Category: Thermo Scientifc Heraeus Centrifuges
Description: Thermo Scientific Heraeus Multifuge X3 Centrifuges
Brand:
Thermo Scientific Heraeus
Model Number:
75004516
YO-17707-80
More Details
Thermo Scientific Orion ROSS Sure-Flow pH electrode, glass body, refil
Category: pH/Ion Laboratory Electrodes
Description: Thermo Scientific Orion ROSS pH Electrodes
Brand:
Thermo Scientific Orion
Model Number:
8172BNWP
YO-05718-51
More Details
Refrigerator/freezer thermometer with bottle probe (4127CP)
Category: Digital Thermometers - Food Industry
Description: Cole-Parmer General-Purpose and Refrigerator/Freezer Thermometers
Brand:
Cole-Parmer
Model Number:
4127CP
YO-94460-72
More Details
Safety Sign, Eye Wash Station, 10 X 14, Plastic
Category: First Aid/Emergency Eyewash & Shower Signs
Description: Safety: Eye Wash Station Signs
Brand:
Accuform
Model Number:
MFSAD31VP
YO-61014-59
More Details
Rotor-X Low-Flow Paddle-Wheel Flow Sensor with polypropylene body, tit (3-2536-PO)
Category: +GF+ Signet Flowmeter System
Description: Rotor-X Low-Flow Paddle-Wheel Flow Sensors
Brand:
+GF+ Signet
Model Number:
3-2536-PO
YO-32500-00
More Details
Thermo Scientific Nalgene Oak Ridge round-bottom tubes; 85-mL; PC; 10/
Category: Centrifuge Tubes
Description: Thermo Scientific Nalgene Oak Ridge Round-Bottom Tubes
Brand:
Thermo Scientific Nalgene
Model Number:
3118-0085
YO-06328-70
More Details
Liquid Nitrogen withdrawal system (D050-8C00)
Category: Taylor Wharton Cryogenic Storage Dewars
Description: Taylor-Wharton Liquid Nitrogen Withdrawal System
Brand:
Taylor-Wharton
Model Number:
D050-8C00
YO-03773-67
More Details
Cole-Parmer Self-Priming 316 SS Centrifugal Pump, High Suction Lift; 1 (YO-70620-22)
Category: Centrifugal Pumps that are Mechanically Coupled (have a Mechanical Seal)
Description: Cole-Parmer Self-Priming 316 SS Centrifugal Pumps
Brand:
Cole-Parmer
YO-70620-22
More Details
Hach Total Phosphorus(USEPA) Test Kit (27426-45)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
27426-45
YO-99574-18
More Details
Shimadzu UV-1800 UV/Visible Scanning Spectrophotometer; 115 VAC
Category: Visible/UV-Visible Spectroscopy
Description: Shimadzu UV-1800 UV/Visible Scanning Spectrophotometer
Brand:
Shimadzu
YO-83400-20
More Details
Immersion heater, flat, 8-3/4" L heated area, 500 watts, 115 VAC (335-230)
Category: 316 Stainless Steel Heaters
Description: Heat-O-Matic Immersion Heaters
Brand:
Heat-O-Matic
Model Number:
335-230
YO-03046-40
More Details
316 Stainless Steel Tubing, high pressure; 1/16" OD; two 3-ft sections (YO-03300-02)
Category: Rigid Tubing
Description: 316 Stainless Steel Tubing
Brand:
Cole-Parmer
YO-03300-02
More Details
Accumet Benchtop Meter Replacement pH/ATC Probe
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Accumet
Model Number:
CP-620-530
YO-59500-81
More Details
Thermo Scientific NanoDrop™ ND-2000 1-position Spectrophotometer
Category: Thermo Scientific NanoDrop Spectrophotometers
Description: Thermo Scientific NanoDrop 2000 & 2000c Spectrophotometers
Brand:
Thermo Scientific NanoDrop
Model Number:
ND-2000
YO-83061-00
More Details
Thermo Scientific SnakeSkin Pleated Dialysis Tubing, 10,000 MWCO, 22 m
Category: Dialysis Membranes and Tubing
Description: Thermo Scientific SnakeSkin Pleated Dialysis Tubes
Brand:
Thermo Scientific
Model Number:
SnakeSkin Pleated Dialysis Tubes
YO-02904-10
More Details
64114
results
<<<Previous
1
2
3
4
5
6
7
8
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Horiba U-52 Multiparameter Meter to Thermo Scientific Snakeskin Pleated Dialysis Tubing
>
31125329-p-toc_2 31125329ptoc2