Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 5

Thermo Scientific Nalgene Cryogenic Vial   to   Fluke 922 Air Flow

Image
  Product Detail
SKU

Thermo Scientific Nalgene Cryogenic vial; 2.0 mL, 25/pack

Category: Cryogenic Vials and Caps
Description: Thermo Scientific Nalgene External Thread Cryogenic Vials
Brand: Thermo Scientific Nalgene
Model Number: 5000-0020


Thermo Scientific Finntip 10 mL Pipette Tips, 2 to 10 mL, 100/bag

Category: Thermo Scientific Finntip Pipette Tips
Description: Thermo Scientific Finntip Pipette Tips (2000 to 10,000 uL)
Brand: Thermo Scientific Finntip
Model Number: 9402-151


Thermo Scientific Nalgene Heavy-Duty Vacuum Carboy, 10 L

Category: Round Carboys
Description: Thermo Scientific Nalgene Heavy-Duty Vacuum Carboys
Brand: Thermo Scientific Nalgene
Model Number: 2226-0020


Threaded adapter, stainless steel, 1/8" female NPT to 1/8" male BSPT (YO-31830-40)

Category: BSP Adapters
Description: British Standard Pipe Tapered Threaded (BSPT) Adapters
Brand: Parker Hannifin


Thermo Scientific Nalgene Heavy-Duty Vacuum Carboy, 20 L

Category: Round Carboys
Description: Thermo Scientific Nalgene Heavy-Duty Vacuum Carboys
Brand: Thermo Scientific Nalgene
Model Number: 2226-0050


Thermo Scientific Nunc™ Sterile Permanox™ Microscope Slides 100/ (YO-01839-08)

Category: Slides/Cover Slips
Description: Thermo Scientific Nunc Sterile Microscope Slides
Brand: Thermo Scientific Nunc


Rigid PVC Tubing 3/8"ID x 5/8"OD - 8/ft

Category: Flexible Tubing
Description: Flexible PVC Tubing
Brand: Cole-Parmer


Gilmont micrometer syringe, 2 mL

Category: Micrometer Burettes and Syringes
Description: Gilmont Micrometer Syringes
Brand: Gilmont
Model Number: GS-1200


Tubing adapter, 1/4" OD compression fitting x 1/4-28 thread, 2/pk (R412-5K-PAIR)

Category: General Accessories
Description: Replacement Accessories
Brand: Lab Products
Model Number: R412-5K-PAIR


LaMotte Sanitizer Strength Chlorine Test Strips

Category: Water Quality Test Strips
Description: LaMotte Sanitizer Strength Test Strips
Brand: LaMotte
Model Number: 4250-BJ


NFPA Hazardous Materials Classification, Sign Only, 14" X 10", adhesiv

Category: Safety Signs & Labels
Description: Right-To-Know NFPA Hazardous Materials Classification Signs and Kits
Brand: Accuform
Model Number: ZFD878VS


Barbed fittings, NPT male pipe adapter, Kynar, 1/4" NPT x 1/4" ID, 3/3 (YO-30704-11)

Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand: Cole-Parmer


Compression pipe adapter, 3/8" NPT x 3/8", Brass, each (YO-31412-36)

Category: Male Threaded Adapters
Description: Brass Compression to NPT(M) Adapters
Brand: Cole-Parmer


Thermo Scientific Nalgene Replacement, polypropylene spigot

Category: Bottle Accessories
Description: Thermo Scientific Nalgene Quick-Action Spigots
Brand: Thermo Scientific Nalgene
Model Number: 6422-0010


Taylor Digital Thermometer with Programmable Temperature Alarms for Mo

Category: Digital Thermometers - General Purpose
Description: Taylor Dual Zone Monitoring Thermometer
Brand: Taylor
Model Number: 1442


Tygon Silicone Tubing, 1/8"ID x 1/4"OD, 50 ft/pack

Category: Tygon Tubing
Description: Tygon Silicone Tubing
Brand: Tygon


Pressure Transducer, I to P;3-15 PSI Output, 4-20 mA Input (961-070-000)

Category: Pressure I/P Transducers
Description: Marsh Bellofram Type 1000 I/P Transducers
Brand: Marsh Bellofram
Model Number: 961-070-000


Volumetric Pipettes, Class A pipettes, 5mL, pack of 12 (PF8170AF)

Category: Volumetric Pipettes
Description: Class A Volumetric Pipettes
Brand: Cole-Parmer
Model Number: PF8170AF


Labels for 1 to 2 mL centrifuges, cryo vials, Eppendorf, PCR tubes - r

Category: Labels and Label Printers
Description: Brady Labelmark 5 Laboratory Labeling Software
Brand: Brady


Solarus Digital Burette 50 mL (9392050)

Category: Electronic Burettes
Description: Hirschmann Solarus Electronic Burettes
Brand: Hirschmann
Model Number: 9392050


Barbed Fittings; NPT Male Pipe Adapter Elbow, Nylon, 1 1/4" NPT(M) x 1 (YO-31408-22)

Category: Barbed to threaded adapters
Description: Barbed to male NPT threaded elbow adapters
Brand: Cole-Parmer


Schedule 40 PVC piping, 1.25", 9' (YO-33312-22)

Category: Piping
Description: Schedule 40 PVC Piping
Brand: Cole-Parmer


Barbed Fittings; NPT Male Pipe Adapter, Nylon, 1 1/2" NPT(M) x 1 1/2" (YO-31408-16)

Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand: Cole-Parmer


Pump, Hand-Operated Rotary Drum Pump, Stainless Steel, 10 GPM, 40" int (219)

Category: Hand-Operated Drum Pumps
Description: Rotary Drum Pumps
Brand: Cole-Parmer
Model Number: 219


Pipette F1 0.5-5 mL (4641110)

Category: Thermo Scientific Finnpipette Pipettors
Description: Thermo Scientific Finnpipette F1 Single-channel Pipettors
Brand: Thermo Scientific Finnpipette
Model Number: 4641110


Vacuum/pressure diaphragm pump with gauges, regulators, and relief val (DOA-P504-BN)

Category: Dry Vacuum Pressure Pumps
Description: High-Capacity Vacuum/Pressure Pumps
Brand: Gast
Model Number: DOA-P504-BN


Ansell Thermaprene™ Heat-Resistant, Lined Neoprene Gloves, size 8,

Category: Temperature-Resistant Gloves
Description: Ansell Thermaprene Heat-Resistant, Lined Neoprene Gloves
Brand: Thermaprene
Model Number: 19-026 214022


Hach 2100Q Handheld Turbidity Meter, EPA compliant

Category: Turbidity Handheld Meters
Description: Hach 2100Q Handheld Turbidity Meter
Brand: Hach
Model Number: 2100Q Portable Turbidity Meter


Thermo Scientific Orion Conductivity Standard 1413 uS, 5 X 60 ml (011007)

Category: Conductivity Calibration Solutions
Description: Thermo Scientific Orion Conductivity Calibration Solutions
Brand: Thermo Scientific Orion
Model Number: 011007


Fluke 922 Air Flow / Digital Manometer (FLUKE-922/KIT)

Category: Manometers
Description: Fluke 922 Airflow Meter
Brand: Fluke
Model Number: FLUKE-922/KIT


64114 results


Thermo Scientific Nalgene Cryogenic Vial to Fluke 922 Air Flow

>