Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 80
Brushless Dc Magnetic Drive Pump
to
Protective Rubber Boot for Wika Stainless Steel Digital Pressure Gauge
Image
Product Detail
SKU
Brushless DC Magnetic Drive Pump, Polypropylene, 8 GPM, 24 VDC
Category: Centrifugal Pumps that are Magnetically Coupled (have a Magnetic Seal)
Description: Brushless DC Magnetic Drive Centrifugal Pumps
Brand:
Cole-Parmer
YO-72020-45
More Details
Chempette digital repetitive dispenser, 2-10 mL (A3-21DG-10)
Category: Repetitive Dispensers/Pipettors
Description: Cole-Parmer Chempette Digital Repetitive Dispensers
Brand:
Chempette
Model Number:
A3-21DG-10
YO-07848-40
More Details
Dwyer Magnehelic Differential Pressure Gauge, 0 to 10 Inh2O
Description: Dwyer Magnehelic Differential Pressure Gauges
Brand:
Dwyer
Model Number:
2010
YO-68462-68
More Details
Buffer capsules, pH 7.0, green (273,PH7)
Category: Buffers and Solutions for pH/Ion Measurement
Description: pH Buffer Capsules
Brand:
Metrepak
Model Number:
273,PH7
YO-05977-14
More Details
Male luer lock plug, PP, 25/pk (YO-45503-56)
Category: Polypropylene Luer Fittings and Adapters
Description: Polypropylene Luer-to-Luer Connectors and Luer Plugs
Brand:
Cole-Parmer
YO-45503-56
More Details
Non-Spill Quick Disconnect, 1/2" Male Pipe Thread (NPT), Polypropylene (NS6D24008)
Category: Quick-Disconnect Fittings
Description: Non-Spill Quick Disconnect Couplings
Brand:
Colder
Model Number:
NS6D24008
YO-31603-36
More Details
Compact/OEM bellows pressure pump, 0.5 LPM 5.8 psi, 3 VDC
Category: Compact/OEM Vacuum Pumps
Description: Miniature OEM Vacuum/Pressure Pumps
Brand:
Cole-Parmer
Model Number:
KPMS08A-3A
YO-79600-06
More Details
Advantec Pressure Filtration Holder, for 142 mm filter; 304 Stainless
Description: Advantec Pressure Filtration Holders
Brand:
Cole-Parmer
YO-02927-50
More Details
Barbed Y-Connector, Stainless Steel, 3/8
Category: Barbed to barbed connectors
Description: Barbed Y connectors
Brand:
Cole-Parmer
YO-30620-24
More Details
Wall clock, Battery operated (M12AB)
Description: Analog Wall Clocks
Brand:
Cole-Parmer
Model Number:
M12AB
YO-08642-01
More Details
All-Plastic Quick Disconnect Barbed Inserts; PS, 1/2" Tube ID, 5/pack (MPX22839M)
Category: Plastic Quick-Disconnect Couplings (APC Series)
Description: Plastic Quick Disconnect Fittings, Barbed Inserts
Brand:
Colder
Model Number:
MPX22839M
YO-31311-73
More Details
Digital Test Gauge, 0 To 10,000 PSI
Category: Digital Pressure Gauges and Meters
Description: GE Druck Digital Test Gauge ~ with 0.05% accuracy!
Brand:
GE Sensing
Model Number:
DPI 104
YO-68975-14
More Details
Compression Fitting, Straight Union, 316 SS, 1/8" OD (YO-31406-01)
Category: Straight Union Connectors
Description: Stainless Steel Straight Compression Unions
Brand:
Cole-Parmer
YO-31406-01
More Details
Millipore Amicon Ultra Centrifugal Filters, 15ML 30K 24PK
Category: Centrifugal Devices and Filters
Description: Millipore Amicon Ultra Centrifugal Filters
Brand:
Millipore
YO-29969-84
More Details
Cole-Parmer Microcentrifuge Tube, 1.7mL, amber, non sterile, 500/pk
Category: Microcentrifuge Tubes
Description: Cole-Parmer Snap Cap Microcentrifuge tubes
Brand:
Cole-Parmer
Model Number:
MCT-175-X
YO-06333-68
More Details
PVC Stopcock with Flow Indication, 1/4" to 1/2" hose barb x 1/4" to 1/ (#1079002)
Category: Stopcocks
Description: Stopcocks with Flow Indicators
Brand:
Guaranteed In Stock Program
Model Number:
#1079002
YO-06225-56
More Details
Straight compression union, PFA, 4mm tubing OD (YO-31321-61)
Category: Straight Union Connectors
Description: PFA Straight Compression Unions
Brand:
Guaranteed In Stock Program
YO-31321-61
More Details
Inner chamber filling solution (900002)
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Lab Products
Model Number:
900002
YO-05730-39
More Details
ZoneMaster Surge Protectors 3-phase surge 120 (ZC120Y)
Category: Surge and Noise Suppressors, and Power Conditioners
Description: ZoneMaster Surge Protectors
Brand:
ZoneMaster
Model Number:
ZC120Y
YO-26874-22
More Details
Millipore Amicon Ultra Centrifugal Filters, 15ML 100K 24PK
Category: Centrifugal Devices and Filters
Description: Millipore Amicon Ultra Centrifugal Filters
Brand:
Millipore
YO-29969-88
More Details
Lightnin small batch mixer with two-speed motor, 115 VAC
Category: Lightnin Batch Industrial Mixers
Description: Lightnin Small Batch, Direct-Drive Industrial Mixers
Brand:
Lightnin
YO-04518-30
More Details
Fluke 87V True RMS Multimeter (FLUKE-87V)
Category: Fluke Digital Multimeters
Description: Fluke 87V Industrial True RMS Multimeter with Temperature
Brand:
Fluke
Model Number:
FLUKE-87V
YO-26087-02
More Details
Qsonica Sonabox™ Sound Enclosure
Category: Accessories for Sonicator Ultrasonic Processors
Description: Sonabox Sound Enclosure
Brand:
Qsonica
Model Number:
432B
YO-04711-99
More Details
Lightnin® 316 SS high-shear radial flow impeller (Lightnin® R- (YO-04560-56)
Category: Lightnin
®
316 Stainless Steel Impellers
Description: High-Shear Radial Flow Impellers (Lightnin
®
R-100)
Brand:
Lightnin
YO-04560-56
More Details
Honeywell conductivity probe, K=0.1
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Honeywell
Model Number:
04973-XX1-333-20-00-000
YO-56611-30
More Details
Masterflex L/S PTFE-tubing pump head
Category: Masterflex L/S Pump Heads
Description: L/S Rigid PTFE-Tubing Pump Head
Brand:
Masterflex
YO-77390-00
More Details
Dry Contact Relay Module With Power Supply
Description: General Accessories
Brand:
Davis
YO-68476-78
More Details
Low Flow Peristaltic Pump, Variable Speed, 3.3 - 10 RPM, 12 VDC, Manua
Category: Microfluidic Pumps
Description: Cole-Parmer Low-Flow Peristaltic Pumps
Brand:
Cole-Parmer
YO-73160-10
More Details
Pyrex Brand 9800 test tube; 14 mL, pack of 72
Category: Test Tubes
Description: Pyrex Brand 9800 Test Tubes
Brand:
Pyrex
Model Number:
9800-15
YO-34569-04
More Details
Protective Rubber Boot For Wika Stainless Steel Digital Pressure Gauge
Category: Digital Pressure Gauges and Meters
Description: WIKA Stainless Steel Digital Gauges
Brand:
Wika
Model Number:
DG-10
YO-68052-90
More Details
64114
results
<<<Previous
76
77
78
79
80
81
82
83
84
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
10
20
30
40
50
60
65
66
67
68
69
70
71
72
73
74
75
76
77
78
79
80
81
82
83
84
85
86
87
88
89
90
91
92
93
94
100
110
120
130
140
150
160
170
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Brushless Dc Magnetic Drive Pump to Protective Rubber Boot for Wika Stainless Steel Digital Pressure Gauge
>
31125329-p-toc_2 31125329ptoc2