Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 9
Labpette Discovery Pipette 20 - 200 Ul (P3940-200)
to
Rtd Probe with 4
Image
Product Detail
SKU
Labpette Discovery Pipette 20 - 200 uL (P3940-200)
Category: Single-Channel Pipettors
Description: Labpette Discovery Pipettes
Brand:
LABPETTE
Model Number:
P3940-200
YO-24601-08
More Details
Bunsen burner, Tirrill burner, natural gas (H-6220)
Category: Bunsen Burners
Description: Tirrill Burners
Brand:
Accuflame
Model Number:
H-6220
YO-36130-10
More Details
Thermo Scientific Nalgene Centrifuge Bottle; 500-mL; PPCO; 69.5 mm dia
Category: Centrifuge Bottles
Description: Thermo Scientific Nalgene 450- and 500-mL Centrifuge Bottles
Brand:
Thermo Scientific Nalgene
Model Number:
3120-9500
YO-06106-25
More Details
316 SS Female Reducer, 1/2" x 1/4" NPT(F) (YO-31410-14)
Category: Female to female reducers
Description: Female reducers, NPT(F) x NPT(F)
Brand:
Cole-Parmer
YO-31410-14
More Details
Low-cost Brix refractometer with ATC, 0 to 32% Brix
Category: Handheld Manual Refractometers
Description: Brix Refractometers
Brand:
Cole-Parmer
Model Number:
RHB-32ATC
YO-81150-08
More Details
Horiba D-51 Waterproof pH Meter
Category: pH/Ion Handheld Meters
Description: Horiba D-51 Waterproof pH Meter
Brand:
Horiba
Model Number:
D-51
YO-58702-20
More Details
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital 2.8 cu ft, 2 (23210606)
Category: Sterilizers
Description: Heidolph Tabletop Autoclaves
Brand:
Brinkmann
Model Number:
23210606
YO-10780-10
More Details
Female luer lock x 1/4" NPT, 316 SS, each (YO-31507-48)
Category: Metal Luer Fittings
Description: 316 SS Fittings
Brand:
Cole-Parmer
YO-31507-48
More Details
Thermocouple,Wire,TYPE-K,20-GAUGE, High Temp Glass Braid Insulation, 1 (K20-2-321* (1300F MAX)) (YO-08542-22)
Category: Wire - Bulk / Spooled Thermocouple Wire
Description: 20 Gauge Thermocouple Wire
Brand:
Cole-Parmer
Model Number:
K20-2-321* (1300F MAX)
YO-08542-22
More Details
Cole-Parmer Economical Glove Box - Small
Category: General Accessories
Description: Cole-Parmer Glove Boxes
Brand:
Cole-Parmer
YO-34788-00
More Details
Remote temperature/humidity monitor with alarm
Category: Humidity Indicators
Description: Thermohygrometer with Remote Probe
Brand:
Holiday Gift Ideas
Model Number:
7110CP
YO-90081-00
More Details
WHEATON BOD Bottle, 300 mL, glass, numbered 49 to 72, robotic stopper,
Category: BOD Bottles
Description: WHEATON BOD Glass Bottles
Brand:
Wheaton
Model Number:
227497-03
YO-03660-18
More Details
Eppendorf Microcentrifuge Tubes; capacity, 1.5-mL; 500/pack
Category: Microcentrifuge Tubes
Description: Eppendorf Flex-Tubes tubes
Brand:
Eppendorf
Model Number:
22 36 411-1
YO-02550-07
More Details
Wheaton TLC Reagent Sprayer Complete Sprayer, 125 mL (YO-34695-10)
Category: Thin Layer Chromatography (TLC) Accessories
Description: WHEATON TLC Reagent Sprayers and Replacement Parts
Brand:
Wheaton
YO-34695-10
More Details
3M BX™ Dual Reader Safety Glasses, 1.5x top and bottom diopters
Category: Safety Glasses
Description: 3M BX Dual Reader Safety Glasses
Brand:
3M
Model Number:
11457-00000-20
YO-86474-31
More Details
Volumetric Pipettes, Class A pipettes, 50mL (PF8170AS)
Category: Volumetric Pipettes
Description: Class A Volumetric Pipettes
Brand:
Cole-Parmer
Model Number:
PF8170AS
YO-25560-32
More Details
Cole-Parmerlog Watch Glass 50mm 12/pk (YO-34508-50)
Category: Watch Glasses
Description: Cole-Parmer Watch Glasses
Brand:
Cole-Parmer
YO-34508-50
More Details
Torbeo Hand-Held Homogenizer; capacity, 1 to 200 mL; stator size; 1.3 (YO-04751-00)
Category: Cole-Parmer Homogenizers
Description: Cole-Parmer Handheld Homogenizers
Brand:
Biospec Products
YO-04751-00
More Details
WTW ProfiLine pH 3310 meter kit with SenTix 41 pH electrode
Category: pH/Ion Handheld Meters
Description: WTW ProfiLine pH 3310 Handheld pH Meters
Brand:
WTW
Model Number:
pH 3310
YO-58890-22
More Details
Fluke 322 Clamp Meter (FLUKE-322)
Category: Fluke Clamp Meters
Description: Fluke 320 Series Clamp Meters
Brand:
Fluke
Model Number:
FLUKE-322
YO-26035-02
More Details
Injection Quill, PVC
Category: Pumps - Metering Accessories
Description: Injection Quills
Brand:
Cole-Parmer
Model Number:
QP-.5-3
YO-74601-06
More Details
WTW ProfiLine pH 3310 meter with Sentix 51 electrode
Category: pH/Ion Handheld Meters
Description: WTW ProfiLine pH 3310 Handheld pH Meters
Brand:
WTW
Model Number:
pH 3310
YO-58890-26
More Details
Pyrex Large Vol Burette 1000 ml (2094-1L)
Category: Glass and Plastic Burettes
Description: Pyrex Brand 2094 Large Volume Burettes
Brand:
Pyrex
Model Number:
2094-1L
YO-34542-34
More Details
Thermo Scientific Nalgene Cryogenic vial; 1.5 mL, 25/pack
Category: Cryogenic Vials and Caps
Description: Thermo Scientific Nalgene External Thread Cryogenic Vials
Brand:
Thermo Scientific Nalgene
Model Number:
5000-1020
YO-06754-94
More Details
Horiba D-55 pH/DO/ORP meter. (352735-1)
Category: Handheld Multiparameter Meters
Description: Horiba D-55 Portable Multiparameter Meter
Brand:
Horiba
Model Number:
352735-1
YO-99591-40
More Details
Thermo Scientific Nunc™ Square Culture Dish; Bioassay dish; 16/case (YO-01929-00)
Category: Tissue Culture
Description: Thermo Scientific Nunc Sterile Bioassay Dishes
Brand:
Thermo Scientific Nunc
YO-01929-00
More Details
Soil Pocket Penetrometer
Category: Soil Testing
Description: Pocket Penetrometer
Brand:
HUMBOLDT
Model Number:
H-4200
YO-99039-10
More Details
CHEMets Colorimetric Dissolved Oxygen Test Kit, 1 to 12 ppm (K-7512)
Category: Titrets and CHEMets Test Kits
Description: CHEMets Colorimetric Dissolved Oxygen Test Kits
Brand:
Titrets
Model Number:
K-7512
YO-05540-40
More Details
Barbed fittings, NPT male pipe adapter, Kynar, 1/16" NPT x 1/8" ID, 3/ (YO-30704-02)
Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30704-02
More Details
RTD Probe with 4 to 20 mA Output, 0 to 400°F, 6" immersion depth (TST-A2-060-PV-08)
Category: RTD Probes and Accessories
Description: Oakton 4 to 20 mA Output RTD probes
Brand:
Cole-Parmer
Model Number:
TST-A2-060-PV-08
YO-32884-02
More Details
64114
results
<<<Previous
5
6
7
8
9
10
11
12
13
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Labpette Discovery Pipette 20 - 200 Ul (P3940-200) to Rtd Probe with 4 to 20 Ma Output
>
31125329-p-toc_2 31125329ptoc2