Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 90
Bulkhead Fitting
to
Pipette F1 100-1000 Μl (4641100)
Image
Product Detail
SKU
Bulkhead Fitting, PTFE, 1/8" NPT(F) Port X 1-5/8" OD (F-TA01)
Category: Bulkhead unions
Description: PTFE Bulkhead Connectors
Brand:
PTFE
Model Number:
F-TA01
YO-34031-11
More Details
Oakton 3000 ppm salt singles, NaCl calibration packets
Category: Conductivity Calibration Solutions
Description: Conductivity Calibration Pouches
Brand:
Oakton
YO-35653-15
More Details
3M personal air monitor, organic vapors, 10/bx
Category: Personal Monitor Badges
Description: 3M Personal Monitor Badges
Brand:
3M
Model Number:
3500
YO-40116-00
More Details
Panel-mount hose barb (SMFPM01)
Category: Miniature Quick-Disconnect Fittings
Description: Panel-mount hose barbs (female fitting halves)
Brand:
Colder
Model Number:
SMFPM01
YO-06363-66
More Details
Bulkhead connectors NPT(F) x OD, LLDPE, 1-1/2" NPT x 2-5/8" OD, 1-3/4" (YO-06924-53)
Category: Bulkhead unions
Description: Bulkhead connectors
Brand:
Cole-Parmer
YO-06924-53
More Details
Rotary Evaporator with Cold-trap glassware, vacuum regulator, vapor te (YO-28623-25)
Category: Yamato Rotary Evaporator System
Description: Yamato Digital Rotary Evaporator Systems
Brand:
Yamato
YO-28623-25
More Details
304 SS dual-blade propeller for shaft 04370-02, 100 mm propeller dia (YO-04370-18)
Category: Laboratory Mixer Propellers
Description: Dual-Blade Propellers
Brand:
metric fitting
YO-04370-18
More Details
Thermo Scientific Barnstead EASYpure® II pretreatment replacement (D50230)
Category: Water Purification
Description: Water Systems Accessories
Brand:
EASYPure
Model Number:
D50230
YO-99261-50
More Details
Fluke-1555 Insulation Tester, 10 kV (FLUKE-1555)
Category: Resistance Testers
Description: Fluke 5 kV and 10 kV Insulation Resistance Testers
Brand:
Fluke
Model Number:
FLUKE-1555
YO-20036-95
More Details
Best Tuff Guard™ glove; size, extra-large; 12/pack
Category: Reusable Latex Gloves
Description: Best Tuff Guard glove
Brand:
Best Glove
Model Number:
574-10
YO-86288-63
More Details
WTW TetraCon 4-electrode conductivity probe, 10-m cable, waterproof DI (301972)
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
WTW
Model Number:
301972
YO-19707-82
More Details
B-D™ Disposable Syringes, Luer-Lock Tips, 30 mL (YO-07940-20)
Category: Syringes for Chormatography Applications
Description: BD Disposable Sterilized Syringes
Brand:
B-D
YO-07940-20
More Details
Burkert Type 3234 SS Diaphragm Valve, 3-Way, 3/4" x 3/4" x 3/4" Tri-Cl (YO-98517-06)
Category: Diaphragm Valves
Description: Burkert Type 3234 Stainless Steel Diaphragm Valves
Brand:
Burkert
YO-98517-06
More Details
Nipple, Sch 80, PTFE, 1/4" NPT(M) (F-N02C)
Category: Threaded nipples
Description: PTFE NPTM Nipples
Brand:
Cole-Parmer
Model Number:
F-N02C
YO-34030-02
More Details
Ascending Irreversible Low Temperature Indicator, 41°F (5°C) (YO-90308-10)
Category: Irreversible Temperature Labels / Indicators
Description: Irreversible Low-Temperature Indicators / Ascending Labels
Brand:
Cole-Parmer
YO-90308-10
More Details
Gilian® Gilibrator-2® Diagnostic Calibration System (800275)
Category: Air Sampling
Description: Gilian
®
Gilibrator-2
®
Diagnostic Calibration System
Brand:
Gilian
Model Number:
800275
YO-86276-96
More Details
Bulkhead Fitting, PVDF, 3/4" NPT(F) Port X 2-3/8" OD (FK-TA06)
Category: Bulkhead unions
Description: PVDF Bulkhead Connectors
Brand:
Cole-Parmer
Model Number:
FK-TA06
YO-34031-22
More Details
Safety Sign, Notice - Safety Glasses Required In This Area, 7" X 10", (MPPE854VA)
Category: Notice Signs
Description: Notice: Safety Glasses Required In This Area Signs
Brand:
Accuform
Model Number:
MPPE854VA
YO-60710-15
More Details
Oakton Waterproof pH/Con 10 Meter with pH/conductivity probe
Category: Handheld Multiparameter Meters
Description: Oakton Waterproof pH/Con 10 pH/Conductivity Meters
Brand:
Oakton
YO-35630-02
More Details
Class II A2 6-ft Biosafety Cabinet with 8" sash opening and fluorescen (YO-33515-80)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic Class II Biosafety Cabinets
Brand:
Labconco
YO-33515-80
More Details
Thermo Scientific Orion 2-Cell Conductivity Electrode, K=0.1, 5-ft cab
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Thermo Scientific
Model Number:
013016MD
YO-19704-74
More Details
Parker Balston H2 Generator for Fuel Gas; 260 mL/min
Category: Gas Generators
Description: Parker Balston's Hydrogen Generators for Fuel Gas
Brand:
FUEL
YO-86442-04
More Details
Push-to-connect Bulkhead Union Connector, PVDF, 1/4" Tube OD, Pack Of (F4BU4)
Category: PVDF Push-to-Connect Fittings
Description: Bulkhead Push-to-Connect Unions, PVDF
Brand:
Parker Hannifin
Model Number:
F4BU4
YO-34040-72
More Details
Open water grab sampler, 18' long
Category: Open Water Samplers
Description: Wheaton Open Water Grab Samplers
Brand:
Wheaton
Model Number:
990450
YO-99152-20
More Details
Parker Balston Zero Air Generator; 1 LPM; 220V/50Hz
Category: Gas Generators
Description: Parker Balston Zero Air Generators
Brand:
Cole-Parmer
YO-86442-17
More Details
Thermo Scientific Barnstead EASYpure II pretreatment replacement cartr
Category: Water Purification
Description: Water Systems Accessories
Brand:
EASYPure
Model Number:
D50231
YO-99261-54
More Details
Injection septa, pack of 10 (71-9507)
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Cole-Parmer
Model Number:
71-9507
YO-94101-30
More Details
Spring tubing clamp for up to 5/16"OD tubing, 3/bag (YO-06834-10)
Category: Tubing Clamps, and Accessories
Description: Flow Control Clamps - Spring Pinchcock
Brand:
Cole-Parmer
YO-06834-10
More Details
Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-24)
Category: Compression Fittings
Description: Metric size stainless steel compression fittings
Brand:
metric fitting
YO-31528-24
More Details
Pipette F1 100-1000 µL (4641100)
Category: Thermo Scientific Finnpipette Pipettors
Description: Thermo Scientific Finnpipette F1 Single-channel Pipettors
Brand:
Thermo Scientific Finnpipette
Model Number:
4641100
YO-25014-20
More Details
64114
results
<<<Previous
86
87
88
89
90
91
92
93
94
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
10
20
30
40
50
60
70
75
76
77
78
79
80
81
82
83
84
85
86
87
88
89
90
91
92
93
94
95
96
97
98
99
100
101
102
103
104
110
120
130
140
150
160
170
180
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Bulkhead Fitting to Pipette F1 100-1000 Μl (4641100)
>
31125329-p-toc_2 31125329ptoc2