Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 900
Tag
to
Vertical Condenser for Cole-Parmer Rotary Evaporator (Yo-28615-55)
Image
Product Detail
SKU
Tag, Danger Do Not Operate Locked Out My Life Is On The Line, 5 7/8" X (MLT611LCP)
Category: Lockout/Tagout Danger Tags
Description: Lock Out/Tag Out; Danger - Do Not Operate - Locked Out - My Life Is On The Line
Brand:
Accuform
Model Number:
MLT611LCP
YO-40503-31
More Details
Smith&Wesson 38 SPECIAL™ Safety Glasses, Black frame, Clear lens
Category: Smith&Wesson Safety Glasses
Description: Smith & Wesson 38 Special Safety Glasses
Brand:
Smith & Wesson
Model Number:
SW890CI
YO-86474-30
More Details
BUTYL BENZOATE 98% 250GR (YO-88272-44)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88272-44
More Details
Cole-Parmer StableTemp Dry Block Heaters with Heated Lid (Digital), 2 (949303HLCP)
Category: Cole-Parmer StableTemp Dry Block Heaters and Blocks
Description: Cole-Parmer StableTemp Dry Block Heaters with Heated Cover (Digital)
Brand:
Cole-Parmer
Model Number:
949303HLCP
YO-36600-70
More Details
IMIDAZOLE 99+% CRYSTALLI 250G (YO-88277-64)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88277-64
More Details
CoorsTek High-Form Crucible, High Alumina; 20ml
Category: High Alumina Crucibles
Description: CoorsTek High-Form Crucibles, High Alumina
Brand:
CoorsTek
YO-17804-03
More Details
Acetal manifold, 4 port 1/4" NPT(F) inlet, 1/8" NPT(F) outlet (YO-31524-06)
Category: Metal and Thermoplastic Manifolds
Description: Acetal manifolds
Brand:
Delrin
YO-31524-06
More Details
Ball valve, 2-way right angled, 1/4" NPT(F) (PVC-657-4F4F-E)
Category: PVC valves
Description: Two-way (right-angled)
Brand:
Cole-Parmer
Model Number:
PVC-657-4F4F-E
YO-01378-48
More Details
Utility Cart, with Three Tuffy Flat Shelves, Black
Category: Service Carts
Description: Specialty Carts
Brand:
LUXOR
YO-47600-50
More Details
STACO 3PN1010BV: Steel cased variable-voltage controller with built-in
Category: Variable AC Voltage Controllers
Description: STACO Steel cased variable-voltage controller with built-in voltmeter
Brand:
Staco
Model Number:
3PN1010BV
YO-01575-26
More Details
Valve-cartridges (YO-03217-92)
Category: Cole-Parmer Single and Multitube Flowmeter Systems
Description: Valve Cartridges
Brand:
Cole-Parmer
YO-03217-92
More Details
Combo Distance Meter (10300)
Category: Measuring Tools
Description: Electronic Distance Measuring Meters
Brand:
Holiday Gift Ideas
Model Number:
10300
YO-97241-10
More Details
Hd. Flat Edge Pocket Thermometer , With Potentiometer For Calibration (RR990)
Category: Pocket Thermometers Digital - General Purpose
Description: Heavy-Duty Recalibratable Digital Pocket Thermometers
Brand:
Cole-Parmer
Model Number:
RR990
YO-90090-05
More Details
Fixed-angle Rotors for 17304-series centrifuges (YO-17302-52)
Category: High-Speed Universal Centrifuges
Description: Rotors for 17304-series centrifuges
Brand:
Cole-Parmer
YO-17302-52
More Details
Chemfluor 367 Tubing, 230 psi at 73°F
Category: Rigid Tubing
Description: Chemfluor 367 Tubing
Brand:
Chemfluor
YO-95638-05
More Details
Service kit for model 07090-10 (400-17)
Category: Guzzler Diaphragm Hand Pumps
Description: Service Kits
Brand:
Guzzler
Model Number:
400-17
YO-07090-01
More Details
Charcoal filter for workstation 33705 (FR-2)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
FR-2
YO-33705-60
More Details
BioBasic Phenyl KAPPA Capillary Column - Length 100mm, 500µm ID, 5 (YO-01899-12)
Category: Thermo Scientific BioBasic HPLC Columns
Description: Thermo Scientific BioBasic KAPPA Capillary and Nanobore Columns
Brand:
Thermo Scientific BioBasic
YO-01899-12
More Details
Electrothermal Electromantle Extraction Heater - 6 positions, 500 mL, (EME6 0500/CEBX6)
Category: Extraction Apparatus
Description: Electrothermal Electromantle Extraction Heaters and Heater/Stirrers
Brand:
Electrothermal
Model Number:
EME6 0500/CEBX6
YO-03076-32
More Details
Reaction vessel with coupling, 12 oz
Category: Bottles
Description: Lab-Crest Pressure Reaction Vessels
Brand:
Lab-Crest
Model Number:
110-106-0012
YO-63400-16
More Details
Centrifugal Magnetic Drive Pump with Enclosed Motor, Moderate-Flow Pol (YO-72010-30)
Category: Centrifugal Pumps that are Magnetically Coupled (have a Magnetic Seal)
Description: Centrifugal Magnetic Drive Pumps with Enclosed Motor, Moderate-Flow Polypropylene
Brand:
Cole-Parmer
YO-72010-30
More Details
ExStikII 3-ft Extension Cable and Probe Weight
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
ExStik
Model Number:
EX010
YO-53026-26
More Details
Hot-Wire Thermo-Anemometer
Category: Hot-Wire Anemometers
Description: Davis Hot-Wire Thermoanemometer
Brand:
E Instrument Group, LLC
YO-30008-25
More Details
Gastight High-Performance Syringes; capacity, 5mL; gauge, 22; syringe (YO-07939-53)
Category: Hamilton Gastight High-Performance Syringes
Description: Hamilton Gastight High-Performance Syringes with Cemented Needle
Brand:
Hamilton
YO-07939-53
More Details
Threaded Union, PTFE, 1/2" NPT(F) (F-UN04)
Category: Threaded Fittings
Description: NPT(F) Unions
Brand:
Cole-Parmer
Model Number:
F-UN04
YO-34030-70
More Details
Antisiphon Kit for Cole-Parmer Syringe Pump, Infusion and Withdrawal d
Category: Syringes and Accessories for Syringe Pumps
Description: Cole-Parmer Touch Screen Syringe Pumps Accessories
Brand:
Cole-Parmer
Model Number:
788303
YO-74905-72
More Details
Ohaus Defender 3000 Washdown Industrial Scale, 30 Kg X 0.005 Kg, 230
Category: Ohaus Industrial Scales
Description: Ohaus Defender 3000 Washdown Industrial Scales
Brand:
Ohaus
Model Number:
D31P30BR-230VAC
YO-01006-72
More Details
Spectra/Mesh Screen Discs, Stainless Steel, 90 mm, 103 µm, 5/pk
Category: Spectra/Mesh Screen Discs and Sheets
Description: Spectra/Mesh Screen Discs and Sheets, Stainless Steel
Brand:
Spectra/Mesh
YO-06635-43
More Details
Quick-Disconnect Compression Elbow Insert; Straight-through, Delrin, 1 (PMC2104)
Category: Compression Elbow Inserts
Description: Straight-through Compression Elbow Inserts
Brand:
Colder
Model Number:
PMC2104
YO-06361-26
More Details
Vertical Condenser for Cole-Parmer Rotary Evaporator (YO-28615-55)
Category: Rotary Evaporator Systems
Description: Cole-Parmer Roto Evaporator Accessories
Brand:
Cole-Parmer
YO-28615-55
More Details
64114
results
<<<Previous
896
897
898
899
900
901
902
903
904
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
100
200
300
400
500
600
700
800
810
820
830
840
850
860
870
880
885
886
887
888
889
890
891
892
893
894
895
896
897
898
899
900
901
902
903
904
905
906
907
908
909
910
911
912
913
914
920
930
940
950
960
970
980
990
1,000
1,100
1,200
1,300
1,400
1,500
1,600
1,700
1,800
2,000
2,138
Tag to Vertical Condenser for Cole-Parmer Rotary Evaporator (Yo-28615-55)
>
31125329-p-toc_2 31125329ptoc2