Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 100
Polypropylene Felt Filter Bag
to
Meta Scrubs Bottom
Image
Product Detail
SKU
Polypropylene felt filter bag, 18-1/2"L, 1 µm (155384-03)
Category: Bag Filters for Polypropylene Bag Filter Housings
Description: Polypropylene Felt Filter Bags
Brand:
Cole-Parmer
Model Number:
155384-03
YO-01516-20
More Details
Polypropylene felt filter bag, 18-1/2"L, 25 µm (155390-03)
Category: Bag Filters for Polypropylene Bag Filter Housings
Description: Polypropylene Felt Filter Bags
Brand:
Cole-Parmer
Model Number:
155390-03
YO-01516-26
More Details
Size 1 nylon monofilament filter bag; 300 µm (255200-03)
Category: Polypropylene Adapter Heads
Description: Nylon monofilament filter bags for adapter heads
Brand:
Cole-Parmer
Model Number:
255200-03
YO-01520-08
More Details
Silicone O-ring 47mm (YO-02930-60)
Category: Advantec Stainless Steel Filter Holders
Description: Advantec Stainless Steel Syringe and In-Line Filter Holders Accessories and Replacement Parts
Brand:
Advantec
YO-02930-60
More Details
Straight glass adapter 1/2" to 1/2" (YO-03354-00)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
YO-03354-00
More Details
Closed Tube For IKA T-18 And T-25 Homogenizers, 50 mL (disposable) (YO-04720-99)
Category: Homogenizer Probes, and Accessories
Description: IKA T-25 Homogenizer Rotor-Stator Generators
Brand:
IKA
YO-04720-99
More Details
Sensor Cap for In-line Mounting, Red (YO-05614-27)
Category: General Accessories
Description: Replacement Accessories
Brand:
Cole-Parmer
YO-05614-27
More Details
Thermo Scientific Nalgene Narrow-Mouth Economy Bottle, PPCO, 30 mL, 12 (YO-06034-22)
Category: Thermo Scientific Nalgene Narrow-Mouth Bottles
Description: Thermo Scientific Nalgene Narrow-Mouth Economy Bottles, PPCO
Brand:
Thermo Scientific Nalgene
YO-06034-22
More Details
Thermo Scientific Nalgene graduated polymethylpentene cylinder, 250 mL
Category: Plastic Graduated Cylinders
Description: Thermo Scientific Nalgene Graduated Cylinders, PMP
Brand:
Thermo Scientific Nalgene
Model Number:
3665-0250
YO-06134-55
More Details
Quick-Disconnect NPT(M) Pipe Adapter; Valved, 1/8 in. NPT(M), 1/8 in. (PMCD100212)
Category: NPT (M) Threaded Adapter Bodies
Description: Valved NPT (M) Threaded Adapter Bodies
Brand:
Colder
Model Number:
PMCD100212
YO-06359-80
More Details
Quick-Disconnect Panel-Mount Compression Insert; Valved, 1/4" Tube OD, (PMCD4004)
Category: Panel-Mount Compression Inserts
Description: Valved Panel-Mount Compression Inserts
Brand:
Colder
Model Number:
PMCD4004
YO-06361-32
More Details
Quick-Disconnect Panel-Mount Hose Barb Fitting Body; Straight-through, (PLC16006)
Category: Panel-Mount Hose Barb Bodies
Description: Straight-through Panel-Mount Hose Barb Bodies
Brand:
Colder
Model Number:
PLC16006
YO-06361-48
More Details
Barbed fitting,Reducing Connector,1/16" x 1/8" Tube ID,Kynar (YO-06362-11)
Category: Barbed Fittings
Description: Antimicrobial Protected Fittings
Brand:
Cole-Parmer
YO-06362-11
More Details
Barbed fitting,Y-connector,3/16" Tube ID,HDPE (YO-06366-22)
Category: Barbed Fittings
Description: Antimicrobial Protected Fittings
Brand:
Cole-Parmer
YO-06366-22
More Details
John Guest Push-to-Connect Collet Cover, 5/16" Tube OD
Category: John Guest acetal push-to-connect fittings
Description: Collet covers for John Guest push-to-connect fittings
Brand:
John Guest
YO-06378-83
More Details
Straight compression union, PVDF, 3/4" tubing OD (YO-06381-95)
Category: Straight Union Connectors
Description: Straight unions
Brand:
Cole-Parmer
YO-06381-95
More Details
Compression male pipe adapter elbow, 1/8" x 1/8", Nylon, 10 Pack (YO-06382-47)
Category: Male Threaded Adapter Elbows
Description: Nylon Compression to NPT(M) Adapter Elbows
Brand:
Cole-Parmer
YO-06382-47
More Details
Steel Plated Beaker Tongs, 9"
Category: Tongs and Tweezers
Description: Beaker Tong, 9", Steel Plated
Brand:
Cole-Parmer
YO-06443-06
More Details
Polyolefin heat-shrink tubing, 0.125 (YO-06850-10)
Category: Heat-Shrink Tubing
Description: Polyolefin tubing
Brand:
Cole-Parmer
YO-06850-10
More Details
Mini Dehumidifier (26910)
Category: Desiccants and Accessories
Description: Mini Dehumidifier
Brand:
W A HAMMOND DRIERITE CO
Model Number:
26910
YO-07193-02
More Details
Precision Scissors, Straight Blade (11.023)
Category: Tongs and Tweezers
Description: Precision Scissors
Brand:
Cole-Parmer
Model Number:
11.023
YO-07292-10
More Details
Disposable Micropipettes, 15 µL, 100/pk (1-000-0150)
Category: Glass Pipettes
Description: Disposable Micropipettes
Brand:
Cole-Parmer
Model Number:
1-000-0150
YO-07950-25
More Details
Disposable Micropipettes, 75 µL, 100/pk (1-000-0750)
Category: Glass Pipettes
Description: Disposable Micropipettes
Brand:
Cole-Parmer
Model Number:
1-000-0750
YO-07950-45
More Details
Enviro-safe Pocket Thermometer, Closed Plastic Case, 0/220°F, 160m
Category: 100% Non-Hazardous Liquid Filled Glass Thermometers
Description: H-B Instrument Standard Enviro-Safe Nonhazardous Pocket Glass Thermometers, Fahrenheit-scale
Brand:
H-B Instrument Company
Model Number:
20559
YO-08008-86
More Details
Quick-Set Epoxy Syringe (YO-08778-40)
Category: Adhesives and Epoxies
Description: Hardman Syringe Epoxies
Brand:
Hardman
YO-08778-40
More Details
3 3/4" Round Metal Mirror (YO-09914-16)
Category: Inspection / Pick-up Tools
Description: Inspection Tools
Brand:
General Tools and Instruments
YO-09914-16
More Details
META Scrubs Bottom, Hunter green color, Small
Category: Safety Apparel
Description: META Unisex Scrubs
Brand:
META
YO-10507-15
More Details
META Scrubs Bottom, Navy Blue color, XX-Large
Category: Safety Apparel
Description: META Unisex Scrubs
Brand:
META
YO-10507-29
More Details
META Scrubs Bottom, Ceil color, Small
Category: Safety Apparel
Description: META Unisex Scrubs
Brand:
META
YO-10507-35
More Details
META Scrubs Bottom, Ceil color, Large
Category: Safety Apparel
Description: META Unisex Scrubs
Brand:
META
YO-10507-37
More Details
64114
results
<<<Previous
96
97
98
99
100
101
102
103
104
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
10
20
30
40
50
60
70
80
85
86
87
88
89
90
91
92
93
94
95
96
97
98
99
100
101
102
103
104
105
106
107
108
109
110
111
112
113
114
120
130
140
150
160
170
180
190
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Polypropylene Felt Filter Bag to Meta Scrubs Bottom
>
31125329-p-toc_2 31125329ptoc2