Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Product Index Page 1031
Hf-Resistant
to
Thermo Scientific Nalgene 489 Lldpe Tubing
Image
Product Detail
SKU
HF-resistant, Cartridge-Style, Double Junction, pH Electode (YO-27113-32)
Category: Cole-Parmer ERP Modular pH and ORP Electrode Platform Systems
Description: Cole-Parmer Modular pH and ORP Electrodes
Brand:
Cole-Parmer
YO-27113-32
More Details
Analog 500 mV Track It™ Logger with display, 500 mV module and long
Category: Multifunction Dataloggers
Description: Track-It DC Voltage and Current Data Loggers
Brand:
Track It
YO-30003-06
More Details
Davis Instruments Weather Envoy, cabled version.
Category: Weather Transmitters
Description: Davis Instruments Weather Envoy
Brand:
Davis Instruments
YO-30003-61
More Details
Four-way stopcocks, nickel-plated brass with satin finish Female l (V5650003)
Category: Luer Stopcocks
Description: Nickel-Plated Brass Stopcocks
Brand:
Cole-Parmer
Model Number:
V5650003
YO-31607-08
More Details
Solar Radiation Sensor for VantagePro2 (wireless) (6450)
Category: Davis Vantage Pro2 Weather Stations
Description: Davis Instruments VantagePro2 Weather Station Add-On Sensors
Brand:
Davis Instruments
Model Number:
6450
YO-32245-46
More Details
Push-to-connect Bulkhead Union Connector, PVDF, 1/4" Tube OD, Pack Of (F4BU4)
Category: PVDF Push-to-Connect Fittings
Description: Bulkhead Push-to-Connect Unions, PVDF
Brand:
Parker Hannifin
Model Number:
F4BU4
YO-34040-72
More Details
Allihn condenser (YO-34510-03)
Category: Analytical Glassware
Description: Accelerated One-Step Modular Extractor/Concentrator
Brand:
One-Step
YO-34510-03
More Details
Kimax Wide-Mouth Volumetric Flask, 10mL, Glass stopper, 6/cs
Category: Glass Volumetric Flasks
Description: Kimax Wide-Mouth Volumetric Glass Flasks
Brand:
Kimax
YO-34558-62
More Details
Pyrex Brand 6220 funnel; 65 mm ID pack of 12
Category: Powder Funnels
Description: Pyrex Brand 6220 Funnels
Brand:
Pyrex
YO-34564-01
More Details
Oakton Waterproof pHTestr 10 Pocket pH Tester
Category: Oakton Waterproof pH Testr Pocket pH Testers
Description: Oakton Waterproof pHTestr 10 Pocket pH Tester
Brand:
Oakton
Model Number:
pHTestr 10
YO-35634-10
More Details
OXFORD Listeria selective supplement, 13 vials (YO-36152-52)
Category: Bacteria test kits
Description: Listeria Test Kits
Brand:
Bel-Art
YO-36152-52
More Details
Additional Single SS Shelf
Category: CO
2
Incubators
Description: Thermo Scientific Water-Jacketed
CO
2
Incubator
Brand:
Thermo Scientific
YO-39320-24
More Details
Mi3 Communication Box, USB
Category: Infrared Sensors
Description: Raytek Compact MI3-Series Industrial Infrared Temperature Sensors
Brand:
Raytek
Model Number:
RAYMID10LTCB3
YO-39661-24
More Details
Adjustable Side Access Rack for 2" box, 12 bxs/rack, for 13 cuft ExF a
Category: Freezer Accessories
Description: Thermo Scientific Revco Accessories for Upright and Chest Ultra-Low (-86°C) Temperature Freezers
Brand:
Thermo Scientific
YO-44146-56
More Details
10.5mm Chuck for Heidolph Mixers
Category: Laboratory Mixers
Description: Heidolph
®
Laboratory Mixers
Brand:
Heido
YO-54400-05
More Details
Delmhorst J-Lite Wood Moisture Meter 6-30%
Category: Wood / Paper Moisture Meters
Description: Delmhorst J-Series Moisture Meters for Wood
Brand:
Delmhorst
Model Number:
J-LITE
YO-59820-00
More Details
Transducer, 200 PSI, 4-20 mA
Category: Pressure Transmitters and Pressure Transducers
Description: Ashcroft G2 Pressure Transducers
Brand:
Ashcroft
Model Number:
G2200PSI
YO-68900-54
More Details
Vacuum Pump Fluid,sil,500cc/ml
Category: Oils and Fluids
Description: Cole-Parmer Vacuum Pump Oil
Brand:
Cole-Parmer
Model Number:
704BB
YO-79754-10
More Details
Adapter; 304 SS, ½" NPT(F)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
A-1716
YO-81005-62
More Details
Drum Tray Dolly
Category: Secondary Containment
Description: Eagle Drum Trays
Brand:
Cole-Parmer
Model Number:
1618
YO-86110-44
More Details
Justrite Translucent Liquid Disposal Can, 5 Gal.
Category: Safety Cans
Description: Justrite Translucent Liquid Disposal Cans
Brand:
Justrite
Model Number:
12754
YO-86395-24
More Details
Salisbury Pro-Wear™ Arc Flash Protective Overpants, 8 cal/cm2, Larg
Category: Arc Flash Apparel
Description: Pro-Wear Arc Flash Protective Overpants
Brand:
Salisbury
Model Number:
ACP830BL-L
YO-86436-41
More Details
2-ACETYL-6-METHOXYNAPHTHA 10GR (YO-88137-92)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88137-92
More Details
THIOUREA PA 1KG (YO-88235-35)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88235-35
More Details
CUMENE-D12 99 ATOM % D 1ML (YO-88276-78)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88276-78
More Details
2 3-DIFLUOROBENZOIC ACID 10G (YO-88285-49)
Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand:
Cole-Parmer
YO-88285-49
More Details
Temperature Switches: temperature trip point; 100F (AD148-12/100F)
Category: Controllers-Accessories
Description: Temperature Switches
Brand:
Cole-Parmer
Model Number:
AD148-12/100F
YO-93880-52
More Details
Tygon® high-purity tubing, 3/8"ID x 1/2"OD, 50 ft/pack (YO-95635-05)
Category: Tubing for Bioprocessing
Description: TYGON High-Purity Tubing
Brand:
Tygon
YO-95635-05
More Details
Tygon High Purity Tubing, 5/16"ID x 7/16"OD, 50 ft/pk
Category: Flexible Tubing
Description: Tygon High-Purity Tubing
Brand:
Tygon
YO-95635-54
More Details
Thermo Scientific Nalgene 489 LLDPE Tubing, 5/8 x 3/4", 100' per (YO-95912-06)
Category: Rigid Tubing
Description: Thermo Scientific Nalgene 489 LLDPE Tubing
Brand:
Thermo Scientific Nalgene
YO-95912-06
More Details
63806
results
<<<Previous
1027
1028
1029
1030
1031
1032
1033
1034
1035
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
100
200
300
400
500
600
700
800
930
940
950
960
970
980
990
1,000
1,010
1,016
1,017
1,018
1,019
1,020
1,021
1,022
1,023
1,024
1,025
1,026
1,027
1,028
1,029
1,030
1,031
1,032
1,033
1,034
1,035
1,036
1,037
1,038
1,039
1,040
1,041
1,042
1,043
1,044
1,045
1,050
1,060
1,070
1,080
1,090
1,100
1,110
1,120
1,130
1,200
1,300
1,400
1,500
1,600
1,700
1,800
1,900
2,000
2,127
Hf-Resistant to Thermo Scientific Nalgene 489 Lldpe Tubing
>
31125329-p-toc_2 31125329ptoc2