Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 400
Microbore Tubing Connector
to
Mounting Kit for Magnehelic Gauges
Image
Product Detail
SKU
Microbore tubing connector, reducer, 24 to 20 Gauge
Category: Micro-bore Tubing Connectors
Description: Microbore Tubing Reducing Connectors
Brand:
Cole-Parmer
YO-34001-13
More Details
Microbore tubing connector, reducer, 20 to 18 Gauge
Category: Micro-bore Tubing Connectors
Description: Microbore Tubing Reducing Connectors
Brand:
Cole-Parmer
YO-34001-14
More Details
Microbore tubing connector, reducer, 12 Gauge to 1/8"
Category: Micro-bore Tubing Connectors
Description: Microbore Tubing Reducing Connectors
Brand:
Cole-Parmer
YO-34001-18
More Details
Bulkhead Union, 5/32, 10 Per Case (YO-34006-59)
Category: John Guest acetal push-to-connect fittings
Description: John Guest acetal push-to-connect bulkhead union connectors
Brand:
John Guest
YO-34006-59
More Details
PYREX Brand 4980 flask; 500 mL, pack of 6
Category: Glass Erlenmeyer Flasks
Description: PYREX Brand 4980 Glass Flasks
Brand:
Pyrex
YO-34503-07
More Details
Testo Wireless Thermocouple Adapter (0554 0222)
Category: General Accessories
Description: Replacement Accessories
Brand:
Testo
Model Number:
0554 0222
YO-37486-03
More Details
Seeker Infrared Thermometer with Star Burst Laser Target (12:1)
Category: Professional Compact Infrared Thermometers
Description: General Tools Infrared Thermometer with Star Burst Laser
Brand:
General Tools
Model Number:
IRT657
YO-37803-74
More Details
Cole-Parmer Digital Thermohygrometer with Dew Point
Category: Cole-Parmer Humidity Indicators
Description: Cole-Parmer Thermohygrometer with Dew Point and Alarm
Brand:
Cole-Parmer
Model Number:
4800CP
YO-37803-83
More Details
Flir Cigarette Lighter Adapter Kit, 12vdc (1.2m Cable) (1910490)
Category: Thermal Imaging Windows and Accessories
Description: Flir Thermal Imager Accessories
Brand:
Flir
YO-39754-26
More Details
Tag, Danger Do Not Operate mAintenance Department, Back B, 5 7/8" X 3 (MDT206PTP)
Category: OSHA/ANSI Compliant Header Tags
Description: Accident Prevention Tags; Danger: Do Not Operate - Maintenance Department
Brand:
Accuform
Model Number:
MDT206PTP
YO-40503-55
More Details
Tag, Danger Out Of Service, Back A, 5 7/8" X 3 3/8", RV-Plastic (MDT158PTP)
Category: OSHA/ANSI Compliant Header Tags
Description: Accident Prevention Tags; Danger: Out Of Service
Brand:
Accuform
Model Number:
MDT158PTP
YO-40503-69
More Details
Tag, Danger, 5 7/8" X 3 3/8", RV-Plastic (MDT260PTP)
Category: OSHA/ANSI Compliant Header Tags
Description: Accident Prevention Tags; Danger: Blank (write in your own message)
Brand:
Accuform
Model Number:
MDT260PTP
YO-40503-97
More Details
Tag, Warning, 5 7/8" X 3 3/8", RV-Plastic (MWGT205PTP)
Category: OSHA/ANSI Compliant Header Tags
Description: Accident Prevention Tags; Warning: Blank (write your own message)
Brand:
Accuform
Model Number:
MWGT205PTP
YO-40503-99
More Details
Cartridge (YO-49004-87)
Category: Recorders
Description: Chart Recorder Paper and Pens
Brand:
Davis
YO-49004-87
More Details
Replacement PINS - 4IN
Category: Moisture Meter Accessories
Description: Delmhorst-Moisture Meter Replacement Pins
Brand:
Delmhorst
Model Number:
608/4
YO-59820-42
More Details
Zip-lip low-density polyethylene bag, 4" x 4"
Category: Sample Bags
Description: Zip-Lip Sample Bags
Brand:
Cole-Parmer
YO-60101-20
More Details
Teeny Tough-Tags™ label, white
Category: Microtube Labels
Description: Teeny Tough-Tags Label
Brand:
Tough-Tags
Model Number:
9188-8690
YO-62205-20
More Details
Multi-Language Wash Bottle, LDPE Set of 4, 250 mL 5/pk
Category: Wash Bottles
Description: Venting Multilingual-labeled Safety Wash Bottles, LDPE
Brand:
Cole-Parmer
YO-62300-89
More Details
Green neoprene stopper, size 5
Category: Stoppers
Description: Solid Laboratory Green Neoprene Stoppers
Brand:
Guaranteed In Stock Program
YO-62991-14
More Details
Green neoprene stopper, size 8
Category: Stoppers
Description: Solid Laboratory Green Neoprene Stoppers
Brand:
Guaranteed In Stock Program
YO-62991-26
More Details
Folding Skirt Stopper Silicone 10.2 Dia 50/pk
Category: Stoppers
Description: Folding Skirt Stoppers, Rubber or Silicone
Brand:
Lab Products
YO-62995-91
More Details
Cole-Parmer Graduated Cylinder Set of 7
Category: Graduated Cylinder Sets
Description: Cole-Parmer Cylinder Set
Brand:
Cole-Parmer
Model Number:
CPSET7
YO-63100-24
More Details
Rubbermaid Yellow LDPE Lid for Storage Container with Handles 63511-05
Category: Plastic Containers
Description: Rubbermaid Yellow LDPE Lids for Storage Containers with Handles
Brand:
Rubbermaid
YO-63511-54
More Details
0 to 3000 psi Industrial Gauge with Back Connection, 2 1/2" Dial
Category: Mechanical Pressure Gauges
Description: Wika Industrial Gauge
Brand:
Wika
Model Number:
213.53 2.5
YO-68046-38
More Details
SPAN Pressure Gauge, 0 to 160 psi (YO-68081-12)
Category: Mechanical Pressure Gauges
Description: Pressure Gauges for Petrochemical Applications
Brand:
Thuemling
YO-68081-12
More Details
SPAN Pressure Gauge, 0 to 200 psi (YO-68081-14)
Category: Mechanical Pressure Gauges
Description: Pressure Gauges for Petrochemical Applications
Brand:
Thuemling
YO-68081-14
More Details
Cooling tower for process gauges, spiral style, 1/4" NPT F/M
Category: Gauge guards, Seals, and Accessories
Description: Industrial Gauge Cooling Towers
Brand:
Dwyer
Model Number:
A-250-A
YO-68332-05
More Details
Pressure Gauge ~ 1/4" NPT(M) process connection; 0 to 10" Water Co
Category: Mechanical Pressure Gauges
Description: Cole-Parmer Low-Pressure Mechanical Gauge
Brand:
Cole-Parmer
Model Number:
KC25-10"H2O
YO-68357-00
More Details
Pressure Gauge ~ 1/4" NPT (M) process connection; 0 to 200" Water
Category: Mechanical Pressure Gauges
Description: Cole-Parmer Low-Pressure Mechanical Gauge
Brand:
Cole-Parmer
Model Number:
KC25-200"H2O
YO-68357-12
More Details
Mounting Kit for Magnehelic Gauges
Category: Dwyer Magnehelic Differential Pressure Gauges
Description: Dwyer Pressure Gauge Accessories
Brand:
Dwyer
Model Number:
A-610
YO-68462-63
More Details
64114
results
<<<Previous
396
397
398
399
400
401
402
403
404
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
100
200
300
310
320
330
340
350
360
370
380
385
386
387
388
389
390
391
392
393
394
395
396
397
398
399
400
401
402
403
404
405
406
407
408
409
410
411
412
413
414
420
430
440
450
460
470
480
490
500
600
700
800
900
1,000
1,100
1,200
1,300
2,000
2,138
Microbore Tubing Connector to Mounting Kit for Magnehelic Gauges
>
31125329-p-toc_2 31125329ptoc2