Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 600
Platform Head with Cover
to
Pcr Preparation Rack
Image
Product Detail
SKU
Platform head with cover, 3" dia, for Vortex Genie Shakers (OK-0500-902)
Category: Shaker Accessories
Description: Vortex-Genie Shaker Accessories
Brand:
Vortex-Genie
Model Number:
OK-0500-902
YO-04724-61
More Details
Universal microtube holder for use with Turbomix attachment
Category: Shaker Accessories
Description: Vortex-Genie Shaker Accessories
Brand:
Vortex-Genie
YO-04724-63
More Details
Additional Motor Support Rod for Cole-Parmer LabGEN Homogenizer 125 an
Category: Homogenizer Probes, and Accessories
Description: Cole-Parmer LabGEN Homogenizer Accessories
Brand:
Cole-Parmer
YO-04727-90
More Details
Standard, transparent, reusable, PVC bailer, 1.5" OD x 3' L (BPCS1.05X36)
Category: Reusable Bailers
Description: Standard PVC Bailers
Brand:
Cole-Parmer
Model Number:
BPCS1.05X36
YO-05484-15
More Details
Replacement turbidity cells, for 05559-00, -05; 3/pack (50051)
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Lab Products
Model Number:
50051
YO-05559-53
More Details
Thermo Scientific Nalgene™ Utility Basin, HDPE, 10 quart
Category: Utility Trays
Description: Thermo Scientific Nalgene Utility Basins, HDPE
Brand:
Thermo Scientific Nalgene
Model Number:
7120-0010
YO-06011-15
More Details
FDA-approved high-density polyethylene container, 5 gallon (HPB 4716)
Category: UN-Approved Carboys
Description: Five-Gallon Container
Brand:
Cole-Parmer
Model Number:
HPB 4716
YO-06060-10
More Details
Thermo Scientific Nalgene high-density polyethylene jerrican with hand (2240-0015)
Category: Square/Rectangular Carboys and Jerricans
Description: Jerricans
Brand:
Thermo Scientific Nalgene
Model Number:
2240-0015
YO-06069-10
More Details
Thermo Scientific Nalgene low-density polyethylene drop-dispenser bott (2411-0060)
Category: Plastic Dropping Bottles
Description: Thermo Scientific Nalgene Low-Density Polypropylene Drop-Dispenser Bottles
Brand:
Thermo Scientific Nalgene
Model Number:
2411-0060
YO-06086-30
More Details
Thermo Scientific Nalgene PP Two-Way Stopcock, 1/4" to 5/16" hose barb (6460-0002)
Category: Stopcocks
Description: Thermo Scientific Nalgene Two- and Three-Way Stopcocks with Hose Barb Connections
Brand:
Thermo Scientific Nalgene
Model Number:
6460-0002
YO-06222-20
More Details
Scienceware stainless steel vibrating spatula with PTFE coating
Category: Spatulas & Stir Rods
Description: Scienceware Vibrating Spatulas, 304 SS
Brand:
Scienceware
Model Number:
H36719-0000
YO-06264-55
More Details
NSF-Approved High-Heat Spatulas, PP/Rubber, 13-1/2"L (1934)
Category: Spatulas & Stir Rods
Description: Spatulas, PP/Rubber
Brand:
Cole-Parmer
Model Number:
1934
YO-06278-25
More Details
T-connector assortment pack, PP, 6 each of sizes 4mm to 14 mm, 36 piec (YO-06294-82)
Category: Fittings Assortment Packs and Kits
Description: Fittings Assortment Packs
Brand:
Cole-Parmer
YO-06294-82
More Details
Miniature Quick-Disconnect Coupling; PC; Straight-through, 1/16" ID, P (SMC0191)
Category: Miniature Quick-Disconnect Fittings
Description: Miniature Quick-Disconnect Fittings, Complete Couplings
Brand:
Colder
Model Number:
SMC0191
YO-06363-90
More Details
Peroxide-Cured Silicone Tubing, 3/8"ID x 1/2"OD, 25 ft/pack
Category: Silicone Tubing
Description: Peroxide-Cured Silicone Tubing, 25 ft/pack
Brand:
Cole-Parmer
YO-06411-12
More Details
Tygon Microbore Tubing, 0.040" x 0.070"OD, 100 ft/roll
Category: Sanitary General-Purpose Tubing
Description: Tygon Microbore Tubing
Brand:
Tygon
YO-06418-04
More Details
Masterflex® Tygon® Food (B 44-4X), L/S® 16, 50 ft. (YO-06419-16)
Category: L/S® Precision Tubing for Masterflex® L/S® Pumps
Description: L/S® Precision Tygon® Food (B-44-4X) Tubing for Masterflex® L/S® Pumps
Brand:
Masterflex
YO-06419-16
More Details
Clear C-FLEX Tubing, 1/4"ID x 7/16"OD, 25 ft/pack
Category: Flexible Tubing
Description: Clear C-FLEX Tubing
Brand:
C-FLEX
YO-06422-11
More Details
Clear C-FLEX Tubing, 3/8"ID x 1/2"OD, 25 ft/pack
Category: Flexible Tubing
Description: Clear C-FLEX Tubing
Brand:
C-FLEX
YO-06422-15
More Details
C-FLEX tubing, 1/16"ID x 1/8"OD, 25 ft/pack
Category: Flexible Tubing
Description: C-FLEX Tubing
Brand:
C-FLEX
YO-06424-62
More Details
C-FLEX tubing, 1/4"ID x 7/16"OD, 25 ft/pack
Category: Flexible Tubing
Description: C-FLEX Tubing
Brand:
C-FLEX
YO-06424-72
More Details
Microbore two-stop tube sets, Tygon; 0.25 mm ID. Pk of 12.
Category: L/S Multichannel Pump Systems
Description: Tygon E-LFL Two-Stop Precision Pump Tubing for Masterflex L/S Multichannel Pump Heads and Systems
Brand:
Masterflex
YO-06447-12
More Details
Microbore two-stop tube sets, Tygon; 0.89 mm ID. Pk of 12
Category: L/S Multichannel Pump Systems
Description: Tygon E-LFL Two-Stop Precision Pump Tubing for Masterflex L/S Multichannel Pump Heads and Systems
Brand:
Masterflex
YO-06447-26
More Details
Microbore two-stop tube sets, Tygon; 2.06 mm ID. Pk of 12.
Category: L/S Multichannel Pump Systems
Description: Tygon E-LFL Two-Stop Precision Pump Tubing for Masterflex L/S Multichannel Pump Heads and Systems
Brand:
Masterflex
YO-06447-42
More Details
Biohazard Autoclave Gloves, 13"L
Category: Temperature-Resistant Gloves
Description: Biohazard Autoclave Gloves
Brand:
Scienceware
Model Number:
13201
YO-06496-40
More Details
Zip-lip low-density polyethylene bag assortment pack; 100 each of 6010 (YO-06503-90)
Category: Sample Bags
Description: Zip-Lip Sample Bags
Brand:
Cole-Parmer
YO-06503-90
More Details
Masterflex Norprene high-pressure tubing, L/S 16HP, 25 ft.
Category: L/S High-Pressure Tubing for Masterflex L/S Pumps
Description: L/S High-Pressure Norprene Tubing for Masterflex L/S Pumps
Brand:
Masterflex
YO-06504-16
More Details
Glass fiber filter, GB100R, 55-mm dia; box of 100
Category: Filter Papers
Description: Advantec Glass and Quartz Fiber Filters
Brand:
Advantec
Model Number:
GB100R55MM
YO-06657-73
More Details
High-density polyethylene utility tray, 17 1/2" x 15 1/2" x 5" (P417-05-2510)
Category: Utility Trays
Description: Cole-Parmer Utility Trays, PP
Brand:
Cole-Parmer
Model Number:
P417-05-2510
YO-06710-30
More Details
PCR preparation rack, blue, 5/pk
Category: Tube Racks
Description: PCR Preparation and Storage Racks, PP
Brand:
Cole-Parmer
YO-06736-01
More Details
64114
results
<<<Previous
596
597
598
599
600
601
602
603
604
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
100
200
300
400
500
510
520
530
540
550
560
570
580
585
586
587
588
589
590
591
592
593
594
595
596
597
598
599
600
601
602
603
604
605
606
607
608
609
610
611
612
613
614
620
630
640
650
660
670
680
690
700
800
900
1,000
1,100
1,200
1,300
1,400
1,500
2,000
2,138
Platform Head with Cover to Pcr Preparation Rack
>
31125329-p-toc_2 31125329ptoc2