Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 7
Light Blue Tweezer
to
Quick Disconnect Air Plug
Image
Product Detail
SKU
Light Blue Tweezers, Style 3C (18056EZF)
Category: Tongs and Tweezers
Description: Color Coded Tweezers
Brand:
Cole-Parmer
Model Number:
18056EZF
YO-07291-02
More Details
Durac General Purpose Thermometer, -20/150°C, 300mm Length, Total
Category: Red Spirit Filled Glass Thermometers
Description: H-B Instrument Durac Red Spirit General Purpose Glass Thermometers, Celsius-Scale
Brand:
H-B Instrument Company
Model Number:
2/1150
YO-08008-21
More Details
Pad F/08057-30 8x8" (PADFOR2036)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
PADFOR2036
YO-08057-94
More Details
Female luer x 1/16" hose barb, smooth bore, Nylon, 25/pk (YO-45502-50)
Category: Nylon Luer Fittings and Adapters
Description: Nylon Barbed Luer Adapters
Brand:
Cole-Parmer
YO-45502-50
More Details
Male luer x 1/16" hose barb (for separate lock ring), Nylon, 25/pk (YO-45505-40)
Category: Nylon Luer Fittings and Adapters
Description: Nylon Barbed Luer Adapters
Brand:
Cole-Parmer
YO-45505-40
More Details
Male luer x 3/32" hose barb (for separate lock ring), Nylon, 25/pk (YO-45505-42)
Category: Nylon Luer Fittings and Adapters
Description: Nylon Barbed Luer Adapters
Brand:
Cole-Parmer
YO-45505-42
More Details
3/4" wide x 36 yards long - white
Category: Tapes
Description: Color-Coded Vinyl Adhesive Tape
Brand:
Cole-Parmer
Model Number:
VT-WHITE-3/4
YO-86209-30
More Details
3/4" wide x 36 yards long - yellow
Category: Tapes
Description: Color-Coded Vinyl Adhesive Tape
Brand:
Cole-Parmer
Model Number:
VT-YELLOW-3/4
YO-86209-31
More Details
3/4" wide x 36 yards long - orange
Category: Tapes
Description: Color-Coded Vinyl Adhesive Tape
Brand:
Cole-Parmer
Model Number:
VT-ORANGE-3/4
YO-86209-32
More Details
3/4" wide x 36 yards long - red
Category: Tapes
Description: Color-Coded Vinyl Adhesive Tape
Brand:
Cole-Parmer
Model Number:
VT-RED-3/4
YO-86209-33
More Details
3/4" wide x 36 yards long - green
Category: Tapes
Description: Color-Coded Vinyl Adhesive Tape
Brand:
Cole-Parmer
Model Number:
VT-GREEN-3/4
YO-86209-34
More Details
3/4" wide x 36 yards long - blue
Category: Tapes
Description: Color-Coded Vinyl Adhesive Tape
Brand:
Cole-Parmer
Model Number:
VT-BLUE-3/4
YO-86209-35
More Details
3/4" wide x 36 yards long - black
Category: Tapes
Description: Color-Coded Vinyl Adhesive Tape
Brand:
Cole-Parmer
Model Number:
VT-BLACK-3/4
YO-86209-37
More Details
For Interconnecti Logic Terminals (YO-94469-65)
Description: General Accessories
Brand:
Davis
YO-94469-65
More Details
Male Cam X 1-in NPT(F) Adapter, PP (QC-A100-PP)
Category: Standard polypropylene cam couplers and adapters
Description: Polypropylene male cam to female NPT threaded adapters
Brand:
Cole-Parmer
Model Number:
QC-A100-PP
YO-34020-12
More Details
Male Cam X 3/4-in NPT(M) Adapter, PP (QC-F075-PP)
Category: Standard polypropylene cam couplers and adapters
Description: Polypropylene male cam to male NPT threaded adapters
Brand:
Cole-Parmer
Model Number:
QC-F075-PP
YO-34020-02
More Details
Small pull-out tub, quantity 50
Category: Hand Cleansers
Description: Antimicrobial Hand Cleaning Wipes
Brand:
SAFETEC OF AMERICA
Model Number:
34405
YO-86106-12
More Details
Minigrabber Patch Cord, 24in Bk
Category: Test Leads and Probes
Description: Pomona Minigrabber Test Clip Patch Cord
Brand:
Pomona
Model Number:
3781
YO-23031-87
More Details
Minigrabber Patch Cord, 24in Red
Category: Test Leads and Probes
Description: Pomona Minigrabber Test Clip Patch Cord
Brand:
Pomona
Model Number:
3781
YO-23031-88
More Details
Size 4 high-flow polyester filter bag; 300°F (149°C) maximum (255101-03)
Category: High-Flow Filter Bags for High-Flow Filter Bag Housings
Description: High-Flow Polyster Felt-Filter Bags
Brand:
Cole-Parmer
Model Number:
255101-03
YO-01524-84
More Details
Nebulizer Inlet Tube, 150-mm length
Category: Flame Photometers
Description: Replacement Parts for Flame Photometers, Automatic Diluter, and Autosampler
Brand:
Cole-Parmer
YO-02655-08
More Details
Mono-Mold Standard Stir Bar, 5/8"L x 5/16" dia
Category: Stir Bars
Description: Mono-Mold Standard Stir Bars
Brand:
Mono-Mold
YO-04770-30
More Details
Shipping Carton for Collapsible Container 06100-30
Category: Plastic Containers
Description: Shipping Cartons for Collapsible Containers
Brand:
Guaranteed In Stock Program
Model Number:
CDC2668
YO-06100-03
More Details
Stackable high-density polyethylene pail, 1 gallon (1W 0627400)
Category: Buckets and Pails
Description: Gallon HDPE FDA and USDA Pails
Brand:
Cole-Parmer
Model Number:
1W 0627400
YO-06274-00
More Details
Quick-Disconnect Hose Barb Insert; Straight-through, 1/4 in. Tube ID, (PMC220412)
Category: Hose Barb Inserts
Description: Straight-through Hose Barb Inserts
Brand:
Colder
Model Number:
PMC220412
YO-06364-47
More Details
Compression union tee, PVDF, 1/8" (YO-06382-45)
Category: Compression Tee Connectors
Description: Union tees
Brand:
Cole-Parmer
YO-06382-45
More Details
Union elbows (YO-06383-50)
Category: Compression Union Elbow Connectors
Description: Union elbows
Brand:
Cole-Parmer
YO-06383-50
More Details
Compression to male threaded adapter, PVDF, 3/8" OD x 3/8" NPT(M) (YO-06385-85)
Category: Male Threaded Adapters
Description: Male pipe adapters (straight)
Brand:
Cole-Parmer
YO-06385-85
More Details
Adapter for 06433-60, fits Rieke Sr covers (ADPT-C01)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
ADPT-C01
YO-06433-62
More Details
Quick Disconnect Air Plug, 3/8 in NPT(F) (MP-3/8F)
Category: Accessories for Compressors - Air Equipment
Description: Quick-Disconnect Couplings for Air Lines
Brand:
Cole-Parmer
Model Number:
MP-3/8F
YO-07123-43
More Details
64114
results
<<<Previous
3
4
5
6
7
8
9
10
11
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Light Blue Tweezer to Quick Disconnect Air Plug
>
31125329-p-toc_2 31125329ptoc2