Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 70
Gel Wrap™ Gasket for Cole-Parmer Dual Vertical Mini-Gel System Mode (Yo-28575-88)
to
Quick-Disconnect Fitting
Image
Product Detail
SKU
Gel Wrap™ Gasket for Cole-Parmer Dual Vertical Mini-Gel System Mode (YO-28575-88)
Category: Cole-Parmer Dual Vertical Mini-Gel Systems
Description: Cole-Parmer Dual Vertical Mini-Gel Gel Wrap
Brand:
Cole-Parmer
YO-28575-88
More Details
Sanitary Gasket, Epdm, 1", 10/pk (YO-30548-62)
Category: Gaskets for Sanitary Clamp Fittings
Description: Cole-Parmer Gaskets for Sanitary Lines
Brand:
Cole-Parmer
YO-30548-62
More Details
Barbed fittings, NPT male tees, Natural PP, 1" NPT x 1" ID, 25/32", (YO-30610-89)
Category: Barbed to male NPT threaded branch T connectors
Description: Natural polypropylene barbed to male NPT threaded branch T connectors
Brand:
Cole-Parmer
YO-30610-89
More Details
Masterflex TEE-Connector, polypropylene, L/S 13-14, 10/pack
Category: Masterflex Barbed Fittings
Description: Masterflex Tee connectors
Brand:
Masterflex
YO-30613-43
More Details
Masterflex barbed reducing union, PVDF, L/S 16 x 3/32" tubing ID, 10/p
Description: Masterflex Barbed Fittings
Brand:
Masterflex
YO-30616-08
More Details
Masterflex Polyethylene RED.,L/S18/35/36,I/P73/70,10/P
Category: Masterflex Barbed Fittings
Description: Masterflex Straight and reducing Connectors
Brand:
Masterflex
YO-30616-17
More Details
Masterflex barbed reducing union, nylon, I/P 89 x 1/2" tubing ID, 10/p
Description: Masterflex Barbed Fittings
Brand:
Masterflex
YO-30616-26
More Details
Masterflex Polypropylene REDUCER,L/S 17/24 & I/P26,10/P
Category: Masterflex Barbed Fittings
Description: Masterflex Straight and reducing Connectors
Brand:
Masterflex
YO-30616-55
More Details
Masterflex Polypropylene RED.,L/S18/35/36,I/P73/70,10/P
Category: Masterflex Barbed Fittings
Description: Masterflex Straight and reducing Connectors
Brand:
Masterflex
YO-30616-59
More Details
MasterflexPolypropyleneREDUCER,I/P 82/88 & BT 88,10/P
Description: Masterflex Barbed Fittings
Brand:
Masterflex
YO-30616-63
More Details
Male Pipe Adapter, Nylon, 1/4" x 5/32", 10 PACK (YO-30621-40)
Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30621-40
More Details
Male Pipe Adapter, Nylon, 1/4" x 1/2", 10 PACK (YO-30621-55)
Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30621-55
More Details
Male Pipe Adapter, Nylon, 3/8" x 5/16", 10 PACK (YO-30621-60)
Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30621-60
More Details
Straight Connector, Nylon, 3/4", 10 PACK (YO-30621-82)
Category: Barbed to barbed connectors
Description: Straight barbed connectors
Brand:
Cole-Parmer
YO-30621-82
More Details
Reducing Connector, Nylon, 5/8" x 1/2", 10 PACK (YO-30622-04)
Category: Barbed to barbed connectors
Description: Straight barbed reducing connectors
Brand:
Cole-Parmer
YO-30622-04
More Details
Reducing Connector, Nylon, 1/4" x 3/16", 10 PACK (YO-30622-30)
Category: Barbed to barbed connectors
Description: Straight barbed reducing connectors
Brand:
Cole-Parmer
YO-30622-30
More Details
Reducing Connector, Polyethylene, 1/2" x 3/8", 10 PACK (YO-30622-42)
Category: Barbed to barbed connectors
Description: Straight barbed reducing connectors
Brand:
Cole-Parmer
YO-30622-42
More Details
Straight Connector, Nylon, 5/32", 10 PACK (YO-30622-51)
Category: Barbed to barbed connectors
Description: Straight barbed connectors
Brand:
Cole-Parmer
YO-30622-51
More Details
Elbow, Nylon, 5/8", 10 PACK (YO-30622-66)
Category: Barbed to barbed connectors
Description: Barbed elbow connectors
Brand:
Cole-Parmer
YO-30622-66
More Details
Nipple, Polyethylene, 1" x , 10 PACK (YO-30623-22)
Category: Threaded nipples
Description: Nipples, NPT(M)
Brand:
Cole-Parmer
YO-30623-22
More Details
Tee, Nylon, 5/8", 10 PACK (YO-30623-59)
Category: Barbed to barbed connectors
Description: Barbed T connectors
Brand:
Cole-Parmer
YO-30623-59
More Details
Tee, Polyethylene, 1", 10 PACK (YO-30623-62)
Category: Barbed to barbed connectors
Description: Barbed T connectors
Brand:
Cole-Parmer
YO-30623-62
More Details
Tee, Nylon, 1", 10 PACK (YO-30623-63)
Category: Barbed to barbed connectors
Description: Barbed T connectors
Brand:
Cole-Parmer
YO-30623-63
More Details
Tee, Nylon, 1/2", 10 PACK (YO-30623-80)
Category: Barbed to barbed connectors
Description: Barbed T connectors
Brand:
Cole-Parmer
YO-30623-80
More Details
Male pipe branch Tee, Nylon, 3/4" x 1/4", 10 PACKS (YO-30623-87)
Category: Barbed to threaded adapters
Description: Barbed to male NPT threaded branch T connectors
Brand:
Cole-Parmer
YO-30623-87
More Details
Male pipe branch Tee, Nylon, 3/4" x 1/2", 10 PACKS (YO-30623-93)
Category: Barbed to threaded adapters
Description: Barbed to male NPT threaded branch T connectors
Brand:
Cole-Parmer
YO-30623-93
More Details
Male pipe branch Tee, Nylon, 3/8" x 1/2", 10 PACKS (YO-30624-17)
Category: Barbed to threaded adapters
Description: Barbed to male NPT threaded branch T connectors
Brand:
Cole-Parmer
YO-30624-17
More Details
Cole-Parmer Animal Free Female Luer Fittings, PP, 25/pk
Category: Polycarbonate Luer Fittings and Adapters
Description: Cole-Parmer Animal Free Luer Fittings
Brand:
Cole-Parmer
YO-30800-12
More Details
Threaded adapter, brass, 3/8" NPTM x 1/4" tubing ID, 5/pack (YO-30904-20)
Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30904-20
More Details
Quick-disconnect fittings, Standard John Guest straight-through fittin (PLC29006)
Category: Standard John Guest Inserts
Description: Straight-through fittings
Brand:
Delrin
Model Number:
PLC29006
YO-31304-22
More Details
64114
results
<<<Previous
66
67
68
69
70
71
72
73
74
Next >>>
View by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Page:
1
10
20
30
40
50
55
56
57
58
59
60
61
62
63
64
65
66
67
68
69
70
71
72
73
74
75
76
77
78
79
80
81
82
83
84
90
100
110
120
130
140
150
160
200
300
400
500
600
700
800
900
1,000
2,000
2,138
Gel Wrap™ Gasket for Cole-Parmer Dual Vertical Mini-Gel System Mode (Yo-28575-88) to Quick-Disconnect Fitting
>
31125329-p-toc_2 31125329ptoc2