|
|
CAS# | Chemical Name | Percent | EINECS/ELINCS |
93-08-3 | 2'-Acetonaphthone | 99 | 202-216-2 |
|
Appearance: white powder.
Caution! Harmful. Causes eye and skin irritation.
May be harmful if swallowed. Causes digestive and respiratory tract
irritation. The toxicological properties of this material have not
been fully investigated.
Target Organs: No data found.
Potential Health Effects
Eye:
Causes eye irritation. May cause chemical conjunctivitis.
Skin:
Causes skin irritation.
Ingestion:
Causes gastrointestinal irritation with nausea, vomiting and
diarrhea. The toxicological properties of this substance have not
been fully investigated.
Inhalation:
The toxicological properties of this substance have not been fully
investigated. Causes irritation of the mucous membrane and upper
respiratory tract.
Chronic:
No information found.
|
|
|
|
|
Chemical Name | ACGIH | NIOSH | OSHA - Final PELs |
2'-Acetonaphthone | none listed | none listed | none listed |
|
|
|
|
|
|
US DOT | Canada TDG | |
Shipping Name: | Not regulated as a hazardous material | TOXIC LIQUIDS, ORGANIC, N.O.S. (Glutaraldehyde) |
Hazard Class: | 6.1 | |
UN Number: | UN2810 | |
Packing Group: | II |
|
|