Material Safety Data Sheet
Tetracosane, 99%
ACC# 37230
|
Section 1 - Chemical Product and Company Identification |
MSDS Name: Tetracosane, 99%
Catalog Numbers:
AC138040000, AC138040250, AC138041000
Synonyms:
None Known.
Company Identification:
Acros Organics N.V.
One Reagent Lane
Fair Lawn, NJ 07410
For information in North America, call: 800-ACROS-01
For emergencies in the US, call CHEMTREC: 800-424-9300
|
Section 2 - Composition, Information on Ingredients
|
| CAS# |
Chemical Name |
Percent |
EINECS/ELINCS |
| 646-31-1 |
Tetracosane |
99% | 211-474-5 |
|
Section 3 - Hazards Identification
|
EMERGENCY OVERVIEW
Appearance: white solid.
Caution! May cause eye and skin irritation. May
cause respiratory tract irritation. The toxicological properties of
this material have not been fully investigated.
Target Organs: No data found.
Potential Health Effects
Eye:
May cause eye irritation.
Skin:
May cause skin irritation.
Ingestion:
May cause irritation of the digestive tract. The toxicological
properties of this substance have not been fully investigated.
Inhalation:
May cause respiratory tract irritation. The toxicological properties
of this substance have not been fully investigated.
Chronic:
No information found.
|
Section 4 - First Aid Measures
|
Eyes:
Flush eyes with plenty of water for at least 15 minutes, occasionally
lifting the upper and lower eyelids. Get medical aid.
Skin:
Get medical aid. Flush skin with plenty of water for at least 15
minutes while removing contaminated clothing and shoes. Wash clothing
before reuse.
Ingestion:
Never give anything by mouth to an unconscious person. Get medical
aid. Do NOT induce vomiting. If conscious and alert, rinse mouth and
drink 2-4 cupfuls of milk or water.
Inhalation:
Remove from exposure and move to fresh air immediately. If not
breathing, give artificial respiration. If breathing is difficult,
give oxygen. Get medical aid.
Notes to Physician:
Treat symptomatically and supportively.
|
Section 5 - Fire Fighting Measures
|
General Information:
As in any fire, wear a self-contained breathing apparatus in
pressure-demand, MSHA/NIOSH (approved or equivalent), and full
protective gear. During a fire, irritating and highly toxic gases
may be generated by thermal decomposition or combustion. Runoff from
fire control or dilution water may cause pollution.
Extinguishing Media:
Use agent most appropriate to extinguish fire. Use water spray, dry
chemical, carbon dioxide, or appropriate foam.
Flash Point: > 110 deg C (> 230.00 deg F)
Autoignition Temperature: Not applicable.
Explosion Limits, Lower:Not available.
Upper: Not available.
NFPA Rating:
(estimated) Health: 1; Flammability: 1; Instability: 0
|
Section 6 - Accidental Release Measures
|
General Information: Use proper personal protective equipment as indicated
in Section 8.
Spills/Leaks:
Vacuum or sweep up material and place into a suitable disposal
container. Clean up spills immediately, observing precautions in the
Protective Equipment section. Avoid generating dusty conditions.
Provide ventilation.
|
Section 7 - Handling and Storage
|
Handling:
Wash thoroughly after handling. Remove contaminated clothing and
wash before reuse. Use with adequate ventilation. Minimize dust
generation and accumulation. Avoid contact with eyes, skin, and
clothing. Keep container tightly closed. Avoid ingestion and
inhalation.
Storage:
Store in a tightly closed container. Store in a cool, dry,
well-ventilated area away from incompatible substances.
|
Section 8 - Exposure Controls, Personal Protection
|
Engineering Controls:
Facilities storing or utilizing this material should be equipped
with an eyewash facility and a safety shower. Use adequate
ventilation to keep airborne concentrations low.
Exposure Limits
| Chemical Name |
ACGIH |
NIOSH |
OSHA - Final PELs |
| Tetracosane
|
none listed
|
none listed
|
none listed
|
OSHA Vacated PELs:
Tetracosane:
No OSHA Vacated PELs are listed for this chemical.
Personal Protective Equipment
Eyes:
Wear appropriate protective eyeglasses or chemical
safety goggles as described by OSHA's eye and face
protection regulations in 29 CFR 1910.133 or European
Standard EN166.
Skin:
Wear appropriate protective gloves to prevent skin
exposure.
Clothing:
Wear appropriate protective clothing to prevent skin
exposure.
Respirators:
Follow the OSHA respirator regulations found in 29
CFR 1910.134 or European Standard EN 149. Use a
NIOSH/MSHA or European Standard EN 149 approved
respirator if exposure limits are exceeded or if
irritation or other symptoms are experienced.
|
Section 9 - Physical and Chemical Properties
|
Physical State: Solid
Appearance: white
Odor: None reported.
pH: Not available.
Vapor Pressure: Not available.
Vapor Density: Not available.
Evaporation Rate:Not available.
Viscosity: Not available.
Boiling Point: 391 deg C @ 760.00mmHg
Freezing/Melting Point:49.00 - 52.00 deg C
Decomposition Temperature:Not available.
Solubility: insoluble
Specific Gravity/Density:Not available.
Molecular Formula:C24H50
Molecular Weight:338.65
|
Section 10 - Stability and Reactivity
|
Chemical Stability:
Stable under normal temperatures and pressures.
Conditions to Avoid:
Incompatible materials, dust generation, excess heat, strong
oxidants.
Incompatibilities with Other Materials:
Oxidizing agents.
Hazardous Decomposition Products:
Carbon monoxide, irritating and toxic fumes and gases, carbon
dioxide.
Hazardous Polymerization: Has not been reported.
|
Section 11 - Toxicological Information
|
RTECS#:
CAS# 646-31-1
unlisted.
LD50/LC50:
Not available.
Carcinogenicity:
CAS# 646-31-1:
Not listed by ACGIH, IARC, NTP, or CA Prop 65.
Epidemiology:
No information found
Teratogenicity:
No information found
Reproductive Effects:
No information found
Mutagenicity:
No information found
Neurotoxicity:
No information found
Other Studies:
|
Section 12 - Ecological Information
|
No information available.
|
Section 13 - Disposal Considerations
|
Chemical waste generators must determine whether a discarded chemical is classified
as a hazardous waste.
US EPA guidelines for the classification determination are listed in 40 CFR Parts 261.3.
Additionally, waste generators must consult state and local hazardous waste regulations to
ensure complete and accurate classification.
RCRA P-Series: None listed.
RCRA U-Series: None listed.
|
Section 14 - Transport Information
|
| |
US DOT |
Canada TDG |
| Shipping Name: |
Not regulated as a hazardous material |
No information available. |
| Hazard Class: |
|
|
| UN Number: |
|
|
| Packing Group: |
|
|
|
Section 15 - Regulatory Information
|
US FEDERAL
TSCA
CAS# 646-31-1 is listed on the TSCA inventory.
Health & Safety Reporting List
None of the chemicals are on the Health & Safety Reporting List.
Chemical Test Rules
None of the chemicals in this product are under a Chemical Test Rule.
Section 12b
None of the chemicals are listed under TSCA Section 12b.
TSCA Significant New Use Rule
None of the chemicals in this material have a SNUR under TSCA.
CERCLA Hazardous Substances and corresponding RQs
None of the chemicals in this material have an RQ.
SARA Section 302 Extremely Hazardous Substances
None of the chemicals in this product have a TPQ.
Section 313
No chemicals are reportable under Section 313.
Clean Air Act:
This material does not contain any hazardous air pollutants.
This material does not contain any Class 1 Ozone depletors.
This material does not contain any Class 2 Ozone depletors.
Clean Water Act:
None of the chemicals in this product
are listed as Hazardous Substances under the CWA.
None of the chemicals in this
product are listed as Priority Pollutants under the CWA.
None of the chemicals in this
product are listed as Toxic Pollutants under the CWA.
OSHA:
None of the chemicals in this product
are considered highly hazardous by OSHA.
STATE
CAS# 646-31-1 is not present on
state lists from CA, PA, MN, MA, FL, or NJ.
California Prop 65
California No Significant Risk Level:
None of the chemicals in this product are listed.
European/International Regulations
European Labeling in Accordance with EC Directives
Hazard Symbols:
Not available.
Risk Phrases:
Safety Phrases:
S 24/25 Avoid contact with skin and eyes.
S 37 Wear suitable gloves.
S 45 In case of accident or if you feel unwell, seek medical advice
immediately (show the label where possible).
S 28A After contact with skin, wash immediately with plenty of water
.
WGK (Water Danger/Protection)
CAS# 646-31-1: No information available.
Canada - DSL/NDSL
CAS# 646-31-1 is listed on Canada's DSL List.
Canada - WHMIS
WHMIS: Not available.
This product has been classified in accordance with the hazard
criteria of the Controlled Products Regulations and the MSDS
contains all of the information required by those regulations.
Canadian Ingredient Disclosure List
|
Section 16 - Additional Information
|
MSDS Creation Date: 9/02/1997
Revision #5 Date: 10/03/2005
The information above is believed to be accurate and represents the best
information currently available to us. However, we make no warranty of
merchantability or any other warranty, express or implied, with respect to
such information, and we assume no liability resulting from its use. Users
should make their own investigations to determine the suitability of the
information for their particular purposes. In no event shall Fisher be liable
for any claims, losses, or damages of any third party or for lost profits
or any special, indirect, incidental, consequential or exemplary
damages, howsoever arising, even if Fisher has been advised of
the possibility of such damages.