Material Safety Data Sheet
Tween 80

ACC# 40200
Section 1 - Chemical Product and Company Identification

MSDS Name: Tween 80
Catalog Numbers: BP338-500, NC9011213, NC9067414, NC9252526, NC9631327, NC9659759, NC9826024, NC9974612, T164-500, T164500LC, XXT164537GM, XXT164ET500M, XXT164ET500ML, XXT164SHK4LI, XXT164TESTIN, XXTWN80FHK4LI, XXTWN80NF4LI
Synonyms:
Polyoxyethylene 20 sorbitan monooleate; tween 81; tween 80A; BW800003
Company Identification:

              Fisher Scientific
              1 Reagent Lane
              Fair Lawn, NJ 07410
For information, call:
201-796-7100
Emergency Number:
201-796-7100
For CHEMTREC assistance, call:
800-424-9300
For International CHEMTREC assistance, call:
703-527-3887

Section 2 - Composition, Information on Ingredients

CAS# Chemical Name Percent EINECS/ELINCS
9005-65-6 Polyoxyethylene 20 sorbitan monooleate 100 unlisted

Section 3 - Hazards Identification

EMERGENCY OVERVIEW

Appearance: yellow amber liquid.
Caution! May cause eye, skin, and respiratory tract irritation. This is expected to be a low hazard for usual industrial handling.
Target Organs: None.


Potential Health Effects
Eye: May cause mild eye irritation.
Skin: May cause skin irritation.
Ingestion: May cause gastrointestinal irritation with nausea, vomiting and diarrhea. Low hazard for usual industrial handling.
Inhalation: May cause respiratory tract irritation. Low hazard for usual industrial handling.
Chronic: Adverse reproductive effects have been reported in animals.

Section 4 - First Aid Measures


Eyes: Flush eyes with plenty of water for at least 15 minutes, occasionally lifting the upper and lower eyelids. Get medical aid.
Skin: Flush skin with plenty of water for at least 15 minutes while removing contaminated clothing and shoes. Get medical aid if irritation develops or persists. Wash clothing before reuse.
Ingestion: If victim is conscious and alert, give 2-4 cupfuls of milk or water. Never give anything by mouth to an unconscious person. Get medical aid if irritation or symptoms occur.
Inhalation: Remove from exposure and move to fresh air immediately. If not breathing, give artificial respiration. If breathing is difficult, give oxygen. Get medical aid if cough or other symptoms appear.
Notes to Physician: Treat symptomatically and supportively.

Section 5 - Fire Fighting Measures


General Information: As in any fire, wear a self-contained breathing apparatus in pressure-demand, MSHA/NIOSH (approved or equivalent), and full protective gear. During a fire, irritating and highly toxic gases may be generated by thermal decomposition or combustion. Use water spray to keep fire-exposed containers cool. Vapors may be heavier than air. They can spread along the ground and collect in low or confined areas. Containers may explode when heated.
Extinguishing Media: Use agent most appropriate to extinguish fire. Cool containers with flooding quantities of water until well after fire is out. Use water spray, dry chemical, carbon dioxide, or appropriate foam.
Flash Point: 148 deg C ( 298.40 deg F)
Autoignition Temperature: Not applicable.
Explosion Limits, Lower:Not available.
Upper: Not available.
NFPA Rating: (estimated) Health: 1; Flammability: 1; Instability: 0

Section 6 - Accidental Release Measures


General Information: Use proper personal protective equipment as indicated in Section 8.
Spills/Leaks: Absorb spill with inert material (e.g. vermiculite, sand or earth), then place in suitable container. Clean up spills immediately, observing precautions in the Protective Equipment section. Provide ventilation.

Section 7 - Handling and Storage


Handling: Wash thoroughly after handling. Wash hands before eating. Avoid contact with eyes, skin, and clothing. Keep container tightly closed. Avoid ingestion and inhalation. Use with adequate ventilation.
Storage: Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances. Do NOT freeze.

Section 8 - Exposure Controls, Personal Protection


Engineering Controls: Use adequate ventilation to keep airborne concentrations low.
Exposure Limits
Chemical Name ACGIH NIOSH OSHA - Final PELs
Polyoxyethylene 20 sorbitan monooleate none listed none listed none listed

OSHA Vacated PELs: Polyoxyethylene 20 sorbitan monooleate: No OSHA Vacated PELs are listed for this chemical.
Personal Protective Equipment
Eyes: Wear appropriate protective eyeglasses or chemical safety goggles as described by OSHA's eye and face protection regulations in 29 CFR 1910.133 or European Standard EN166.
Skin: Wear appropriate protective gloves to prevent skin exposure.
Clothing: Wear appropriate protective clothing to prevent skin exposure.
Respirators: Follow the OSHA respirator regulations found in 29 CFR 1910.134 or European Standard EN 149. Use a NIOSH/MSHA or European Standard EN 149 approved respirator if exposure limits are exceeded or if irritation or other symptoms are experienced.

Section 9 - Physical and Chemical Properties


Physical State: Liquid
Appearance: yellow amber
Odor: characteristic odor
pH: 5-7 for 5% solution.
Vapor Pressure: < 1.0 mm Hg @20
Vapor Density: >1.0
Evaporation Rate:>1 (ether=1)
Viscosity: Not available.
Boiling Point: 100 deg C
Freezing/Melting Point:Not available.
Decomposition Temperature:Not available.
Solubility: Soluble in water.
Specific Gravity/Density: 1.08
Molecular Formula:Not applicable.
Molecular Weight:Not available.

Section 10 - Stability and Reactivity


Chemical Stability: Stable under normal temperatures and pressures.
Conditions to Avoid: Incompatible materials, excess heat.
Incompatibilities with Other Materials: Oxidizing agents.
Hazardous Decomposition Products: Carbon monoxide, irritating and toxic fumes and gases, carbon dioxide.
Hazardous Polymerization: Has not been reported.

Section 11 - Toxicological Information


RTECS#:      
CAS# 9005-65-6: WG2932500
LD50/LC50:
CAS# 9005-65-6:
     Draize test, rabbit, eye: 150 mg Mild;
     Oral, mouse: LD50 = 25 gm/kg;
     Oral, rat: LD50 = 34500 uL/kg;
.

Carcinogenicity:
CAS# 9005-65-6: Not listed by ACGIH, IARC, NTP, or CA Prop 65.

Epidemiology: No information available.
Teratogenicity: No information available.
Reproductive Effects: May cause reproductive effects based on animal studies.
Mutagenicity: No information available.
Neurotoxicity: No information available.
Other Studies:

Section 12 - Ecological Information


Ecotoxicity: No data available. No information available.
Environmental: No information found.
Physical: No information found.
Other: No information available.

Section 13 - Disposal Considerations

Chemical waste generators must determine whether a discarded chemical is classified as a hazardous waste. US EPA guidelines for the classification determination are listed in 40 CFR Parts 261.3. Additionally, waste generators must consult state and local hazardous waste regulations to ensure complete and accurate classification.
RCRA P-Series: None listed.
RCRA U-Series: None listed.

Section 14 - Transport Information

US DOT Canada TDG
Shipping Name: Not regulated as a hazardous material No information available.
Hazard Class:
UN Number:
Packing Group:

Section 15 - Regulatory Information

US FEDERAL

TSCA
     CAS# 9005-65-6 is listed on the TSCA inventory.
Health & Safety Reporting List
     None of the chemicals are on the Health & Safety Reporting List.
Chemical Test Rules
     None of the chemicals in this product are under a Chemical Test Rule.
Section 12b
     None of the chemicals are listed under TSCA Section 12b.
TSCA Significant New Use Rule
     None of the chemicals in this material have a SNUR under TSCA.
CERCLA Hazardous Substances and corresponding RQs
     None of the chemicals in this material have an RQ.
SARA Section 302 Extremely Hazardous Substances
     None of the chemicals in this product have a TPQ.
Section 313      No chemicals are reportable under Section 313.
Clean Air Act:
     This material does not contain any hazardous air pollutants.
     This material does not contain any Class 1 Ozone depletors.
     This material does not contain any Class 2 Ozone depletors.
Clean Water Act:
     None of the chemicals in this product are listed as Hazardous Substances under the CWA.
     None of the chemicals in this product are listed as Priority Pollutants under the CWA.
     None of the chemicals in this product are listed as Toxic Pollutants under the CWA.
OSHA:
     None of the chemicals in this product are considered highly hazardous by OSHA.
STATE
     CAS# 9005-65-6 is not present on state lists from CA, PA, MN, MA, FL, or NJ.

California Prop 65

California No Significant Risk Level: None of the chemicals in this product are listed.

European/International Regulations

European Labeling in Accordance with EC Directives
Hazard Symbols:
      Not available.
Risk Phrases:


Safety Phrases:


WGK (Water Danger/Protection)

     CAS# 9005-65-6: 1
Canada - DSL/NDSL
     CAS# 9005-65-6 is listed on Canada's DSL List.
Canada - WHMIS
     This product has a WHMIS classification of D2B.
This product has been classified in accordance with the hazard criteria of the Controlled Products Regulations and the MSDS contains all of the information required by those regulations.
Canadian Ingredient Disclosure List

Section 16 - Additional Information

MSDS Creation Date: 7/15/1999
Revision #6 Date: 5/31/2005

The information above is believed to be accurate and represents the best information currently available to us. However, we make no warranty of merchantability or any other warranty, express or implied, with respect to such information, and we assume no liability resulting from its use. Users should make their own investigations to determine the suitability of the information for their particular purposes. In no event shall Fisher be liable for any claims, losses, or damages of any third party or for lost profits or any special, indirect, incidental, consequential or exemplary damages, howsoever arising, even if Fisher has been advised of the possibility of such damages.