Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
More Unique Lab & Industrial Supplies
Conductivity/Resistivity/TDS Meters and Testers
Conductivity/Resistivity/TDS Meters and Testers
Cole-Parmer offers a wide range of conductivity meters and conductivity controllers for a variety of environments and applications.
Common features from portable to benchtop
Our low cost pocket conductivity testers and portable conductivity meters are ideal for field based conductivity measurement. Many models have an onboard memory to store and recall conductivity readings taken. For laboratory based applications, select from benchtop conductivity meters including leading brands such as Oakton, YSI, and Thermo Orion. Features of this style of conductivity meter can include a computer interface either by RS-232 or USB. Speak to our Applications Specialists for free technical support on conductivity measurement to help solve your application needs.
How conductivity measurements are made
When considering features for a conductivity meter it is important to first understand how conductivity measurements are made. Conductivity is the ability of a material to conduct electric current. Determination of conductivity is achieved by the instrument by placing two plates in the sample, applying a potential across the plates and measuring the current that passes through the solution. Conductivity, the opposite of resistivity, is determined from the voltage and current values according to Ohm’s Law.
Conductivity Meter Selection Tips
  • Autoranging: Meter automatically selects the most accurate range for measurement. There is no need to manually change the range.
  • TDS Conversion Factor: When a solution does not have a similar ionic water content to natural water or salt water, then a TDS conversion factor is needed to automatically adjust the TDS readings
  • Temperature Compensation: A cell with built-in temperature sensor allows the meter to make adjustments to the conductivity or TDS reading based on changes in solution temperature
  • Adjustable temperature coefficients: Alcohols and pure water are affected by changes in temperature differently than typical samples. An adjustable temperature coefficient allows the user to compensate for the properties of the measured solution
  • Adjustable cell constant: Adjusts the measurement to reflect use of a cell with a constant other than K = 1.0. Wide range meters may except cells between K = 0.01 and 10
Need
Assistance?
Get help from
our Experts!
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price
Oakton EcoTestr TDS Low pocket TDS tester
  • Product Type: TDS Meter
  • Meter style: Tester
  • Range: 0 to 1990 ppm TDS
EW-35462-10
In stock.
$62.25 USD/EACH
$49.99 USD/EACH
Qty:
Oakton ECTestr 11 dual-range, pin-style pocket conductivity tester
  • Product Type: Conductivity Meter
  • Meter style: Tester
  • Range: 0 µS to 20.00 mS Conductivity
EW-35662-30
In stock.
$88.25 USD/EACH
$79.42 USD/EACH
Qty:
Oakton ECTestr 11+ multi-range, cup-style pocket conductivity tester
  • Product Type: Conductivity Meter
  • Meter style: Tester
  • Range: 0 µS to 20.00 mS Conductivity
EW-35662-35
In stock.
$120.00 USD/EACH
$108.00 USD/EACH
Qty:
Oakton CON 6+ handheld conductivity meter with probe
  • Product Type: Conductivity Meter
  • Meter style: Portable
  • Range: 0 µS to 200 mS Conductivity
EW-35604-00
In stock.
$360.00 USD/EACH
Qty:
Oakton CON 6+ handheld conductivity meter kit
  • Product Type: Conductivity Meter
  • Meter style: Portable
  • Range: 0 µS to 200 mS Conductivity
EW-35604-04
In stock.
$406.00 USD/EACH
Qty:
Oakton TDSTestr 11 dual-range, pin-style pocket TDS tester
  • Product Type: TDS Meter
  • Meter style: Tester
  • Range: 0 to 10.0 ppt TDS
EW-35662-10
In stock.
$87.25 USD/EACH
Qty:
Myron L 4PII Waterproof Conductivity/TDS/Resistivity Meter 19401-12
  • Product Type: Conductivity Meter
  • Meter style: Portable
  • Range: 0 µS to 200 mS Conductivity
EW-19401-12
In stock.
$835.00 USD/EACH
Qty:
Oakton TDSTestr11+ multi-range, cup-style pocket TDS tester
  • Product Type: TDS Meter
  • Meter style: Tester
  • Range: 0-2000 ppm Conductivity
EW-35662-15
In stock.
$118.00 USD/EACH
Qty:
Myron L EP-10, Analog Conductivity Meter 01489-35
  • Product Type: Conductivity Meter
  • Meter style: Portable
  • Conductivity accuracy: ±2% full-scale
EW-01489-35
In stock.
$422.00 USD/EACH
$337.60 USD/EACH
Qty:
Oakton EcoTestr EC High pocket conductivity tester
  • Product Type: Conductivity Meter
  • Meter style: Tester
  • Conductivity accuracy: ±1% from 0 to 9.9 mS, ±2% above 10 mS
EW-35462-35
In stock.
$63.25 USD/EACH
$49.99 USD/EACH
Qty:
Oakton EcoTestr EC Low pocket conductivity tester
  • Product Type: Conductivity Meter
  • Meter style: Tester
  • Conductivity accuracy: ±1% FS
EW-35462-30
In stock.
$63.25 USD/EACH
$49.99 USD/EACH
Qty:
Oakton EcoTestr Salt pocket salinity tester
  • Product Type: Salinity Meter
  • Meter style: Tester
  • Range: 0 to 10 ppt Salinity
EW-35462-50
In stock.
$72.50 USD/EACH
$56.99 USD/EACH
Qty:
Oakton Waterproof CON 150 Portable Meter with Probe
  • Product Type: Conductivity Meter
  • Meter style: Portable
  • Range: 0 to 200 mS Conductivity
EW-35607-32
In stock.
$503.00 USD/EACH
Qty:
Oakton Benchtop CON 700 Meter with Probe and Stand
  • Product Type: Conductivity Meter
  • Meter style: Benchtop
  • Conductivity accuracy: ±1% full scale
EW-35411-00
In stock.
$663.00 USD/EACH
$596.70 USD/EACH
Qty:
Oakton SaltTestr 11 pocket salinity tester
  • Product Type: Salinity Meter
  • Meter style: Tester
  • Range: 0 to 10 ppt Salinity
EW-35662-52
In stock.
$107.00 USD/EACH
$85.60 USD/EACH
Qty:
108 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#
#PRODUCTHEADER#
#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#